Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 7712780-7713306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
UniGene
Hs.75285
Gene Chromosome Location
Chr.10: 7703273-7749520 on Build GRCh38
Transcripts
NM_002216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6197075 c.452C>T p.T151M Chr.10: 7713270-7713270 on Build GRCh38
1965500 c.453G>A p.T151T Chr.10: 7713271-7713271 on Build GRCh38
4641379 c.386G>A p.R129K Chr.10: 7713204-7713204 on Build GRCh38
1965497 c.414C>T p.G138G Chr.10: 7713232-7713232 on Build GRCh38
7116397 c.402G>C p.E134D Chr.10: 7713220-7713220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765726132 Chr.10: 7712947-7712947 on Build GRCh38
rs764481949 Chr.10: 7713134-7713135 on Build GRCh38
rs555417936 Chr.10: 7713204-7713204 on Build GRCh38
rs749554375 Chr.10: 7713190-7713190 on Build GRCh38
rs753013136 Chr.10: 7712859-7712859 on Build GRCh38
rs752032119 Chr.10: 7713140-7713140 on Build GRCh38
rs143792235 Chr.10: 7713208-7713208 on Build GRCh38
rs999347529 Chr.10: 7712987-7712987 on Build GRCh38
rs942344905 Chr.10: 7713125-7713125 on Build GRCh38
rs777351046 Chr.10: 7712846-7712846 on Build GRCh38
rs557890208 Chr.10: 7713048-7713048 on Build GRCh38
rs917372210 Chr.10: 7712820-7712820 on Build GRCh38
rs763273800 Chr.10: 7712950-7712959 on Build GRCh38
rs1039036382 Chr.10: 7713121-7713122 on Build GRCh38
rs999798857 Chr.10: 7713229-7713229 on Build GRCh38
rs61834849 Chr.10: 7713122-7713122 on Build GRCh38
rs879071993 Chr.10: 7713126-7713126 on Build GRCh38
rs369929097 Chr.10: 7712947-7712957 on Build GRCh38
rs375021724 Chr.10: 7713019-7713019 on Build GRCh38
rs757713933 Chr.10: 7713134-7713134 on Build GRCh38
rs529268099 Chr.10: 7712849-7712849 on Build GRCh38
rs770447604 Chr.10: 7713232-7713232 on Build GRCh38
rs778587511 Chr.10: 7713198-7713198 on Build GRCh38
rs769425686 Chr.10: 7713258-7713258 on Build GRCh38
rs767867921 Chr.10: 7713211-7713211 on Build GRCh38
rs887800287 Chr.10: 7712809-7712809 on Build GRCh38
rs909947584 Chr.10: 7713042-7713042 on Build GRCh38
rs780234408 Chr.10: 7713174-7713174 on Build GRCh38
rs927854294 Chr.10: 7713016-7713016 on Build GRCh38
rs553496491 Chr.10: 7712959-7712959 on Build GRCh38
rs148172382 Chr.10: 7713263-7713263 on Build GRCh38
rs766572351 Chr.10: 7713285-7713285 on Build GRCh38
rs112895650 Chr.10: 7713097-7713097 on Build GRCh38
rs762282644 Chr.10: 7713266-7713266 on Build GRCh38
rs988547357 Chr.10: 7712925-7712925 on Build GRCh38
rs761225692 Chr.10: 7713274-7713274 on Build GRCh38
rs756678189 Chr.10: 7712876-7712876 on Build GRCh38
rs569527507 Chr.10: 7712939-7712939 on Build GRCh38
rs979802773 Chr.10: 7712961-7712961 on Build GRCh38
rs1030190109 Chr.10: 7713128-7713128 on Build GRCh38
rs1043890256 Chr.10: 7713126-7713126 on Build GRCh38
rs756313919 Chr.10: 7713170-7713170 on Build GRCh38
rs376866436 Chr.10: 7712992-7712992 on Build GRCh38
rs12411796 Chr.10: 7713046-7713046 on Build GRCh38
rs762073382 Chr.10: 7713198-7713199 on Build GRCh38
rs1024315599 Chr.10: 7713014-7713014 on Build GRCh38
rs200935424 Chr.10: 7713270-7713270 on Build GRCh38
rs941550897 Chr.10: 7713045-7713045 on Build GRCh38
rs7091826 Chr.10: 7712887-7712887 on Build GRCh38
rs897388704 Chr.10: 7712921-7712921 on Build GRCh38
rs766611100 Chr.10: 7712947-7712948 on Build GRCh38
rs918118783 Chr.10: 7713018-7713018 on Build GRCh38
rs968501433 Chr.10: 7713181-7713181 on Build GRCh38
rs1013517853 Chr.10: 7713282-7713282 on Build GRCh38
rs779990202 Chr.10: 7713028-7713028 on Build GRCh38
rs750866146 Chr.10: 7712998-7712999 on Build GRCh38
rs1050711310 Chr.10: 7712955-7712955 on Build GRCh38
rs960648165 Chr.10: 7712993-7712993 on Build GRCh38
rs141925309 Chr.10: 7713271-7713271 on Build GRCh38
rs1006601295 Chr.10: 7712969-7712969 on Build GRCh38
rs746925560 Chr.10: 7713230-7713230 on Build GRCh38
rs566244998 Chr.10: 7713060-7713060 on Build GRCh38
rs776076565 Chr.10: 7713234-7713234 on Build GRCh38
rs914307060 Chr.10: 7712930-7712930 on Build GRCh38
rs141590355 Chr.10: 7713191-7713191 on Build GRCh38
rs1006683878 Chr.10: 7713116-7713116 on Build GRCh38
rs889460644 Chr.10: 7713120-7713120 on Build GRCh38
rs568338043 Chr.10: 7712847-7712847 on Build GRCh38
rs577078012 Chr.10: 7713189-7713189 on Build GRCh38
rs934245917 Chr.10: 7713099-7713099 on Build GRCh38
rs903700711 Chr.10: 7712986-7712986 on Build GRCh38
rs1021844393 Chr.10: 7713145-7713145 on Build GRCh38
rs955863952 Chr.10: 7712899-7712899 on Build GRCh38
rs386740611 Chr.10: 7712948-7712959 on Build GRCh38
rs750932895 Chr.10: 7713138-7713138 on Build GRCh38
rs941509600 Chr.10: 7712886-7712886 on Build GRCh38
rs1051251542 Chr.10: 7713110-7713110 on Build GRCh38
rs11255301 Chr.10: 7713003-7713003 on Build GRCh38
rs1017993574 Chr.10: 7712839-7712839 on Build GRCh38
rs4497303 Chr.10: 7712805-7712805 on Build GRCh38
rs768578232 Chr.10: 7713249-7713249 on Build GRCh38
rs190963409 Chr.10: 7712813-7712813 on Build GRCh38
rs1021969577 Chr.10: 7712980-7712980 on Build GRCh38
rs777804600 Chr.10: 7713227-7713227 on Build GRCh38
rs964666841 Chr.10: 7712883-7712883 on Build GRCh38

View Full Product Details