Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 30016500-30017001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1462
Entrez Gene ID
57608
Gene Name
KIAA1462
Gene Aliases
JCAD
UniGene
Hs.533953
Gene Chromosome Location
Chr.10: 30012800-30069816 on Build GRCh38
Transcripts
NM_020848.2 XM_011519608.1 XM_011519609.2

SNP Detail

SNP ID Chromosome Location
rs999575943 Chr.10: 30016675-30016675 on Build GRCh38
rs950686046 Chr.10: 30016551-30016551 on Build GRCh38
rs983474306 Chr.10: 30016589-30016589 on Build GRCh38
rs989458879 Chr.10: 30016755-30016755 on Build GRCh38
rs1031054540 Chr.10: 30016713-30016713 on Build GRCh38
rs879151888 Chr.10: 30016732-30016732 on Build GRCh38
rs368927533 Chr.10: 30016917-30016917 on Build GRCh38
rs532180789 Chr.10: 30016948-30016948 on Build GRCh38
rs142765150 Chr.10: 30016546-30016546 on Build GRCh38
rs1012539622 Chr.10: 30016961-30016961 on Build GRCh38
rs564737089 Chr.10: 30016926-30016926 on Build GRCh38
rs942308756 Chr.10: 30016612-30016612 on Build GRCh38
rs1051479728 Chr.10: 30016657-30016657 on Build GRCh38
rs1024309709 Chr.10: 30016977-30016977 on Build GRCh38
rs975730774 Chr.10: 30016829-30016829 on Build GRCh38
rs956760530 Chr.10: 30016737-30016737 on Build GRCh38
rs982261761 Chr.10: 30016709-30016709 on Build GRCh38
rs1012899940 Chr.10: 30016558-30016558 on Build GRCh38
rs886874277 Chr.10: 30016874-30016874 on Build GRCh38
rs942664793 Chr.10: 30016791-30016791 on Build GRCh38
rs1016583654 Chr.10: 30016537-30016537 on Build GRCh38
rs902268900 Chr.10: 30016661-30016661 on Build GRCh38
rs981489908 Chr.10: 30016531-30016536 on Build GRCh38
rs557102292 Chr.10: 30016867-30016867 on Build GRCh38
rs771734781 Chr.10: 30016642-30016642 on Build GRCh38
rs922074559 Chr.10: 30016639-30016639 on Build GRCh38
rs774878067 Chr.10: 30016665-30016669 on Build GRCh38
rs915223615 Chr.10: 30016774-30016774 on Build GRCh38
rs1039720687 Chr.10: 30016633-30016633 on Build GRCh38
rs941068883 Chr.10: 30016876-30016876 on Build GRCh38
rs183003946 Chr.10: 30016885-30016885 on Build GRCh38
rs190413463 Chr.10: 30016818-30016818 on Build GRCh38
rs1023839414 Chr.10: 30016607-30016607 on Build GRCh38
rs922501689 Chr.10: 30016842-30016842 on Build GRCh38
rs933822500 Chr.10: 30016855-30016855 on Build GRCh38

View Full Product Details