Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 30017621-30018134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1462
Entrez Gene ID
57608
Gene Name
KIAA1462
Gene Aliases
JCAD
UniGene
Hs.533953
Gene Chromosome Location
Chr.10: 30012800-30069816 on Build GRCh38
Transcripts
NM_020848.2 XM_011519608.1 XM_011519609.2

SNP Detail

SNP ID Chromosome Location
rs762199377 Chr.10: 30017930-30017930 on Build GRCh38
rs750384557 Chr.10: 30017696-30017696 on Build GRCh38
rs376706706 Chr.10: 30017851-30017851 on Build GRCh38
rs955499590 Chr.10: 30017683-30017683 on Build GRCh38
rs777023178 Chr.10: 30017924-30017924 on Build GRCh38
rs752702227 Chr.10: 30017927-30017931 on Build GRCh38
rs747291054 Chr.10: 30017899-30017899 on Build GRCh38
rs767117425 Chr.10: 30017955-30017955 on Build GRCh38
rs982686291 Chr.10: 30017684-30017684 on Build GRCh38
rs765138074 Chr.10: 30017658-30017658 on Build GRCh38
rs765746381 Chr.10: 30017844-30017844 on Build GRCh38
rs770009857 Chr.10: 30017943-30017943 on Build GRCh38
rs1016314029 Chr.10: 30017843-30017843 on Build GRCh38
rs915658174 Chr.10: 30017747-30017747 on Build GRCh38
rs763299579 Chr.10: 30017952-30017952 on Build GRCh38
rs879028384 Chr.10: 30017695-30017695 on Build GRCh38
rs374891227 Chr.10: 30017796-30017796 on Build GRCh38
rs150699597 Chr.10: 30018087-30018087 on Build GRCh38
rs777765976 Chr.10: 30017866-30017866 on Build GRCh38
rs990057916 Chr.10: 30017962-30017962 on Build GRCh38
rs776968011 Chr.10: 30017836-30017836 on Build GRCh38
rs955355433 Chr.10: 30017893-30017893 on Build GRCh38
rs962539064 Chr.10: 30017699-30017699 on Build GRCh38
rs142972033 Chr.10: 30017882-30017882 on Build GRCh38
rs1030108929 Chr.10: 30017886-30017886 on Build GRCh38
rs766876687 Chr.10: 30017852-30017852 on Build GRCh38
rs755821724 Chr.10: 30017861-30017861 on Build GRCh38
rs868030902 Chr.10: 30017751-30017751 on Build GRCh38
rs947684553 Chr.10: 30017755-30017755 on Build GRCh38
rs576596543 Chr.10: 30017797-30017797 on Build GRCh38
rs186219328 Chr.10: 30017991-30017991 on Build GRCh38
rs981207757 Chr.10: 30018018-30018018 on Build GRCh38
rs947820019 Chr.10: 30018030-30018030 on Build GRCh38
rs530528706 Chr.10: 30018093-30018093 on Build GRCh38
rs1052636122 Chr.10: 30017830-30017830 on Build GRCh38
rs980516729 Chr.10: 30018049-30018049 on Build GRCh38
rs1046988683 Chr.10: 30017814-30017814 on Build GRCh38
rs948474808 Chr.10: 30017770-30017770 on Build GRCh38
rs934332871 Chr.10: 30018043-30018043 on Build GRCh38
rs760452207 Chr.10: 30017961-30017961 on Build GRCh38
rs906430225 Chr.10: 30017795-30017795 on Build GRCh38
rs77154162 Chr.10: 30018019-30018019 on Build GRCh38
rs750859058 Chr.10: 30017847-30017847 on Build GRCh38
rs558441047 Chr.10: 30017780-30017780 on Build GRCh38
rs752239495 Chr.10: 30017960-30017960 on Build GRCh38
rs927614822 Chr.10: 30018070-30018070 on Build GRCh38
rs374219802 Chr.10: 30017837-30017837 on Build GRCh38
rs12762440 Chr.10: 30017950-30017950 on Build GRCh38
rs370794553 Chr.10: 30017873-30017873 on Build GRCh38
rs1052687247 Chr.10: 30018074-30018074 on Build GRCh38
rs1011039166 Chr.10: 30017854-30017854 on Build GRCh38
rs2478839 Chr.10: 30017875-30017875 on Build GRCh38
rs543268865 Chr.10: 30017733-30017733 on Build GRCh38
rs939639954 Chr.10: 30017816-30017816 on Build GRCh38
rs914983717 Chr.10: 30017700-30017700 on Build GRCh38
rs989237024 Chr.10: 30017687-30017687 on Build GRCh38
rs754197429 Chr.10: 30018025-30018025 on Build GRCh38
rs996152761 Chr.10: 30017856-30017856 on Build GRCh38
rs768844722 Chr.10: 30017917-30017917 on Build GRCh38
rs745306243 Chr.10: 30017966-30017966 on Build GRCh38
rs962590568 Chr.10: 30017953-30017953 on Build GRCh38
rs561341505 Chr.10: 30018024-30018024 on Build GRCh38
rs778939984 Chr.10: 30017874-30017874 on Build GRCh38
rs752398867 Chr.10: 30017857-30017857 on Build GRCh38

View Full Product Details