Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89417767-89418268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT5
Entrez Gene ID
24138
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
ISG58, P58, RI58
UniGene
Hs.252839
Gene Chromosome Location
Chr.10: 89414568-89421002 on Build GRCh38
Transcripts
NM_012420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1968752 c.933G>A p.K311K Chr.10: 89418132-89418132 on Build GRCh38
6683843 c.631A>G p.N211D Chr.10: 89417830-89417830 on Build GRCh38
6683844 c.1041G>A p.M347I Chr.10: 89418240-89418240 on Build GRCh38
295276 c.940C>T p.L314L Chr.10: 89418139-89418139 on Build GRCh38
311918 c.1004C>T p.S335F Chr.10: 89418203-89418203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029117055 Chr.10: 89417840-89417840 on Build GRCh38
rs772821401 Chr.10: 89418204-89418204 on Build GRCh38
rs138567361 Chr.10: 89417806-89417806 on Build GRCh38
rs754773069 Chr.10: 89417907-89417907 on Build GRCh38
rs777262908 Chr.10: 89418204-89418204 on Build GRCh38
rs968249503 Chr.10: 89417793-89417793 on Build GRCh38
rs752370089 Chr.10: 89418116-89418116 on Build GRCh38
rs137961574 Chr.10: 89418200-89418200 on Build GRCh38
rs541824745 Chr.10: 89418206-89418206 on Build GRCh38
rs373762041 Chr.10: 89418171-89418171 on Build GRCh38
rs1015092578 Chr.10: 89417925-89417925 on Build GRCh38
rs749609057 Chr.10: 89418090-89418090 on Build GRCh38
rs747098320 Chr.10: 89418157-89418157 on Build GRCh38
rs537711619 Chr.10: 89417977-89417977 on Build GRCh38
rs760523848 Chr.10: 89417905-89417905 on Build GRCh38
rs765034469 Chr.10: 89418167-89418167 on Build GRCh38
rs772734921 Chr.10: 89418119-89418119 on Build GRCh38
rs771602485 Chr.10: 89418041-89418043 on Build GRCh38
rs773652579 Chr.10: 89417830-89417830 on Build GRCh38
rs757943926 Chr.10: 89417933-89417933 on Build GRCh38
rs558900926 Chr.10: 89417989-89417989 on Build GRCh38
rs757239514 Chr.10: 89417803-89417803 on Build GRCh38
rs767044177 Chr.10: 89418205-89418205 on Build GRCh38
rs780908003 Chr.10: 89418085-89418085 on Build GRCh38
rs749932525 Chr.10: 89417912-89417912 on Build GRCh38
rs748481306 Chr.10: 89417810-89417810 on Build GRCh38
rs867721703 Chr.10: 89418140-89418140 on Build GRCh38
rs914362521 Chr.10: 89418213-89418213 on Build GRCh38
rs984930247 Chr.10: 89417860-89417860 on Build GRCh38
rs771476722 Chr.10: 89417837-89417837 on Build GRCh38
rs978004153 Chr.10: 89417801-89417801 on Build GRCh38
rs369610086 Chr.10: 89418113-89418113 on Build GRCh38
rs767844750 Chr.10: 89417877-89417877 on Build GRCh38
rs775734590 Chr.10: 89417979-89417979 on Build GRCh38
rs775644758 Chr.10: 89418137-89418137 on Build GRCh38
rs745398622 Chr.10: 89417805-89417805 on Build GRCh38
rs759654456 Chr.10: 89417970-89417970 on Build GRCh38
rs747404756 Chr.10: 89418196-89418196 on Build GRCh38
rs535431435 Chr.10: 89417881-89417881 on Build GRCh38
rs768414149 Chr.10: 89417794-89417795 on Build GRCh38
rs141518852 Chr.10: 89418035-89418035 on Build GRCh38
rs372927340 Chr.10: 89418054-89418054 on Build GRCh38
rs754532465 Chr.10: 89418077-89418077 on Build GRCh38
rs369805092 Chr.10: 89417957-89417957 on Build GRCh38
rs304448 Chr.10: 89418243-89418243 on Build GRCh38
rs780952714 Chr.10: 89418087-89418087 on Build GRCh38
rs778968540 Chr.10: 89417909-89417909 on Build GRCh38
rs774234616 Chr.10: 89417844-89417844 on Build GRCh38
rs750409421 Chr.10: 89418026-89418026 on Build GRCh38
rs763331428 Chr.10: 89417836-89417836 on Build GRCh38
rs1040033922 Chr.10: 89418070-89418070 on Build GRCh38
rs761492728 Chr.10: 89418166-89418166 on Build GRCh38
rs768331042 Chr.10: 89417949-89417949 on Build GRCh38
rs779379603 Chr.10: 89418235-89418235 on Build GRCh38
rs777935444 Chr.10: 89417950-89417950 on Build GRCh38
rs780773937 Chr.10: 89418246-89418246 on Build GRCh38
rs146899400 Chr.10: 89418006-89418006 on Build GRCh38
rs779503083 Chr.10: 89418047-89418047 on Build GRCh38
rs778244463 Chr.10: 89417804-89417804 on Build GRCh38
rs920114660 Chr.10: 89417987-89417987 on Build GRCh38
rs142605779 Chr.10: 89417944-89417944 on Build GRCh38
rs757074614 Chr.10: 89417908-89417908 on Build GRCh38
rs776952897 Chr.10: 89418161-89418161 on Build GRCh38
rs200078169 Chr.10: 89418041-89418041 on Build GRCh38
rs761267068 Chr.10: 89417985-89417986 on Build GRCh38
rs376665939 Chr.10: 89417906-89417906 on Build GRCh38
rs751483932 Chr.10: 89417871-89417871 on Build GRCh38
rs556953266 Chr.10: 89417956-89417956 on Build GRCh38
rs372707015 Chr.10: 89417857-89417857 on Build GRCh38
rs770196223 Chr.10: 89417825-89417825 on Build GRCh38
rs771362408 Chr.10: 89418108-89418108 on Build GRCh38
rs534834408 Chr.10: 89418032-89418032 on Build GRCh38
rs774985156 Chr.10: 89417963-89417963 on Build GRCh38
rs774364102 Chr.10: 89417961-89417961 on Build GRCh38
rs755743504 Chr.10: 89418222-89418222 on Build GRCh38
rs142354157 Chr.10: 89418238-89418238 on Build GRCh38
rs150755193 Chr.10: 89418197-89418197 on Build GRCh38
rs267602615 Chr.10: 89418051-89418051 on Build GRCh38
rs148357594 Chr.10: 89418018-89418018 on Build GRCh38
rs200178716 Chr.10: 89418007-89418007 on Build GRCh38

View Full Product Details