Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89419269-89419775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT5
Entrez Gene ID
24138
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
ISG58, P58, RI58
UniGene
Hs.252839
Gene Chromosome Location
Chr.10: 89414568-89421002 on Build GRCh38
Transcripts
NM_012420.2

SNP Detail

SNP ID Chromosome Location
rs979009811 Chr.10: 89419348-89419348 on Build GRCh38
rs200039526 Chr.10: 89419380-89419380 on Build GRCh38
rs530645593 Chr.10: 89419361-89419364 on Build GRCh38
rs917707580 Chr.10: 89419421-89419421 on Build GRCh38
rs1043279457 Chr.10: 89419684-89419684 on Build GRCh38
rs140611897 Chr.10: 89419643-89419643 on Build GRCh38
rs931229711 Chr.10: 89419522-89419522 on Build GRCh38
rs758493301 Chr.10: 89419369-89419381 on Build GRCh38
rs976964106 Chr.10: 89419406-89419414 on Build GRCh38
rs371002806 Chr.10: 89419361-89419361 on Build GRCh38
rs555050244 Chr.10: 89419606-89419606 on Build GRCh38
rs1004202063 Chr.10: 89419659-89419659 on Build GRCh38
rs1019557479 Chr.10: 89419691-89419691 on Build GRCh38
rs575253268 Chr.10: 89419515-89419515 on Build GRCh38
rs954038616 Chr.10: 89419505-89419505 on Build GRCh38
rs201404641 Chr.10: 89419373-89419373 on Build GRCh38
rs1031353706 Chr.10: 89419751-89419751 on Build GRCh38
rs987803810 Chr.10: 89419391-89419391 on Build GRCh38
rs397698880 Chr.10: 89419372-89419373 on Build GRCh38
rs201018447 Chr.10: 89419381-89419381 on Build GRCh38
rs58331391 Chr.10: 89419361-89419361 on Build GRCh38
rs568301706 Chr.10: 89419752-89419752 on Build GRCh38
rs145790187 Chr.10: 89419378-89419381 on Build GRCh38
rs200106760 Chr.10: 89419358-89419359 on Build GRCh38
rs942004618 Chr.10: 89419559-89419559 on Build GRCh38
rs773840373 Chr.10: 89419362-89419365 on Build GRCh38
rs748562220 Chr.10: 89419360-89419366 on Build GRCh38
rs1057468106 Chr.10: 89419649-89419650 on Build GRCh38
rs919321216 Chr.10: 89419586-89419586 on Build GRCh38
rs542480478 Chr.10: 89419614-89419614 on Build GRCh38
rs147725383 Chr.10: 89419708-89419708 on Build GRCh38
rs894638226 Chr.10: 89419662-89419662 on Build GRCh38
rs904996225 Chr.10: 89419469-89419469 on Build GRCh38
rs938764780 Chr.10: 89419648-89419648 on Build GRCh38
rs67858414 Chr.10: 89419360-89419363 on Build GRCh38
rs61855171 Chr.10: 89419377-89419377 on Build GRCh38
rs949152285 Chr.10: 89419436-89419436 on Build GRCh38
rs1022333339 Chr.10: 89419314-89419314 on Build GRCh38
rs869160108 Chr.10: 89419381-89419383 on Build GRCh38
rs1031384666 Chr.10: 89419363-89419363 on Build GRCh38
rs998497605 Chr.10: 89419710-89419710 on Build GRCh38
rs972043089 Chr.10: 89419566-89419566 on Build GRCh38
rs374809691 Chr.10: 89419361-89419362 on Build GRCh38
rs887022988 Chr.10: 89419568-89419568 on Build GRCh38
rs1045325884 Chr.10: 89419462-89419462 on Build GRCh38
rs145505742 Chr.10: 89419707-89419707 on Build GRCh38
rs998528357 Chr.10: 89419358-89419361 on Build GRCh38
rs948932742 Chr.10: 89419682-89419682 on Build GRCh38
rs1013149862 Chr.10: 89419297-89419297 on Build GRCh38
rs11387675 Chr.10: 89419366-89419367 on Build GRCh38
rs1028145633 Chr.10: 89419498-89419501 on Build GRCh38
rs770532812 Chr.10: 89419377-89419378 on Build GRCh38
rs57143517 Chr.10: 89419381-89419384 on Build GRCh38
rs934947321 Chr.10: 89419365-89419365 on Build GRCh38
rs184236 Chr.10: 89419538-89419538 on Build GRCh38
rs755751959 Chr.10: 89419359-89419360 on Build GRCh38
rs760774941 Chr.10: 89419340-89419340 on Build GRCh38

View Full Product Details