Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101061134-101061627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAZALD1
Entrez Gene ID
81621
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
BONO1, FKSG28, FKSG40, IGFBP-rP10
UniGene
Hs.534859
Gene Chromosome Location
Chr.10: 101061141-101068141 on Build GRCh38
Transcripts
NM_001319303.1 NM_030929.4 NR_135067.1 XM_005270194.3

SNP Detail

SNP ID Chromosome Location
rs1022619221 Chr.10: 101061466-101061466 on Build GRCh38
rs948995710 Chr.10: 101061222-101061222 on Build GRCh38
rs1051204501 Chr.10: 101061385-101061385 on Build GRCh38
rs1031898729 Chr.10: 101061221-101061221 on Build GRCh38
rs981188426 Chr.10: 101061263-101061263 on Build GRCh38
rs1036522426 Chr.10: 101061252-101061252 on Build GRCh38
rs373979754 Chr.10: 101061173-101061173 on Build GRCh38
rs1023053801 Chr.10: 101061242-101061242 on Build GRCh38
rs370310325 Chr.10: 101061165-101061165 on Build GRCh38
rs969722129 Chr.10: 101061472-101061472 on Build GRCh38
rs772465864 Chr.10: 101061281-101061281 on Build GRCh38
rs909079476 Chr.10: 101061576-101061576 on Build GRCh38
rs916177722 Chr.10: 101061186-101061186 on Build GRCh38
rs1035998414 Chr.10: 101061497-101061497 on Build GRCh38
rs893347260 Chr.10: 101061445-101061445 on Build GRCh38
rs999089764 Chr.10: 101061191-101061191 on Build GRCh38
rs1048624991 Chr.10: 101061227-101061227 on Build GRCh38
rs989796384 Chr.10: 101061236-101061236 on Build GRCh38
rs1029829530 Chr.10: 101061171-101061171 on Build GRCh38
rs997770032 Chr.10: 101061265-101061265 on Build GRCh38
rs377155388 Chr.10: 101061368-101061368 on Build GRCh38
rs1035375388 Chr.10: 101061529-101061529 on Build GRCh38
rs1007523742 Chr.10: 101061271-101061271 on Build GRCh38
rs1042998066 Chr.10: 101061561-101061561 on Build GRCh38
rs998994742 Chr.10: 101061406-101061406 on Build GRCh38
rs1038370107 Chr.10: 101061336-101061336 on Build GRCh38
rs957869733 Chr.10: 101061228-101061228 on Build GRCh38
rs367892507 Chr.10: 101061278-101061278 on Build GRCh38
rs941946267 Chr.10: 101061329-101061329 on Build GRCh38
rs977944995 Chr.10: 101061156-101061156 on Build GRCh38
rs969774728 Chr.10: 101061257-101061257 on Build GRCh38
rs545621811 Chr.10: 101061324-101061324 on Build GRCh38
rs766430753 Chr.10: 101061161-101061161 on Build GRCh38
rs941707505 Chr.10: 101061487-101061487 on Build GRCh38
rs537004149 Chr.10: 101061155-101061155 on Build GRCh38
rs1032262592 Chr.10: 101061423-101061423 on Build GRCh38
rs1021547310 Chr.10: 101061275-101061275 on Build GRCh38
rs765066233 Chr.10: 101061469-101061469 on Build GRCh38
rs928456097 Chr.10: 101061269-101061269 on Build GRCh38
rs1030200385 Chr.10: 101061267-101061267 on Build GRCh38
rs975094644 Chr.10: 101061583-101061583 on Build GRCh38
rs530672161 Chr.10: 101061464-101061464 on Build GRCh38
rs755196947 Chr.10: 101061403-101061403 on Build GRCh38
rs941950465 Chr.10: 101061582-101061582 on Build GRCh38
rs899865196 Chr.10: 101061339-101061339 on Build GRCh38
rs921256605 Chr.10: 101061588-101061588 on Build GRCh38
rs574247858 Chr.10: 101061170-101061170 on Build GRCh38
rs998827598 Chr.10: 101061310-101061310 on Build GRCh38
rs983263017 Chr.10: 101061299-101061299 on Build GRCh38
rs1007368217 Chr.10: 101061544-101061544 on Build GRCh38
rs891233437 Chr.10: 101061401-101061401 on Build GRCh38
rs981093843 Chr.10: 101061475-101061475 on Build GRCh38
rs560105393 Chr.10: 101061224-101061224 on Build GRCh38
rs983266684 Chr.10: 101061570-101061570 on Build GRCh38
rs370581889 Chr.10: 101061533-101061533 on Build GRCh38
rs970192293 Chr.10: 101061392-101061392 on Build GRCh38
rs909098496 Chr.10: 101061328-101061328 on Build GRCh38
rs578169627 Chr.10: 101061601-101061601 on Build GRCh38
rs560969664 Chr.10: 101061536-101061536 on Build GRCh38

View Full Product Details