Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119326343-119326840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK5
Entrez Gene ID
2869
Gene Name
G protein-coupled receptor kinase 5
Gene Aliases
GPRK5
UniGene
Hs.524625
Gene Chromosome Location
Chr.10: 119207685-119455619 on Build GRCh38
Transcripts
NM_005308.2 XM_005269707.1 XM_005269708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5096724 c.86A>G p.K29R Chr.10: 119326549-119326549 on Build GRCh38
6675756 c.68G>A p.R23H Chr.10: 119326531-119326531 on Build GRCh38
4984984 c.122A>T p.Q41L Chr.10: 119326585-119326585 on Build GRCh38
6064961 c.126T>A p.C42* Chr.10: 119326589-119326589 on Build GRCh38
6064960 c.126T>A p.C42* Chr.10: 119326589-119326589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747058699 Chr.10: 119326608-119326608 on Build GRCh38
rs996363540 Chr.10: 119326740-119326740 on Build GRCh38
rs190852531 Chr.10: 119326670-119326670 on Build GRCh38
rs199919771 Chr.10: 119326579-119326579 on Build GRCh38
rs750666305 Chr.10: 119326502-119326502 on Build GRCh38
rs369263199 Chr.10: 119326465-119326465 on Build GRCh38
rs1053536722 Chr.10: 119326402-119326402 on Build GRCh38
rs916437484 Chr.10: 119326622-119326622 on Build GRCh38
rs772304616 Chr.10: 119326602-119326602 on Build GRCh38
rs928692325 Chr.10: 119326387-119326387 on Build GRCh38
rs1005223941 Chr.10: 119326479-119326479 on Build GRCh38
rs913271616 Chr.10: 119326367-119326367 on Build GRCh38
rs148332973 Chr.10: 119326459-119326459 on Build GRCh38
rs116973720 Chr.10: 119326386-119326386 on Build GRCh38
rs377304963 Chr.10: 119326600-119326600 on Build GRCh38
rs749921571 Chr.10: 119326532-119326532 on Build GRCh38
rs1028740942 Chr.10: 119326742-119326742 on Build GRCh38
rs565807282 Chr.10: 119326443-119326443 on Build GRCh38
rs1048962399 Chr.10: 119326460-119326460 on Build GRCh38
rs11198880 Chr.10: 119326785-119326785 on Build GRCh38
rs762994858 Chr.10: 119326648-119326648 on Build GRCh38
rs778295658 Chr.10: 119326411-119326411 on Build GRCh38
rs372390813 Chr.10: 119326466-119326466 on Build GRCh38
rs556145937 Chr.10: 119326497-119326497 on Build GRCh38
rs773337501 Chr.10: 119326474-119326474 on Build GRCh38
rs901607457 Chr.10: 119326492-119326492 on Build GRCh38
rs1018535861 Chr.10: 119326485-119326485 on Build GRCh38
rs201715604 Chr.10: 119326518-119326518 on Build GRCh38
rs775497739 Chr.10: 119326644-119326644 on Build GRCh38
rs576302427 Chr.10: 119326578-119326578 on Build GRCh38
rs568154624 Chr.10: 119326463-119326463 on Build GRCh38
rs543414778 Chr.10: 119326778-119326778 on Build GRCh38
rs761548495 Chr.10: 119326522-119326522 on Build GRCh38
rs777904638 Chr.10: 119326606-119326606 on Build GRCh38
rs370900336 Chr.10: 119326662-119326662 on Build GRCh38
rs1040018952 Chr.10: 119326707-119326707 on Build GRCh38
rs897311 Chr.10: 119326792-119326792 on Build GRCh38
rs529612408 Chr.10: 119326684-119326684 on Build GRCh38
rs940588338 Chr.10: 119326700-119326700 on Build GRCh38
rs373525505 Chr.10: 119326496-119326496 on Build GRCh38
rs552329080 Chr.10: 119326814-119326814 on Build GRCh38
rs148368301 Chr.10: 119326521-119326521 on Build GRCh38
rs945030488 Chr.10: 119326741-119326741 on Build GRCh38
rs370069160 Chr.10: 119326494-119326494 on Build GRCh38
rs572157375 Chr.10: 119326638-119326638 on Build GRCh38
rs991578886 Chr.10: 119326609-119326609 on Build GRCh38
rs554107912 Chr.10: 119326382-119326382 on Build GRCh38
rs768774762 Chr.10: 119326650-119326650 on Build GRCh38
rs536374803 Chr.10: 119326430-119326430 on Build GRCh38
rs1036300945 Chr.10: 119326760-119326760 on Build GRCh38
rs930473712 Chr.10: 119326641-119326641 on Build GRCh38
rs922036915 Chr.10: 119326432-119326432 on Build GRCh38
rs886370094 Chr.10: 119326694-119326694 on Build GRCh38
rs2230345 Chr.10: 119326585-119326585 on Build GRCh38
rs1051830400 Chr.10: 119326691-119326691 on Build GRCh38
rs1020437475 Chr.10: 119326809-119326809 on Build GRCh38
rs541167969 Chr.10: 119326664-119326664 on Build GRCh38
rs376334654 Chr.10: 119326469-119326469 on Build GRCh38
rs554571626 Chr.10: 119326468-119326468 on Build GRCh38
rs989038286 Chr.10: 119326379-119326379 on Build GRCh38
rs767144498 Chr.10: 119326531-119326531 on Build GRCh38
rs901476134 Chr.10: 119326736-119326736 on Build GRCh38
rs2230345 Chr.10: 119326585-119326585 on Build GRCh38
rs1023421469 Chr.10: 119326530-119326530 on Build GRCh38
rs978966474 Chr.10: 119326661-119326661 on Build GRCh38
rs755630855 Chr.10: 119326533-119326533 on Build GRCh38
rs780564165 Chr.10: 119326571-119326571 on Build GRCh38
rs143234689 Chr.10: 119326414-119326414 on Build GRCh38
rs534817780 Chr.10: 119326457-119326457 on Build GRCh38

View Full Product Details