Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132537818-132538349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5A
Entrez Gene ID
3632
Gene Name
inositol polyphosphate-5-phosphatase A
Gene Aliases
5PTASE
UniGene
Hs.523360
Gene Chromosome Location
Chr.10: 132537779-132783480 on Build GRCh38
Transcripts
NM_005539.4 XM_017016205.1

SNP Detail

SNP ID Chromosome Location
rs1042814668 Chr.10: 132537893-132537893 on Build GRCh38
rs1055469706 Chr.10: 132537909-132537909 on Build GRCh38
rs533815336 Chr.10: 132538247-132538247 on Build GRCh38
rs763430676 Chr.10: 132538087-132538087 on Build GRCh38
rs753459311 Chr.10: 132538183-132538183 on Build GRCh38
rs192496226 Chr.10: 132538058-132538058 on Build GRCh38
rs941076346 Chr.10: 132538278-132538278 on Build GRCh38
rs973613432 Chr.10: 132538299-132538299 on Build GRCh38
rs1007483114 Chr.10: 132538050-132538050 on Build GRCh38
rs1020230355 Chr.10: 132538062-132538062 on Build GRCh38
rs1033761668 Chr.10: 132538201-132538201 on Build GRCh38
rs540258601 Chr.10: 132537902-132537902 on Build GRCh38
rs73395322 Chr.10: 132538328-132538328 on Build GRCh38
rs898098022 Chr.10: 132538215-132538215 on Build GRCh38
rs1027726363 Chr.10: 132537838-132537838 on Build GRCh38
rs1036653869 Chr.10: 132538006-132538006 on Build GRCh38
rs911099964 Chr.10: 132537855-132537855 on Build GRCh38
rs897968414 Chr.10: 132538008-132538008 on Build GRCh38
rs781535537 Chr.10: 132538129-132538129 on Build GRCh38
rs192044971 Chr.10: 132538307-132538307 on Build GRCh38
rs377744450 Chr.10: 132538274-132538275 on Build GRCh38
rs1018895027 Chr.10: 132538052-132538052 on Build GRCh38
rs950339297 Chr.10: 132537962-132537962 on Build GRCh38
rs374186663 Chr.10: 132538221-132538221 on Build GRCh38
rs768269758 Chr.10: 132538204-132538204 on Build GRCh38
rs986115614 Chr.10: 132537853-132537853 on Build GRCh38
rs977960895 Chr.10: 132538224-132538224 on Build GRCh38
rs936965934 Chr.10: 132537905-132537905 on Build GRCh38
rs534663009 Chr.10: 132537902-132537905 on Build GRCh38
rs995032253 Chr.10: 132538015-132538015 on Build GRCh38
rs542636664 Chr.10: 132537884-132537884 on Build GRCh38
rs966111602 Chr.10: 132538057-132538057 on Build GRCh38
rs112197169 Chr.10: 132537885-132537885 on Build GRCh38
rs896035578 Chr.10: 132537926-132537926 on Build GRCh38
rs959681372 Chr.10: 132538203-132538203 on Build GRCh38
rs568037162 Chr.10: 132538245-132538245 on Build GRCh38
rs559874623 Chr.10: 132538019-132538019 on Build GRCh38
rs929572713 Chr.10: 132538304-132538304 on Build GRCh38
rs751836665 Chr.10: 132538121-132538121 on Build GRCh38
rs1007596688 Chr.10: 132538248-132538248 on Build GRCh38
rs953352519 Chr.10: 132537848-132537848 on Build GRCh38
rs764513704 Chr.10: 132538118-132538118 on Build GRCh38
rs187283988 Chr.10: 132538298-132538298 on Build GRCh38
rs183215914 Chr.10: 132538138-132538138 on Build GRCh38
rs1036994704 Chr.10: 132538207-132538207 on Build GRCh38
rs1003661653 Chr.10: 132538185-132538185 on Build GRCh38
rs1022519669 Chr.10: 132538205-132538205 on Build GRCh38
rs371807331 Chr.10: 132538289-132538290 on Build GRCh38
rs562559842 Chr.10: 132538171-132538171 on Build GRCh38
rs913054862 Chr.10: 132537892-132537892 on Build GRCh38
rs920741224 Chr.10: 132538303-132538303 on Build GRCh38
rs1027673974 Chr.10: 132538018-132538018 on Build GRCh38
rs967587607 Chr.10: 132538093-132538093 on Build GRCh38
rs752908045 Chr.10: 132538086-132538086 on Build GRCh38
rs976731350 Chr.10: 132537885-132537886 on Build GRCh38
rs757827008 Chr.10: 132538125-132538125 on Build GRCh38
rs1005446045 Chr.10: 132537952-132537952 on Build GRCh38
rs889203899 Chr.10: 132538049-132538050 on Build GRCh38
rs748904085 Chr.10: 132538196-132538196 on Build GRCh38
rs978609524 Chr.10: 132538096-132538096 on Build GRCh38
rs910978551 Chr.10: 132538275-132538275 on Build GRCh38
rs11146414 Chr.10: 132538238-132538238 on Build GRCh38
rs112667537 Chr.10: 132537982-132537982 on Build GRCh38

View Full Product Details