Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2147557-2148058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2
Entrez Gene ID
3481
Gene Name
insulin like growth factor 2
Gene Aliases
C11orf43, GRDF, IGF-II, PP9974
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2149603 on Build GRCh38
Transcripts
NM_001007139.5 NR_028043.2 NR_133657.1 NM_001042376.2 NR_003512.3

Gene Symbol
IGF2-AS
Entrez Gene ID
51214
Gene Name
IGF2 antisense RNA
Gene Aliases
IGF2-AS1, IGF2AS, PEG8
UniGene
Hs.716962
Gene Chromosome Location
Chr.11: 2140512-2148666 on Build GRCh38
Transcripts
NM_001007139.5 NR_028043.2 NR_133657.1 NM_001042376.2 NR_003512.3

Gene Symbol
INS-IGF2
Entrez Gene ID
723961
Gene Name
INS-IGF2 readthrough
Gene Aliases
INSIGF
UniGene
Hs.272259
Gene Chromosome Location
Chr.11: 2129112-2161209 on Build GRCh38
Transcripts
NM_001007139.5 NR_028043.2 NR_133657.1 NM_001042376.2 NR_003512.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7372291 c.580C>G p.R194G Chr.11: 2147635-2147635 on Build GRCh38
6378672 c.563G>T p.C188F Chr.11: 2147652-2147652 on Build GRCh38
395440 c.589C>A p.R197S Chr.11: 2147626-2147626 on Build GRCh38
5372269 c.472T>C p.S158P Chr.11: 2147743-2147743 on Build GRCh38
5009189 c.590G>A p.R197H Chr.11: 2147625-2147625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997545933 Chr.11: 2147837-2147837 on Build GRCh38
rs948648025 Chr.11: 2147722-2147722 on Build GRCh38
rs974141698 Chr.11: 2147908-2147908 on Build GRCh38
rs923016450 Chr.11: 2147881-2147881 on Build GRCh38
rs548444408 Chr.11: 2147972-2147972 on Build GRCh38
rs760579823 Chr.11: 2147896-2147896 on Build GRCh38
rs536534912 Chr.11: 2147966-2147966 on Build GRCh38
rs992741620 Chr.11: 2147841-2147841 on Build GRCh38
rs1006881751 Chr.11: 2147701-2147701 on Build GRCh38
rs368323611 Chr.11: 2147788-2147788 on Build GRCh38
rs145087135 Chr.11: 2147660-2147660 on Build GRCh38
rs373049451 Chr.11: 2147825-2147825 on Build GRCh38
rs955781796 Chr.11: 2147863-2147863 on Build GRCh38
rs542049186 Chr.11: 2147759-2147759 on Build GRCh38
rs898470835 Chr.11: 2147714-2147714 on Build GRCh38
rs964326772 Chr.11: 2147831-2147831 on Build GRCh38
rs569860697 Chr.11: 2148011-2148011 on Build GRCh38
rs566008391 Chr.11: 2147946-2147946 on Build GRCh38
rs373622226 Chr.11: 2147758-2147758 on Build GRCh38
rs975676181 Chr.11: 2147627-2147627 on Build GRCh38
rs986992206 Chr.11: 2147875-2147875 on Build GRCh38
rs3741211 Chr.11: 2147880-2147880 on Build GRCh38
rs781110309 Chr.11: 2147795-2147795 on Build GRCh38
rs1052250040 Chr.11: 2147601-2147601 on Build GRCh38
rs1025644567 Chr.11: 2147994-2147994 on Build GRCh38
rs10770125 Chr.11: 2147784-2147784 on Build GRCh38
rs764042641 Chr.11: 2148026-2148026 on Build GRCh38
rs368782521 Chr.11: 2147885-2147885 on Build GRCh38
rs752989920 Chr.11: 2147752-2147752 on Build GRCh38
rs769367976 Chr.11: 2147614-2147614 on Build GRCh38
rs907064161 Chr.11: 2147745-2147745 on Build GRCh38
rs547735779 Chr.11: 2147899-2147899 on Build GRCh38
rs968282810 Chr.11: 2147626-2147626 on Build GRCh38
rs369566517 Chr.11: 2147688-2147688 on Build GRCh38
rs889035223 Chr.11: 2147909-2147909 on Build GRCh38
rs374993400 Chr.11: 2147577-2147577 on Build GRCh38
rs771064056 Chr.11: 2147665-2147665 on Build GRCh38
rs961083715 Chr.11: 2148005-2148005 on Build GRCh38
rs1021272922 Chr.11: 2147827-2147827 on Build GRCh38
rs765516077 Chr.11: 2147746-2147746 on Build GRCh38
rs762472654 Chr.11: 2147642-2147642 on Build GRCh38
rs564901104 Chr.11: 2147864-2147864 on Build GRCh38
rs1025698499 Chr.11: 2147834-2147834 on Build GRCh38
rs754199654 Chr.11: 2147735-2147735 on Build GRCh38
rs766966005 Chr.11: 2147716-2147716 on Build GRCh38
rs994147879 Chr.11: 2147990-2147990 on Build GRCh38
rs919948759 Chr.11: 2147983-2147983 on Build GRCh38
rs377731345 Chr.11: 2147847-2147847 on Build GRCh38
rs1006573731 Chr.11: 2147934-2147934 on Build GRCh38
rs368865985 Chr.11: 2147789-2147789 on Build GRCh38
rs755400457 Chr.11: 2147736-2147736 on Build GRCh38
rs934996772 Chr.11: 2147876-2147876 on Build GRCh38
rs1038398154 Chr.11: 2147700-2147700 on Build GRCh38
rs773951027 Chr.11: 2147677-2147677 on Build GRCh38
rs531475925 Chr.11: 2147802-2147802 on Build GRCh38
rs1013748673 Chr.11: 2148027-2148027 on Build GRCh38
rs375522759 Chr.11: 2147634-2147634 on Build GRCh38

View Full Product Details