Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62713748-62714251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL2
Entrez Gene ID
221092
Gene Name
heterogeneous nuclear ribonucleoprotein U like 2
Gene Aliases
HNRPUL2, SAF-A2
UniGene
Hs.657058
Gene Chromosome Location
Chr.11: 62712625-62727385 on Build GRCh38
Transcripts
NM_001079559.2 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001079559.2 NR_037946.1

SNP Detail

SNP ID Chromosome Location
rs754535791 Chr.11: 62714013-62714027 on Build GRCh38
rs911542692 Chr.11: 62714092-62714092 on Build GRCh38
rs919141236 Chr.11: 62713787-62713787 on Build GRCh38
rs550475868 Chr.11: 62713824-62713824 on Build GRCh38
rs1034968940 Chr.11: 62714000-62714000 on Build GRCh38
rs975204152 Chr.11: 62714154-62714154 on Build GRCh38
rs962776202 Chr.11: 62713788-62713789 on Build GRCh38
rs962834923 Chr.11: 62714047-62714047 on Build GRCh38
rs79417283 Chr.11: 62714195-62714195 on Build GRCh38
rs893973988 Chr.11: 62713914-62713914 on Build GRCh38
rs950586281 Chr.11: 62714086-62714086 on Build GRCh38
rs943068870 Chr.11: 62714123-62714123 on Build GRCh38
rs117949236 Chr.11: 62713858-62713858 on Build GRCh38
rs1011527484 Chr.11: 62713935-62713935 on Build GRCh38
rs758036002 Chr.11: 62714224-62714224 on Build GRCh38
rs917809360 Chr.11: 62713890-62713890 on Build GRCh38
rs992389243 Chr.11: 62714122-62714122 on Build GRCh38
rs78082051 Chr.11: 62714016-62714016 on Build GRCh38
rs962364597 Chr.11: 62714047-62714051 on Build GRCh38
rs112941223 Chr.11: 62714071-62714071 on Build GRCh38
rs187741131 Chr.11: 62713888-62713888 on Build GRCh38
rs926426522 Chr.11: 62713828-62713828 on Build GRCh38
rs1017118957 Chr.11: 62714147-62714147 on Build GRCh38
rs899897423 Chr.11: 62714146-62714146 on Build GRCh38
rs937861899 Chr.11: 62713830-62713830 on Build GRCh38
rs1021132139 Chr.11: 62713936-62713936 on Build GRCh38
rs548677470 Chr.11: 62714040-62714040 on Build GRCh38
rs753411971 Chr.11: 62713900-62713900 on Build GRCh38
rs754495850 Chr.11: 62714046-62714046 on Build GRCh38
rs917792550 Chr.11: 62714123-62714123 on Build GRCh38
rs530730294 Chr.11: 62713800-62713800 on Build GRCh38
rs752417125 Chr.11: 62714098-62714098 on Build GRCh38
rs945282235 Chr.11: 62714125-62714125 on Build GRCh38
rs184438627 Chr.11: 62713869-62713869 on Build GRCh38
rs547315410 Chr.11: 62714137-62714138 on Build GRCh38
rs1004838807 Chr.11: 62714205-62714205 on Build GRCh38
rs902736130 Chr.11: 62713984-62713984 on Build GRCh38
rs890140538 Chr.11: 62713863-62713863 on Build GRCh38
rs879745383 Chr.11: 62714037-62714037 on Build GRCh38
rs59226130 Chr.11: 62714146-62714147 on Build GRCh38
rs140720798 Chr.11: 62714178-62714179 on Build GRCh38
rs919109989 Chr.11: 62714084-62714084 on Build GRCh38
rs1054552091 Chr.11: 62714001-62714001 on Build GRCh38
rs1051691698 Chr.11: 62714189-62714189 on Build GRCh38
rs76527578 Chr.11: 62714193-62714193 on Build GRCh38
rs1038777777 Chr.11: 62714174-62714174 on Build GRCh38
rs566124760 Chr.11: 62713894-62713894 on Build GRCh38
rs755155139 Chr.11: 62714138-62714138 on Build GRCh38
rs955798408 Chr.11: 62714041-62714041 on Build GRCh38
rs867587794 Chr.11: 62714137-62714137 on Build GRCh38
rs1003762162 Chr.11: 62713998-62713998 on Build GRCh38
rs914942916 Chr.11: 62714183-62714183 on Build GRCh38
rs887352390 Chr.11: 62714133-62714133 on Build GRCh38
rs573645453 Chr.11: 62714140-62714140 on Build GRCh38
rs777560378 Chr.11: 62714225-62714225 on Build GRCh38
rs973797806 Chr.11: 62713789-62713789 on Build GRCh38
rs959139073 Chr.11: 62714121-62714121 on Build GRCh38
rs964186309 Chr.11: 62714150-62714150 on Build GRCh38
rs987415828 Chr.11: 62714088-62714088 on Build GRCh38
rs568565432 Chr.11: 62714070-62714070 on Build GRCh38
rs1042103547 Chr.11: 62713974-62713974 on Build GRCh38
rs764773117 Chr.11: 62714054-62714054 on Build GRCh38
rs982835941 Chr.11: 62713777-62713777 on Build GRCh38
rs556788282 Chr.11: 62714017-62714017 on Build GRCh38
rs756420542 Chr.11: 62714043-62714043 on Build GRCh38
rs879151241 Chr.11: 62714144-62714144 on Build GRCh38
rs370183371 Chr.11: 62713937-62713937 on Build GRCh38
rs1039218720 Chr.11: 62713905-62713905 on Build GRCh38
rs57919774 Chr.11: 62714141-62714141 on Build GRCh38
rs564420309 Chr.11: 62713827-62713827 on Build GRCh38
rs373006936 Chr.11: 62713930-62713930 on Build GRCh38
rs978035711 Chr.11: 62714139-62714139 on Build GRCh38
rs1051407555 Chr.11: 62713829-62713829 on Build GRCh38
rs1056286849 Chr.11: 62713881-62713881 on Build GRCh38

View Full Product Details