Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62715740-62716247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL2
Entrez Gene ID
221092
Gene Name
heterogeneous nuclear ribonucleoprotein U like 2
Gene Aliases
HNRPUL2, SAF-A2
UniGene
Hs.657058
Gene Chromosome Location
Chr.11: 62712625-62727385 on Build GRCh38
Transcripts
NM_001079559.2 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001079559.2 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5807136 c.2045G>C p.G682A Chr.11: 62715874-62715874 on Build GRCh38
3451075 c.2005G>A p.D669N Chr.11: 62715914-62715914 on Build GRCh38
2039523 c.2041C>T p.P681S Chr.11: 62715878-62715878 on Build GRCh38
4034873 c.1996C>A p.R666R Chr.11: 62715923-62715923 on Build GRCh38
5566625 c.1993C>T p.R665C Chr.11: 62715926-62715926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756840852 Chr.11: 62715979-62715979 on Build GRCh38
rs533366736 Chr.11: 62715936-62715936 on Build GRCh38
rs553530053 Chr.11: 62716080-62716080 on Build GRCh38
rs750787445 Chr.11: 62715824-62715824 on Build GRCh38
rs747224169 Chr.11: 62715925-62715925 on Build GRCh38
rs755534411 Chr.11: 62715866-62715866 on Build GRCh38
rs760963379 Chr.11: 62715957-62715957 on Build GRCh38
rs567991857 Chr.11: 62715811-62715811 on Build GRCh38
rs527903393 Chr.11: 62715803-62715803 on Build GRCh38
rs371447865 Chr.11: 62715865-62715865 on Build GRCh38
rs886832220 Chr.11: 62716003-62716003 on Build GRCh38
rs759978479 Chr.11: 62716070-62716070 on Build GRCh38
rs201181290 Chr.11: 62715857-62715857 on Build GRCh38
rs1003670170 Chr.11: 62716147-62716147 on Build GRCh38
rs770222606 Chr.11: 62716210-62716210 on Build GRCh38
rs370000407 Chr.11: 62715960-62715960 on Build GRCh38
rs780856606 Chr.11: 62715901-62715901 on Build GRCh38
rs201735517 Chr.11: 62715870-62715870 on Build GRCh38
rs182240817 Chr.11: 62716175-62716175 on Build GRCh38
rs1048973746 Chr.11: 62716070-62716071 on Build GRCh38
rs901361824 Chr.11: 62716038-62716038 on Build GRCh38
rs1019780851 Chr.11: 62716036-62716036 on Build GRCh38
rs777045003 Chr.11: 62716029-62716029 on Build GRCh38
rs572473752 Chr.11: 62716030-62716030 on Build GRCh38
rs917764300 Chr.11: 62715959-62715959 on Build GRCh38
rs376392244 Chr.11: 62715958-62715958 on Build GRCh38
rs1026937840 Chr.11: 62715856-62715856 on Build GRCh38
rs571276897 Chr.11: 62715786-62715786 on Build GRCh38
rs1013941618 Chr.11: 62715782-62715782 on Build GRCh38
rs367678403 Chr.11: 62715900-62715900 on Build GRCh38
rs888417342 Chr.11: 62716088-62716088 on Build GRCh38
rs751021478 Chr.11: 62715888-62715888 on Build GRCh38
rs375092244 Chr.11: 62715986-62715986 on Build GRCh38
rs1031138202 Chr.11: 62716168-62716168 on Build GRCh38
rs980636332 Chr.11: 62716182-62716182 on Build GRCh38
rs994746315 Chr.11: 62715849-62715849 on Build GRCh38
rs371971357 Chr.11: 62715918-62715918 on Build GRCh38
rs748222976 Chr.11: 62715933-62715933 on Build GRCh38
rs373890076 Chr.11: 62715976-62715976 on Build GRCh38
rs917741609 Chr.11: 62716114-62716114 on Build GRCh38
rs754451295 Chr.11: 62715851-62715851 on Build GRCh38
rs201075565 Chr.11: 62715950-62715950 on Build GRCh38
rs771267565 Chr.11: 62715992-62715992 on Build GRCh38
rs755246 Chr.11: 62715914-62715914 on Build GRCh38
rs760131465 Chr.11: 62715965-62715965 on Build GRCh38
rs540053290 Chr.11: 62716076-62716076 on Build GRCh38
rs547977650 Chr.11: 62715808-62715808 on Build GRCh38
rs778423083 Chr.11: 62716056-62716056 on Build GRCh38
rs367899280 Chr.11: 62715915-62715915 on Build GRCh38
rs761158059 Chr.11: 62715842-62715842 on Build GRCh38
rs992903723 Chr.11: 62716113-62716113 on Build GRCh38
rs956097891 Chr.11: 62716090-62716090 on Build GRCh38
rs747907520 Chr.11: 62715957-62715958 on Build GRCh38
rs930939115 Chr.11: 62715987-62715987 on Build GRCh38
rs774305592 Chr.11: 62715844-62715846 on Build GRCh38
rs779320303 Chr.11: 62716101-62716102 on Build GRCh38
rs371770781 Chr.11: 62715912-62715912 on Build GRCh38
rs573647000 Chr.11: 62716127-62716127 on Build GRCh38
rs766824486 Chr.11: 62715848-62715848 on Build GRCh38
rs564517044 Chr.11: 62715804-62715804 on Build GRCh38
rs1019810748 Chr.11: 62715899-62715899 on Build GRCh38
rs1036597284 Chr.11: 62716035-62716035 on Build GRCh38
rs944501260 Chr.11: 62716032-62716032 on Build GRCh38
rs772443171 Chr.11: 62715812-62715812 on Build GRCh38
rs911611923 Chr.11: 62716007-62716007 on Build GRCh38
rs745497599 Chr.11: 62715908-62715908 on Build GRCh38
rs774320558 Chr.11: 62715932-62715932 on Build GRCh38
rs896368593 Chr.11: 62715772-62715772 on Build GRCh38
rs767734482 Chr.11: 62715817-62715817 on Build GRCh38
rs1048092774 Chr.11: 62715991-62715991 on Build GRCh38
rs765725887 Chr.11: 62715972-62715972 on Build GRCh38
rs1029025996 Chr.11: 62716055-62716055 on Build GRCh38
rs906603073 Chr.11: 62716139-62716139 on Build GRCh38
rs377016295 Chr.11: 62715813-62715813 on Build GRCh38
rs997104936 Chr.11: 62716046-62716046 on Build GRCh38
rs542504198 Chr.11: 62716170-62716170 on Build GRCh38

View Full Product Details