Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119043817-119044357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
UniGene
Hs.277704
Gene Chromosome Location
Chr.11: 119044185-119057246 on Build GRCh38
Transcripts
NM_001130991.2 NM_006389.4 XM_005271392.3 XM_005271393.3 XM_005271394.3 XM_017017095.1 XM_017017096.1 XM_017017097.1

SNP Detail

SNP ID Chromosome Location
rs995095916 Chr.11: 119043987-119043987 on Build GRCh38
rs782586583 Chr.11: 119043872-119043873 on Build GRCh38
rs1042449641 Chr.11: 119043898-119043898 on Build GRCh38
rs905528112 Chr.11: 119044328-119044328 on Build GRCh38
rs928031662 Chr.11: 119043905-119043905 on Build GRCh38
rs1024354171 Chr.11: 119044316-119044316 on Build GRCh38
rs915483291 Chr.11: 119043872-119043872 on Build GRCh38
rs995909085 Chr.11: 119044120-119044120 on Build GRCh38
rs932007411 Chr.11: 119043970-119043970 on Build GRCh38
rs1804691 Chr.11: 119044288-119044288 on Build GRCh38
rs1004497517 Chr.11: 119044088-119044088 on Build GRCh38
rs527558241 Chr.11: 119043919-119043919 on Build GRCh38
rs892652353 Chr.11: 119044174-119044174 on Build GRCh38
rs978069412 Chr.11: 119044301-119044301 on Build GRCh38
rs921464403 Chr.11: 119043858-119043858 on Build GRCh38
rs965560514 Chr.11: 119043941-119043941 on Build GRCh38
rs11358416 Chr.11: 119044027-119044027 on Build GRCh38
rs952629966 Chr.11: 119043866-119043866 on Build GRCh38
rs782317953 Chr.11: 119043950-119043950 on Build GRCh38
rs1026893200 Chr.11: 119043861-119043861 on Build GRCh38
rs986527746 Chr.11: 119044067-119044067 on Build GRCh38
rs939440979 Chr.11: 119043907-119043907 on Build GRCh38
rs368870827 Chr.11: 119043940-119043940 on Build GRCh38
rs886365332 Chr.11: 119044036-119044036 on Build GRCh38
rs1029171580 Chr.11: 119044156-119044156 on Build GRCh38
rs936889917 Chr.11: 119044127-119044127 on Build GRCh38
rs376470761 Chr.11: 119043926-119043926 on Build GRCh38
rs200310947 Chr.11: 119044212-119044213 on Build GRCh38
rs1784303 Chr.11: 119043878-119043878 on Build GRCh38
rs1048931545 Chr.11: 119044023-119044023 on Build GRCh38
rs2508952 Chr.11: 119043844-119043844 on Build GRCh38
rs1046154598 Chr.11: 119044194-119044194 on Build GRCh38
rs538629851 Chr.11: 119043965-119043965 on Build GRCh38
rs188410530 Chr.11: 119043849-119043849 on Build GRCh38
rs1052634002 Chr.11: 119044133-119044133 on Build GRCh38
rs114412931 Chr.11: 119043886-119043886 on Build GRCh38
rs193219264 Chr.11: 119044197-119044197 on Build GRCh38
rs927980878 Chr.11: 119044307-119044308 on Build GRCh38
rs376736100 Chr.11: 119044076-119044076 on Build GRCh38
rs782035211 Chr.11: 119044028-119044028 on Build GRCh38
rs796534893 Chr.11: 119044043-119044043 on Build GRCh38
rs73560986 Chr.11: 119044180-119044180 on Build GRCh38
rs976212870 Chr.11: 119043947-119043947 on Build GRCh38
rs34129550 Chr.11: 119043871-119043871 on Build GRCh38
rs35588561 Chr.11: 119044287-119044287 on Build GRCh38
rs1001915569 Chr.11: 119044199-119044199 on Build GRCh38
rs965747252 Chr.11: 119044104-119044104 on Build GRCh38
rs1018602919 Chr.11: 119044093-119044093 on Build GRCh38
rs1026588545 Chr.11: 119044223-119044223 on Build GRCh38
rs782439850 Chr.11: 119044027-119044028 on Build GRCh38

View Full Product Details