Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119045397-119045869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
UniGene
Hs.277704
Gene Chromosome Location
Chr.11: 119044185-119057246 on Build GRCh38
Transcripts
NM_001130991.2 NM_006389.4 XM_005271392.3 XM_005271393.3 XM_005271394.3 XM_017017095.1 XM_017017096.1 XM_017017097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466430 c.2933G>T p.G978V Chr.11: 119045786-119045786 on Build GRCh38
3738883 c.2978C>A p.P993H Chr.11: 119045615-119045615 on Build GRCh38
2106233 c.2899G>T p.G967* Chr.11: 119045820-119045820 on Build GRCh38
7251991 c.2888-10C>G p.? Chr.11: 119045841-119045841 on Build GRCh38
6131183 c.2888G>T p.G963V Chr.11: 119045831-119045831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200381445 Chr.11: 119045667-119045667 on Build GRCh38
rs781817732 Chr.11: 119045616-119045616 on Build GRCh38
rs757441842 Chr.11: 119045440-119045440 on Build GRCh38
rs972914098 Chr.11: 119045567-119045567 on Build GRCh38
rs895432402 Chr.11: 119045828-119045828 on Build GRCh38
rs781877508 Chr.11: 119045838-119045838 on Build GRCh38
rs1054343633 Chr.11: 119045708-119045708 on Build GRCh38
rs373944521 Chr.11: 119045826-119045826 on Build GRCh38
rs782012379 Chr.11: 119045791-119045791 on Build GRCh38
rs782755118 Chr.11: 119045614-119045614 on Build GRCh38
rs782427249 Chr.11: 119045449-119045454 on Build GRCh38
rs782613468 Chr.11: 119045664-119045664 on Build GRCh38
rs781979070 Chr.11: 119045845-119045849 on Build GRCh38
rs782266299 Chr.11: 119045688-119045688 on Build GRCh38
rs150272156 Chr.11: 119045652-119045652 on Build GRCh38
rs782171065 Chr.11: 119045779-119045779 on Build GRCh38
rs782014862 Chr.11: 119045492-119045492 on Build GRCh38
rs782601314 Chr.11: 119045549-119045549 on Build GRCh38
rs1003994885 Chr.11: 119045825-119045825 on Build GRCh38
rs782556723 Chr.11: 119045501-119045501 on Build GRCh38
rs782246082 Chr.11: 119045661-119045661 on Build GRCh38
rs782482598 Chr.11: 119045662-119045662 on Build GRCh38
rs782133967 Chr.11: 119045613-119045613 on Build GRCh38
rs138891090 Chr.11: 119045811-119045811 on Build GRCh38
rs782760728 Chr.11: 119045801-119045801 on Build GRCh38
rs782395218 Chr.11: 119045691-119045691 on Build GRCh38
rs782310949 Chr.11: 119045555-119045555 on Build GRCh38
rs541561621 Chr.11: 119045639-119045639 on Build GRCh38
rs45619037 Chr.11: 119045589-119045589 on Build GRCh38
rs782510386 Chr.11: 119045843-119045843 on Build GRCh38
rs782562037 Chr.11: 119045759-119045759 on Build GRCh38
rs781959451 Chr.11: 119045696-119045696 on Build GRCh38
rs573944250 Chr.11: 119045602-119045602 on Build GRCh38
rs782458703 Chr.11: 119045444-119045444 on Build GRCh38
rs781823008 Chr.11: 119045758-119045758 on Build GRCh38
rs923127159 Chr.11: 119045438-119045438 on Build GRCh38
rs782158571 Chr.11: 119045733-119045733 on Build GRCh38
rs953254593 Chr.11: 119045446-119045446 on Build GRCh38
rs782662603 Chr.11: 119045510-119045510 on Build GRCh38
rs781867851 Chr.11: 119045749-119045749 on Build GRCh38
rs781912639 Chr.11: 119045730-119045730 on Build GRCh38
rs782684099 Chr.11: 119045632-119045632 on Build GRCh38
rs189673760 Chr.11: 119045845-119045845 on Build GRCh38
rs45461798 Chr.11: 119045590-119045590 on Build GRCh38
rs373526795 Chr.11: 119045633-119045633 on Build GRCh38
rs765434164 Chr.11: 119045588-119045588 on Build GRCh38
rs782811813 Chr.11: 119045827-119045827 on Build GRCh38
rs782335558 Chr.11: 119045768-119045768 on Build GRCh38
rs906607472 Chr.11: 119045786-119045786 on Build GRCh38
rs58946406 Chr.11: 119045536-119045536 on Build GRCh38
rs782124085 Chr.11: 119045494-119045494 on Build GRCh38
rs782714519 Chr.11: 119045814-119045814 on Build GRCh38
rs782416423 Chr.11: 119045487-119045487 on Build GRCh38
rs531266604 Chr.11: 119045810-119045810 on Build GRCh38
rs781985105 Chr.11: 119045566-119045566 on Build GRCh38
rs782316376 Chr.11: 119045672-119045672 on Build GRCh38
rs781929414 Chr.11: 119045775-119045775 on Build GRCh38
rs938433081 Chr.11: 119045527-119045527 on Build GRCh38
rs535575901 Chr.11: 119045422-119045422 on Build GRCh38
rs373092360 Chr.11: 119045705-119045705 on Build GRCh38
rs782051553 Chr.11: 119045478-119045478 on Build GRCh38
rs781793271 Chr.11: 119045821-119045821 on Build GRCh38
rs782193367 Chr.11: 119045550-119045550 on Build GRCh38
rs782337963 Chr.11: 119045473-119045473 on Build GRCh38
rs150671584 Chr.11: 119045742-119045742 on Build GRCh38
rs369729316 Chr.11: 119045700-119045700 on Build GRCh38
rs562147605 Chr.11: 119045542-119045542 on Build GRCh38
rs572270319 Chr.11: 119045447-119045447 on Build GRCh38
rs780587471 Chr.11: 119045715-119045715 on Build GRCh38
rs533088185 Chr.11: 119045535-119045535 on Build GRCh38
rs1009667489 Chr.11: 119045449-119045449 on Build GRCh38
rs953296689 Chr.11: 119045650-119045650 on Build GRCh38
rs370908954 Chr.11: 119045766-119045766 on Build GRCh38
rs11607916 Chr.11: 119045606-119045606 on Build GRCh38
rs115207968 Chr.11: 119045844-119045844 on Build GRCh38
rs782355459 Chr.11: 119045550-119045550 on Build GRCh38
rs781892243 Chr.11: 119045476-119045476 on Build GRCh38
rs1014902861 Chr.11: 119045596-119045596 on Build GRCh38
rs377515064 Chr.11: 119045526-119045526 on Build GRCh38
rs782459894 Chr.11: 119045618-119045618 on Build GRCh38
rs555551 Chr.11: 119045594-119045594 on Build GRCh38
rs782206320 Chr.11: 119045495-119045496 on Build GRCh38
rs782817998 Chr.11: 119045605-119045605 on Build GRCh38
rs782578988 Chr.11: 119045453-119045455 on Build GRCh38
rs370905937 Chr.11: 119045450-119045450 on Build GRCh38
rs782125422 Chr.11: 119045797-119045797 on Build GRCh38
rs112560647 Chr.11: 119045780-119045780 on Build GRCh38
rs782294686 Chr.11: 119045450-119045450 on Build GRCh38
rs782254110 Chr.11: 119045767-119045767 on Build GRCh38
rs782502562 Chr.11: 119045750-119045750 on Build GRCh38
rs184161571 Chr.11: 119045717-119045717 on Build GRCh38
rs181046821 Chr.11: 119045496-119045496 on Build GRCh38
rs369505393 Chr.11: 119045558-119045558 on Build GRCh38
rs141932266 Chr.11: 119045475-119045475 on Build GRCh38
rs782042727 Chr.11: 119045687-119045687 on Build GRCh38
rs782475037 Chr.11: 119045548-119045548 on Build GRCh38
rs781797882 Chr.11: 119045499-119045499 on Build GRCh38
rs782682283 Chr.11: 119045557-119045557 on Build GRCh38
rs782281844 Chr.11: 119045447-119045447 on Build GRCh38
rs755613446 Chr.11: 119045794-119045794 on Build GRCh38
rs782227677 Chr.11: 119045465-119045465 on Build GRCh38
rs782338668 Chr.11: 119045587-119045587 on Build GRCh38
rs113118803 Chr.11: 119045833-119045833 on Build GRCh38
rs539602273 Chr.11: 119045460-119045460 on Build GRCh38
rs961497748 Chr.11: 119045561-119045561 on Build GRCh38
rs141977574 Chr.11: 119045625-119045625 on Build GRCh38
rs782779471 Chr.11: 119045495-119045495 on Build GRCh38
rs782101752 Chr.11: 119045576-119045576 on Build GRCh38
rs77766253 Chr.11: 119045423-119045423 on Build GRCh38
rs782427966 Chr.11: 119045783-119045783 on Build GRCh38

View Full Product Details