Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119048386-119048895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
UniGene
Hs.277704
Gene Chromosome Location
Chr.11: 119044185-119057246 on Build GRCh38
Transcripts
NM_001130991.2 NM_006389.4 XM_005271392.3 XM_005271393.3 XM_005271394.3 XM_017017095.1 XM_017017096.1 XM_017017097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291140 c.2234C>T p.A745V Chr.11: 119048495-119048495 on Build GRCh38
1561061 c.2029G>A p.V677I Chr.11: 119048850-119048850 on Build GRCh38
161647 c.2050G>C p.E684Q Chr.11: 119048829-119048829 on Build GRCh38
6371735 c.2231A>G p.E744G Chr.11: 119048498-119048498 on Build GRCh38
4018441 c.2084T>C p.M695T Chr.11: 119048795-119048795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368780055 Chr.11: 119048829-119048829 on Build GRCh38
rs115520137 Chr.11: 119048448-119048448 on Build GRCh38
rs782782755 Chr.11: 119048853-119048853 on Build GRCh38
rs782738483 Chr.11: 119048722-119048722 on Build GRCh38
rs782163982 Chr.11: 119048590-119048590 on Build GRCh38
rs946497294 Chr.11: 119048480-119048480 on Build GRCh38
rs782342968 Chr.11: 119048558-119048558 on Build GRCh38
rs987838110 Chr.11: 119048581-119048581 on Build GRCh38
rs782457061 Chr.11: 119048597-119048597 on Build GRCh38
rs781827314 Chr.11: 119048782-119048782 on Build GRCh38
rs782634334 Chr.11: 119048514-119048514 on Build GRCh38
rs782290466 Chr.11: 119048821-119048823 on Build GRCh38
rs2276059 Chr.11: 119048666-119048666 on Build GRCh38
rs782586020 Chr.11: 119048598-119048598 on Build GRCh38
rs201533554 Chr.11: 119048714-119048714 on Build GRCh38
rs782611769 Chr.11: 119048447-119048447 on Build GRCh38
rs372581628 Chr.11: 119048801-119048801 on Build GRCh38
rs782018600 Chr.11: 119048451-119048451 on Build GRCh38
rs141569642 Chr.11: 119048641-119048641 on Build GRCh38
rs781798213 Chr.11: 119048730-119048730 on Build GRCh38
rs782749634 Chr.11: 119048592-119048592 on Build GRCh38
rs200405558 Chr.11: 119048490-119048490 on Build GRCh38
rs782215503 Chr.11: 119048431-119048431 on Build GRCh38
rs372068451 Chr.11: 119048544-119048544 on Build GRCh38
rs782480667 Chr.11: 119048788-119048788 on Build GRCh38
rs899627532 Chr.11: 119048866-119048866 on Build GRCh38
rs782712882 Chr.11: 119048582-119048582 on Build GRCh38
rs781933792 Chr.11: 119048689-119048689 on Build GRCh38
rs376359962 Chr.11: 119048579-119048579 on Build GRCh38
rs183067184 Chr.11: 119048504-119048504 on Build GRCh38
rs781984097 Chr.11: 119048702-119048702 on Build GRCh38
rs782542840 Chr.11: 119048604-119048604 on Build GRCh38
rs782451647 Chr.11: 119048489-119048489 on Build GRCh38
rs782126475 Chr.11: 119048721-119048721 on Build GRCh38
rs375091924 Chr.11: 119048851-119048851 on Build GRCh38
rs201754853 Chr.11: 119048470-119048470 on Build GRCh38
rs781795601 Chr.11: 119048445-119048446 on Build GRCh38
rs564777944 Chr.11: 119048603-119048603 on Build GRCh38
rs1038941854 Chr.11: 119048458-119048458 on Build GRCh38
rs782567377 Chr.11: 119048446-119048446 on Build GRCh38
rs199521624 Chr.11: 119048816-119048816 on Build GRCh38
rs782680299 Chr.11: 119048812-119048812 on Build GRCh38
rs965679428 Chr.11: 119048701-119048701 on Build GRCh38
rs148386060 Chr.11: 119048847-119048847 on Build GRCh38
rs143389635 Chr.11: 119048548-119048548 on Build GRCh38
rs782490675 Chr.11: 119048864-119048864 on Build GRCh38
rs544181749 Chr.11: 119048628-119048628 on Build GRCh38
rs781895996 Chr.11: 119048495-119048495 on Build GRCh38
rs782163186 Chr.11: 119048452-119048452 on Build GRCh38
rs782220789 Chr.11: 119048823-119048823 on Build GRCh38
rs372725914 Chr.11: 119048434-119048434 on Build GRCh38
rs920325598 Chr.11: 119048436-119048436 on Build GRCh38
rs907429448 Chr.11: 119048435-119048436 on Build GRCh38
rs782506751 Chr.11: 119048411-119048411 on Build GRCh38
rs782306849 Chr.11: 119048683-119048683 on Build GRCh38
rs782316888 Chr.11: 119048522-119048522 on Build GRCh38
rs782790888 Chr.11: 119048765-119048765 on Build GRCh38
rs782328296 Chr.11: 119048832-119048832 on Build GRCh38
rs568922 Chr.11: 119048494-119048494 on Build GRCh38
rs781947923 Chr.11: 119048819-119048819 on Build GRCh38
rs146651379 Chr.11: 119048523-119048523 on Build GRCh38
rs973131070 Chr.11: 119048433-119048433 on Build GRCh38
rs983551572 Chr.11: 119048671-119048671 on Build GRCh38
rs572630019 Chr.11: 119048516-119048516 on Build GRCh38
rs782462531 Chr.11: 119048446-119048447 on Build GRCh38
rs782148656 Chr.11: 119048690-119048690 on Build GRCh38
rs373956190 Chr.11: 119048453-119048453 on Build GRCh38
rs782506920 Chr.11: 119048506-119048506 on Build GRCh38
rs201071396 Chr.11: 119048464-119048464 on Build GRCh38
rs148836750 Chr.11: 119048543-119048543 on Build GRCh38
rs782784022 Chr.11: 119048781-119048781 on Build GRCh38
rs782284822 Chr.11: 119048531-119048531 on Build GRCh38
rs782671293 Chr.11: 119048685-119048686 on Build GRCh38
rs781901605 Chr.11: 119048860-119048860 on Build GRCh38
rs191259381 Chr.11: 119048447-119048447 on Build GRCh38
rs202126223 Chr.11: 119048593-119048593 on Build GRCh38
rs782395503 Chr.11: 119048669-119048669 on Build GRCh38
rs782025304 Chr.11: 119048833-119048833 on Build GRCh38
rs781884275 Chr.11: 119048583-119048583 on Build GRCh38
rs571007163 Chr.11: 119048844-119048844 on Build GRCh38
rs782191000 Chr.11: 119048802-119048802 on Build GRCh38
rs566850822 Chr.11: 119048700-119048700 on Build GRCh38
rs2276060 Chr.11: 119048420-119048420 on Build GRCh38
rs782164696 Chr.11: 119048835-119048835 on Build GRCh38
rs147081612 Chr.11: 119048850-119048850 on Build GRCh38
rs147525277 Chr.11: 119048649-119048649 on Build GRCh38
rs782387110 Chr.11: 119048817-119048817 on Build GRCh38
rs782513791 Chr.11: 119048683-119048694 on Build GRCh38
rs532331360 Chr.11: 119048627-119048627 on Build GRCh38
rs782542366 Chr.11: 119048811-119048811 on Build GRCh38
rs782225234 Chr.11: 119048679-119048679 on Build GRCh38
rs782744198 Chr.11: 119048466-119048466 on Build GRCh38
rs782242670 Chr.11: 119048574-119048576 on Build GRCh38
rs782687466 Chr.11: 119048606-119048606 on Build GRCh38
rs782051651 Chr.11: 119048580-119048580 on Build GRCh38
rs782072886 Chr.11: 119048750-119048750 on Build GRCh38
rs45450894 Chr.11: 119048848-119048848 on Build GRCh38
rs1043196731 Chr.11: 119048482-119048482 on Build GRCh38
rs926056665 Chr.11: 119048808-119048808 on Build GRCh38
rs781974126 Chr.11: 119048449-119048449 on Build GRCh38
rs200858668 Chr.11: 119048772-119048772 on Build GRCh38
rs782595910 Chr.11: 119048674-119048674 on Build GRCh38
rs782506174 Chr.11: 119048779-119048779 on Build GRCh38

View Full Product Details