Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122728586-122729098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCAR1
Entrez Gene ID
27198
Gene Name
hydroxycarboxylic acid receptor 1
Gene Aliases
FKSG80, GPR104, GPR81, HCA1, LACR1, TA-GPCR, TAGPCR
UniGene
Hs.610873
Gene Chromosome Location
Chr.12: 122727606-122730582 on Build GRCh38
Transcripts
NM_032554.3

SNP Detail

SNP ID Chromosome Location
rs1039650237 Chr.12: 122729069-122729069 on Build GRCh38
rs753886801 Chr.12: 122728847-122728847 on Build GRCh38
rs539964008 Chr.12: 122728742-122728742 on Build GRCh38
rs997817577 Chr.12: 122728883-122728883 on Build GRCh38
rs143291434 Chr.12: 122728688-122728688 on Build GRCh38
rs534251205 Chr.12: 122728872-122728872 on Build GRCh38
rs566887621 Chr.12: 122728836-122728836 on Build GRCh38
rs548536087 Chr.12: 122728656-122728656 on Build GRCh38
rs556827420 Chr.12: 122728628-122728628 on Build GRCh38
rs1048597170 Chr.12: 122728915-122728915 on Build GRCh38
rs888581755 Chr.12: 122728948-122728948 on Build GRCh38
rs963698429 Chr.12: 122728862-122728862 on Build GRCh38
rs887566272 Chr.12: 122728631-122728631 on Build GRCh38
rs1017925947 Chr.12: 122728614-122728614 on Build GRCh38
rs559884998 Chr.12: 122728765-122728765 on Build GRCh38
rs572857687 Chr.12: 122728694-122728694 on Build GRCh38
rs1047369106 Chr.12: 122729032-122729032 on Build GRCh38
rs1014600345 Chr.12: 122728662-122728662 on Build GRCh38
rs991318957 Chr.12: 122728778-122728778 on Build GRCh38
rs766843591 Chr.12: 122728618-122728618 on Build GRCh38
rs513567 Chr.12: 122728689-122728689 on Build GRCh38
rs774924948 Chr.12: 122728629-122728629 on Build GRCh38
rs1043066258 Chr.12: 122728735-122728735 on Build GRCh38
rs913385428 Chr.12: 122728909-122728909 on Build GRCh38
rs192270496 Chr.12: 122728916-122728916 on Build GRCh38
rs947633711 Chr.12: 122728718-122728718 on Build GRCh38
rs927332467 Chr.12: 122728984-122728984 on Build GRCh38
rs1029828812 Chr.12: 122728886-122728886 on Build GRCh38
rs895868644 Chr.12: 122728764-122728764 on Build GRCh38
rs752940610 Chr.12: 122728758-122728758 on Build GRCh38
rs551238573 Chr.12: 122728822-122728822 on Build GRCh38
rs182289540 Chr.12: 122729035-122729035 on Build GRCh38
rs994409999 Chr.12: 122728803-122728803 on Build GRCh38
rs563160893 Chr.12: 122728823-122728823 on Build GRCh38
rs937353187 Chr.12: 122728992-122728992 on Build GRCh38
rs551962621 Chr.12: 122729020-122729020 on Build GRCh38
rs956457916 Chr.12: 122728891-122728891 on Build GRCh38
rs958603733 Chr.12: 122728705-122728705 on Build GRCh38
rs35614826 Chr.12: 122728958-122728958 on Build GRCh38
rs532595534 Chr.12: 122728809-122728809 on Build GRCh38
rs369101746 Chr.12: 122728675-122728675 on Build GRCh38
rs772873000 Chr.12: 122729033-122729033 on Build GRCh38
rs865936912 Chr.12: 122729018-122729018 on Build GRCh38
rs548574879 Chr.12: 122728831-122728831 on Build GRCh38
rs900065978 Chr.12: 122728611-122728611 on Build GRCh38
rs35439674 Chr.12: 122729022-122729022 on Build GRCh38
rs61955054 Chr.12: 122728824-122728824 on Build GRCh38
rs888283289 Chr.12: 122729039-122729039 on Build GRCh38
rs763382326 Chr.12: 122728610-122728610 on Build GRCh38
rs1057139380 Chr.12: 122728761-122728761 on Build GRCh38
rs538138122 Chr.12: 122729042-122729042 on Build GRCh38
rs932887136 Chr.12: 122728911-122728911 on Build GRCh38
rs536154445 Chr.12: 122729064-122729064 on Build GRCh38
rs1057094935 Chr.12: 122728999-122728999 on Build GRCh38
rs989396232 Chr.12: 122728903-122728903 on Build GRCh38
rs146228231 Chr.12: 122728829-122728829 on Build GRCh38
rs767902089 Chr.12: 122728695-122728695 on Build GRCh38
rs764105730 Chr.12: 122728839-122728839 on Build GRCh38
rs538872935 Chr.12: 122728622-122728622 on Build GRCh38
rs1036499702 Chr.12: 122728972-122728972 on Build GRCh38
rs865906805 Chr.12: 122728704-122728704 on Build GRCh38

View Full Product Details