Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122849785-122850283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1R
Entrez Gene ID
9026
Gene Name
huntingtin interacting protein 1 related
Gene Aliases
HIP12, HIP3, ILWEQ
UniGene
Hs.524815
Gene Chromosome Location
Chr.12: 122834498-122862961 on Build GRCh38
Transcripts
NM_001303097.1 NM_001303099.1 NM_003959.2 XM_011538963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7265305 c.379G>A p.G127R Chr.12: 122849896-122849896 on Build GRCh38
4039681 c.397T>G p.Y133D Chr.12: 122849914-122849914 on Build GRCh38
4039680 c.373C>T p.R125C Chr.12: 122849890-122849890 on Build GRCh38
3457410 c.426C>T p.S142S Chr.12: 122849943-122849943 on Build GRCh38
1726285 c.358-2A>G p.? Chr.12: 122849873-122849873 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs774873603 Chr.12: 122849839-122849839 on Build GRCh38
rs200886571 Chr.12: 122850197-122850197 on Build GRCh38
rs745988369 Chr.12: 122849830-122849830 on Build GRCh38
rs763711375 Chr.12: 122850052-122850052 on Build GRCh38
rs761378510 Chr.12: 122850057-122850057 on Build GRCh38
rs992066071 Chr.12: 122850202-122850202 on Build GRCh38
rs201536073 Chr.12: 122850200-122850200 on Build GRCh38
rs773451715 Chr.12: 122850027-122850027 on Build GRCh38
rs752410967 Chr.12: 122849854-122849854 on Build GRCh38
rs1051141839 Chr.12: 122849826-122849826 on Build GRCh38
rs148746514 Chr.12: 122850201-122850201 on Build GRCh38
rs772557306 Chr.12: 122850026-122850026 on Build GRCh38
rs59065210 Chr.12: 122849992-122849992 on Build GRCh38
rs534393785 Chr.12: 122850096-122850096 on Build GRCh38
rs763818021 Chr.12: 122849859-122849859 on Build GRCh38
rs753945873 Chr.12: 122850122-122850122 on Build GRCh38
rs755167763 Chr.12: 122850128-122850128 on Build GRCh38
rs35904336 Chr.12: 122849873-122849874 on Build GRCh38
rs769026117 Chr.12: 122849926-122849926 on Build GRCh38
rs748380036 Chr.12: 122850013-122850013 on Build GRCh38
rs375497752 Chr.12: 122850056-122850056 on Build GRCh38
rs200665144 Chr.12: 122849971-122849971 on Build GRCh38
rs543373503 Chr.12: 122850220-122850220 on Build GRCh38
rs953433346 Chr.12: 122849928-122849928 on Build GRCh38
rs556979438 Chr.12: 122850024-122850025 on Build GRCh38
rs370283420 Chr.12: 122850014-122850014 on Build GRCh38
rs956893630 Chr.12: 122849907-122849907 on Build GRCh38
rs753257768 Chr.12: 122849844-122849844 on Build GRCh38
rs866800765 Chr.12: 122850109-122850109 on Build GRCh38
rs145255391 Chr.12: 122849889-122849889 on Build GRCh38
rs774937442 Chr.12: 122850037-122850037 on Build GRCh38
rs943062424 Chr.12: 122850085-122850085 on Build GRCh38
rs986190714 Chr.12: 122850183-122850183 on Build GRCh38
rs762485840 Chr.12: 122850041-122850041 on Build GRCh38
rs748564101 Chr.12: 122849947-122849947 on Build GRCh38
rs921518178 Chr.12: 122850112-122850112 on Build GRCh38
rs769872722 Chr.12: 122849831-122849831 on Build GRCh38
rs774816650 Chr.12: 122849933-122849933 on Build GRCh38
rs770270467 Chr.12: 122849857-122849860 on Build GRCh38
rs936455820 Chr.12: 122850242-122850242 on Build GRCh38
rs557340185 Chr.12: 122850171-122850171 on Build GRCh38
rs918456306 Chr.12: 122850196-122850196 on Build GRCh38
rs752896487 Chr.12: 122850162-122850162 on Build GRCh38
rs548584172 Chr.12: 122850010-122850010 on Build GRCh38
rs775827866 Chr.12: 122849836-122849836 on Build GRCh38
rs769078882 Chr.12: 122849838-122849838 on Build GRCh38
rs903611917 Chr.12: 122850235-122850235 on Build GRCh38
rs373368900 Chr.12: 122849886-122849886 on Build GRCh38
rs758962482 Chr.12: 122849966-122849966 on Build GRCh38
rs779275507 Chr.12: 122850152-122850152 on Build GRCh38
rs953382636 Chr.12: 122850178-122850178 on Build GRCh38
rs747365400 Chr.12: 122850184-122850184 on Build GRCh38
rs200503518 Chr.12: 122850203-122850203 on Build GRCh38
rs770995513 Chr.12: 122850164-122850165 on Build GRCh38
rs758967757 Chr.12: 122849850-122849850 on Build GRCh38
rs750210581 Chr.12: 122850108-122850108 on Build GRCh38
rs552700131 Chr.12: 122850113-122850113 on Build GRCh38
rs755220709 Chr.12: 122850019-122850019 on Build GRCh38
rs1013349103 Chr.12: 122850254-122850254 on Build GRCh38
rs747406090 Chr.12: 122849857-122849857 on Build GRCh38
rs373090169 Chr.12: 122849997-122849997 on Build GRCh38
rs760133007 Chr.12: 122850156-122850156 on Build GRCh38
rs749468189 Chr.12: 122849925-122849925 on Build GRCh38
rs773830136 Chr.12: 122849964-122849964 on Build GRCh38
rs867590556 Chr.12: 122850182-122850182 on Build GRCh38
rs375128048 Chr.12: 122849891-122849891 on Build GRCh38
rs765282596 Chr.12: 122850185-122850185 on Build GRCh38
rs997799611 Chr.12: 122850167-122850167 on Build GRCh38
rs780289203 Chr.12: 122849899-122849899 on Build GRCh38
rs750310252 Chr.12: 122850000-122850000 on Build GRCh38
rs763762868 Chr.12: 122849985-122849985 on Build GRCh38
rs774461427 Chr.12: 122850171-122850205 on Build GRCh38
rs767287238 Chr.12: 122850081-122850081 on Build GRCh38
rs903852663 Chr.12: 122850189-122850189 on Build GRCh38
rs757121306 Chr.12: 122849993-122849993 on Build GRCh38
rs779328558 Chr.12: 122850023-122850023 on Build GRCh38
rs748779758 Chr.12: 122850234-122850234 on Build GRCh38
rs773722661 Chr.12: 122849860-122849869 on Build GRCh38
rs758050117 Chr.12: 122849858-122849858 on Build GRCh38
rs375845202 Chr.12: 122850137-122850137 on Build GRCh38
rs147270360 Chr.12: 122850163-122850163 on Build GRCh38
rs35465127 Chr.12: 122849895-122849895 on Build GRCh38
rs771449328 Chr.12: 122850036-122850036 on Build GRCh38
rs748449156 Chr.12: 122850024-122850024 on Build GRCh38
rs768100959 Chr.12: 122849843-122849843 on Build GRCh38
rs199779424 Chr.12: 122850179-122850179 on Build GRCh38
rs1038313669 Chr.12: 122849815-122849815 on Build GRCh38
rs757099141 Chr.12: 122849890-122849890 on Build GRCh38
rs539232427 Chr.12: 122850170-122850170 on Build GRCh38
rs745910954 Chr.12: 122849894-122849894 on Build GRCh38
rs1033569352 Chr.12: 122850207-122850207 on Build GRCh38
rs929593604 Chr.12: 122850070-122850070 on Build GRCh38
rs780387887 Chr.12: 122849853-122849853 on Build GRCh38
rs762544672 Chr.12: 122849981-122849981 on Build GRCh38
rs1056055591 Chr.12: 122850093-122850093 on Build GRCh38
rs766460516 Chr.12: 122850119-122850119 on Build GRCh38
rs774993045 Chr.12: 122849976-122849976 on Build GRCh38
rs899989884 Chr.12: 122849805-122849805 on Build GRCh38
rs759135728 Chr.12: 122849846-122849846 on Build GRCh38
rs533193550 Chr.12: 122849825-122849825 on Build GRCh38
rs917566292 Chr.12: 122850208-122850208 on Build GRCh38
rs772438231 Chr.12: 122849963-122849963 on Build GRCh38

View Full Product Details