Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123633603-123634005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B1
Entrez Gene ID
1967
Gene Name
eukaryotic translation initiation factor 2B subunit alpha
Gene Aliases
EIF2B, EIF2BA
UniGene
Hs.741273
Gene Chromosome Location
Chr.12: 123621023-123633776 on Build GRCh38
Transcripts
NM_001414.3 NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Gene Symbol
GTF2H3
Entrez Gene ID
2967
Gene Name
general transcription factor IIH subunit 3
Gene Aliases
BTF2, P34, TFB4, TFIIH
UniGene
Hs.355348
Gene Chromosome Location
Chr.12: 123633739-123662604 on Build GRCh38
Transcripts
NM_001414.3 NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4952027 c.13+2T>C p.? Chr.12: 123633874-123633874 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201984053 Chr.12: 123633852-123633852 on Build GRCh38
rs747907480 Chr.12: 123633909-123633909 on Build GRCh38
rs1054550347 Chr.12: 123633887-123633887 on Build GRCh38
rs946200742 Chr.12: 123633927-123633927 on Build GRCh38
rs775386431 Chr.12: 123633641-123633641 on Build GRCh38
rs530313748 Chr.12: 123633788-123633788 on Build GRCh38
rs749563752 Chr.12: 123633623-123633623 on Build GRCh38
rs750114147 Chr.12: 123633668-123633668 on Build GRCh38
rs199613674 Chr.12: 123633819-123633819 on Build GRCh38
rs531836165 Chr.12: 123633677-123633677 on Build GRCh38
rs866388073 Chr.12: 123633680-123633680 on Build GRCh38
rs1017864215 Chr.12: 123633973-123633973 on Build GRCh38
rs756472352 Chr.12: 123633622-123633622 on Build GRCh38
rs552019052 Chr.12: 123633970-123633970 on Build GRCh38
rs1007159574 Chr.12: 123633968-123633968 on Build GRCh38
rs758936507 Chr.12: 123633866-123633866 on Build GRCh38
rs748626099 Chr.12: 123633825-123633825 on Build GRCh38
rs535812755 Chr.12: 123633703-123633703 on Build GRCh38
rs774307674 Chr.12: 123633902-123633902 on Build GRCh38
rs146287209 Chr.12: 123633948-123633948 on Build GRCh38
rs891979220 Chr.12: 123633771-123633771 on Build GRCh38
rs1047214963 Chr.12: 123633868-123633868 on Build GRCh38
rs560179155 Chr.12: 123633830-123633830 on Build GRCh38
rs1029299006 Chr.12: 123633694-123633694 on Build GRCh38
rs376275529 Chr.12: 123633881-123633881 on Build GRCh38
rs1010918302 Chr.12: 123633693-123633693 on Build GRCh38
rs752753800 Chr.12: 123633834-123633834 on Build GRCh38
rs370745202 Chr.12: 123633921-123633921 on Build GRCh38
rs775614281 Chr.12: 123633649-123633650 on Build GRCh38
rs371062007 Chr.12: 123633836-123633836 on Build GRCh38
rs772047192 Chr.12: 123633911-123633911 on Build GRCh38
rs760198554 Chr.12: 123633648-123633649 on Build GRCh38
rs199831647 Chr.12: 123633842-123633842 on Build GRCh38
rs11572913 Chr.12: 123633862-123633862 on Build GRCh38
rs34827093 Chr.12: 123633701-123633701 on Build GRCh38
rs886049052 Chr.12: 123633648-123633648 on Build GRCh38
rs200812386 Chr.12: 123633833-123633833 on Build GRCh38
rs921712126 Chr.12: 123633632-123633632 on Build GRCh38
rs950090384 Chr.12: 123633747-123633747 on Build GRCh38
rs974878405 Chr.12: 123633631-123633631 on Build GRCh38
rs886049051 Chr.12: 123633627-123633627 on Build GRCh38
rs576953141 Chr.12: 123633750-123633750 on Build GRCh38
rs755379165 Chr.12: 123633856-123633856 on Build GRCh38
rs945961716 Chr.12: 123633697-123633697 on Build GRCh38
rs191107270 Chr.12: 123633719-123633719 on Build GRCh38
rs113832585 Chr.12: 123633759-123633759 on Build GRCh38
rs1001399707 Chr.12: 123633709-123633709 on Build GRCh38
rs760509266 Chr.12: 123633692-123633692 on Build GRCh38
rs762049228 Chr.12: 123633652-123633652 on Build GRCh38
rs1051394759 Chr.12: 123633658-123633658 on Build GRCh38
rs373212441 Chr.12: 123633947-123633947 on Build GRCh38
rs779339725 Chr.12: 123633634-123633634 on Build GRCh38
rs1010450879 Chr.12: 123633777-123633777 on Build GRCh38
rs112250456 Chr.12: 123633721-123633721 on Build GRCh38
rs1051302453 Chr.12: 123633941-123633941 on Build GRCh38
rs769177882 Chr.12: 123633625-123633625 on Build GRCh38
rs955571029 Chr.12: 123633925-123633925 on Build GRCh38
rs765567277 Chr.12: 123633860-123633860 on Build GRCh38
rs550230155 Chr.12: 123633700-123633700 on Build GRCh38
rs766686037 Chr.12: 123633854-123633854 on Build GRCh38
rs529377445 Chr.12: 123633930-123633930 on Build GRCh38
rs936088749 Chr.12: 123633760-123633765 on Build GRCh38
rs7294420 Chr.12: 123633942-123633942 on Build GRCh38
rs774415968 Chr.12: 123633651-123633651 on Build GRCh38
rs11572912 Chr.12: 123633701-123633702 on Build GRCh38
rs917217939 Chr.12: 123633743-123633743 on Build GRCh38
rs778832726 Chr.12: 123633765-123633765 on Build GRCh38
rs372604299 Chr.12: 123633822-123633822 on Build GRCh38
rs955219931 Chr.12: 123633699-123633699 on Build GRCh38
rs1054599700 Chr.12: 123633762-123633762 on Build GRCh38
rs1020015345 Chr.12: 123633931-123633931 on Build GRCh38
rs764139707 Chr.12: 123633644-123633644 on Build GRCh38
rs769282951 Chr.12: 123633640-123633640 on Build GRCh38
rs770426974 Chr.12: 123633637-123633637 on Build GRCh38
rs898172220 Chr.12: 123633981-123633981 on Build GRCh38
rs778042487 Chr.12: 123633885-123633885 on Build GRCh38
rs925921624 Chr.12: 123633839-123633839 on Build GRCh38
rs756811458 Chr.12: 123633827-123633827 on Build GRCh38
rs951577270 Chr.12: 123633761-123633761 on Build GRCh38
rs556198143 Chr.12: 123633800-123633800 on Build GRCh38
rs746584137 Chr.12: 123633636-123633636 on Build GRCh38
rs773368651 Chr.12: 123633849-123633849 on Build GRCh38
rs773013763 Chr.12: 123633920-123633920 on Build GRCh38
rs375308721 Chr.12: 123633779-123633779 on Build GRCh38
rs759205613 Chr.12: 123633638-123633638 on Build GRCh38
rs61954890 Chr.12: 123633664-123633664 on Build GRCh38
rs768111723 Chr.12: 123633655-123633667 on Build GRCh38
rs886049053 Chr.12: 123633707-123633707 on Build GRCh38
rs767670920 Chr.12: 123633809-123633809 on Build GRCh38
rs768618924 Chr.12: 123633886-123633886 on Build GRCh38
rs779821385 Chr.12: 123633882-123633882 on Build GRCh38
rs951291970 Chr.12: 123633850-123633850 on Build GRCh38
rs779466193 Chr.12: 123633823-123633823 on Build GRCh38
rs973857010 Chr.12: 123633712-123633712 on Build GRCh38
rs547242579 Chr.12: 123633855-123633855 on Build GRCh38
rs917817876 Chr.12: 123633858-123633858 on Build GRCh38
rs558453538 Chr.12: 123633740-123633740 on Build GRCh38
rs550128760 Chr.12: 123633880-123633880 on Build GRCh38
rs1000353832 Chr.12: 123633648-123633648 on Build GRCh38
rs756377630 Chr.12: 123633818-123633818 on Build GRCh38
rs1044433068 Chr.12: 123633758-123633758 on Build GRCh38
rs376458210 Chr.12: 123633812-123633812 on Build GRCh38
rs201242296 Chr.12: 123633829-123633829 on Build GRCh38
rs571053026 Chr.12: 123633642-123633642 on Build GRCh38
rs547389286 Chr.12: 123633717-123633717 on Build GRCh38
rs1025654793 Chr.12: 123633755-123633755 on Build GRCh38
rs774468940 Chr.12: 123633835-123633835 on Build GRCh38
rs537660940 Chr.12: 123633780-123633780 on Build GRCh38

View Full Product Details