Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123650973-123651515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H3
Entrez Gene ID
2967
Gene Name
general transcription factor IIH subunit 3
Gene Aliases
BTF2, P34, TFB4, TFIIH
UniGene
Hs.355348
Gene Chromosome Location
Chr.12: 123633739-123662604 on Build GRCh38
Transcripts
NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5548583 c.402A>G p.A134A Chr.12: 123651031-123651031 on Build GRCh38
3871003 c.407C>T p.S136F Chr.12: 123651036-123651036 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs919184532 Chr.12: 123651206-123651206 on Build GRCh38
rs1052053032 Chr.12: 123651494-123651494 on Build GRCh38
rs974858239 Chr.12: 123651451-123651451 on Build GRCh38
rs142387582 Chr.12: 123651356-123651356 on Build GRCh38
rs921895661 Chr.12: 123651469-123651469 on Build GRCh38
rs948136697 Chr.12: 123651233-123651233 on Build GRCh38
rs765277832 Chr.12: 123651054-123651054 on Build GRCh38
rs1052031345 Chr.12: 123651283-123651283 on Build GRCh38
rs892683628 Chr.12: 123651485-123651485 on Build GRCh38
rs1030007151 Chr.12: 123651284-123651284 on Build GRCh38
rs1020677657 Chr.12: 123651129-123651129 on Build GRCh38
rs189227743 Chr.12: 123651486-123651486 on Build GRCh38
rs903500029 Chr.12: 123651155-123651155 on Build GRCh38
rs11572968 Chr.12: 123651253-123651253 on Build GRCh38
rs558493386 Chr.12: 123651333-123651333 on Build GRCh38
rs183748434 Chr.12: 123651436-123651436 on Build GRCh38
rs756417238 Chr.12: 123651094-123651094 on Build GRCh38
rs11572969 Chr.12: 123651337-123651337 on Build GRCh38
rs780601445 Chr.12: 123651108-123651108 on Build GRCh38
rs1053042478 Chr.12: 123651482-123651482 on Build GRCh38
rs1030058005 Chr.12: 123651053-123651053 on Build GRCh38
rs200262579 Chr.12: 123651032-123651032 on Build GRCh38
rs759197890 Chr.12: 123651050-123651050 on Build GRCh38
rs117510880 Chr.12: 123651493-123651493 on Build GRCh38
rs750813241 Chr.12: 123651087-123651087 on Build GRCh38
rs894817877 Chr.12: 123651364-123651364 on Build GRCh38
rs950579274 Chr.12: 123651403-123651403 on Build GRCh38
rs963014701 Chr.12: 123651303-123651312 on Build GRCh38
rs776493836 Chr.12: 123651040-123651040 on Build GRCh38
rs1004708128 Chr.12: 123651410-123651410 on Build GRCh38
rs936825959 Chr.12: 123651472-123651472 on Build GRCh38
rs751137438 Chr.12: 123651277-123651277 on Build GRCh38
rs913366863 Chr.12: 123651297-123651297 on Build GRCh38
rs892737280 Chr.12: 123651265-123651265 on Build GRCh38
rs573864034 Chr.12: 123651251-123651251 on Build GRCh38
rs375738368 Chr.12: 123651104-123651104 on Build GRCh38
rs765730937 Chr.12: 123651138-123651138 on Build GRCh38
rs539115923 Chr.12: 123651065-123651065 on Build GRCh38
rs948084821 Chr.12: 123651395-123651395 on Build GRCh38
rs376292620 Chr.12: 123651066-123651066 on Build GRCh38
rs963486822 Chr.12: 123651257-123651257 on Build GRCh38
rs11572970 Chr.12: 123651434-123651434 on Build GRCh38
rs900744746 Chr.12: 123651048-123651048 on Build GRCh38
rs909045648 Chr.12: 123651069-123651069 on Build GRCh38
rs1009690347 Chr.12: 123651088-123651088 on Build GRCh38
rs35564084 Chr.12: 123651301-123651301 on Build GRCh38
rs1026194999 Chr.12: 123651348-123651348 on Build GRCh38
rs1013536437 Chr.12: 123651204-123651204 on Build GRCh38
rs144416237 Chr.12: 123651211-123651211 on Build GRCh38
rs531361902 Chr.12: 123651342-123651342 on Build GRCh38
rs113228730 Chr.12: 123651267-123651267 on Build GRCh38
rs1054709580 Chr.12: 123651358-123651358 on Build GRCh38
rs528780262 Chr.12: 123651445-123651445 on Build GRCh38
rs369307451 Chr.12: 123651068-123651068 on Build GRCh38
rs373248339 Chr.12: 123651074-123651074 on Build GRCh38
rs1013879737 Chr.12: 123651382-123651382 on Build GRCh38
rs775565025 Chr.12: 123651059-123651059 on Build GRCh38
rs1962085 Chr.12: 123651268-123651268 on Build GRCh38
rs574843932 Chr.12: 123651377-123651377 on Build GRCh38
rs149629171 Chr.12: 123651161-123651161 on Build GRCh38
rs1056397260 Chr.12: 123651213-123651214 on Build GRCh38
rs569242086 Chr.12: 123651188-123651188 on Build GRCh38
rs959547169 Chr.12: 123651196-123651196 on Build GRCh38
rs763168615 Chr.12: 123651063-123651063 on Build GRCh38
rs950585042 Chr.12: 123651249-123651249 on Build GRCh38
rs555591298 Chr.12: 123651250-123651250 on Build GRCh38
rs1007309534 Chr.12: 123651313-123651313 on Build GRCh38
rs189511157 Chr.12: 123651260-123651260 on Build GRCh38
rs575717186 Chr.12: 123651214-123651214 on Build GRCh38
rs556247001 Chr.12: 123650999-123650999 on Build GRCh38
rs1024912620 Chr.12: 123651383-123651383 on Build GRCh38
rs370948488 Chr.12: 123651222-123651222 on Build GRCh38
rs1045591319 Chr.12: 123651235-123651235 on Build GRCh38

View Full Product Details