Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123652322-123652730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H3
Entrez Gene ID
2967
Gene Name
general transcription factor IIH subunit 3
Gene Aliases
BTF2, P34, TFB4, TFIIH
UniGene
Hs.355348
Gene Chromosome Location
Chr.12: 123633739-123662604 on Build GRCh38
Transcripts
NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2225999 c.434A>G p.H145R Chr.12: 123652538-123652538 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747077234 Chr.12: 123652599-123652599 on Build GRCh38
rs1008942000 Chr.12: 123652593-123652593 on Build GRCh38
rs532706684 Chr.12: 123652433-123652433 on Build GRCh38
rs776138850 Chr.12: 123652557-123652557 on Build GRCh38
rs866350614 Chr.12: 123652549-123652549 on Build GRCh38
rs530113603 Chr.12: 123652589-123652589 on Build GRCh38
rs745743714 Chr.12: 123652570-123652570 on Build GRCh38
rs750602227 Chr.12: 123652658-123652658 on Build GRCh38
rs903161614 Chr.12: 123652644-123652644 on Build GRCh38
rs11572974 Chr.12: 123652385-123652385 on Build GRCh38
rs987589872 Chr.12: 123652466-123652466 on Build GRCh38
rs577804454 Chr.12: 123652602-123652602 on Build GRCh38
rs373533149 Chr.12: 123652572-123652572 on Build GRCh38
rs765745044 Chr.12: 123652550-123652550 on Build GRCh38
rs563117338 Chr.12: 123652573-123652573 on Build GRCh38
rs879029344 Chr.12: 123652591-123652591 on Build GRCh38
rs368867982 Chr.12: 123652543-123652543 on Build GRCh38
rs761740142 Chr.12: 123652609-123652609 on Build GRCh38
rs777299151 Chr.12: 123652506-123652506 on Build GRCh38
rs991656465 Chr.12: 123652353-123652353 on Build GRCh38
rs754165334 Chr.12: 123652680-123652680 on Build GRCh38
rs982277453 Chr.12: 123652428-123652428 on Build GRCh38
rs376413036 Chr.12: 123652606-123652606 on Build GRCh38
rs756539205 Chr.12: 123652662-123652662 on Build GRCh38
rs917447653 Chr.12: 123652374-123652374 on Build GRCh38
rs370690399 Chr.12: 123652514-123652514 on Build GRCh38
rs767833434 Chr.12: 123652610-123652610 on Build GRCh38
rs752074593 Chr.12: 123652680-123652680 on Build GRCh38
rs1044162709 Chr.12: 123652678-123652678 on Build GRCh38
rs763007606 Chr.12: 123652574-123652574 on Build GRCh38
rs1041801374 Chr.12: 123652618-123652618 on Build GRCh38
rs907500099 Chr.12: 123652470-123652470 on Build GRCh38
rs771937909 Chr.12: 123652482-123652482 on Build GRCh38
rs964931425 Chr.12: 123652489-123652489 on Build GRCh38
rs548696384 Chr.12: 123652630-123652630 on Build GRCh38
rs766705286 Chr.12: 123652674-123652674 on Build GRCh38
rs757949917 Chr.12: 123652498-123652498 on Build GRCh38

View Full Product Details