Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16015518-16016021 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB7
Entrez Gene ID
27129
Gene Name
heat shock protein family B (small) member 7
Gene Aliases
cvHSP
UniGene
Hs.502612
Gene Chromosome Location
Chr.1: 16014028-16018796 on Build GRCh38
Transcripts
NM_014424.4 XM_011541249.1 XM_011541250.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1335855 c.378C>T p.C126C Chr.1: 16015715-16015715 on Build GRCh38
4829622 c.449G>A p.R150Q Chr.1: 16015644-16015644 on Build GRCh38
5844331 c.431C>T p.S144L Chr.1: 16015677-16015677 on Build GRCh38
4576527 c.360C>T p.N120N Chr.1: 16015733-16015733 on Build GRCh38
5733474 c.509G>A p.R170Q Chr.1: 16015599-16015599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768245961 Chr.1: 16015676-16015676 on Build GRCh38
rs962795567 Chr.1: 16015746-16015746 on Build GRCh38
rs543729331 Chr.1: 16015545-16015545 on Build GRCh38
rs369621959 Chr.1: 16015778-16015778 on Build GRCh38
rs868049004 Chr.1: 16015819-16015819 on Build GRCh38
rs757356294 Chr.1: 16015707-16015707 on Build GRCh38
rs758535730 Chr.1: 16015600-16015600 on Build GRCh38
rs992078932 Chr.1: 16015668-16015668 on Build GRCh38
rs1000561368 Chr.1: 16015561-16015561 on Build GRCh38
rs776139790 Chr.1: 16015810-16015810 on Build GRCh38
rs370204847 Chr.1: 16015570-16015570 on Build GRCh38
rs11807575 Chr.1: 16015706-16015706 on Build GRCh38
rs769371992 Chr.1: 16015623-16015623 on Build GRCh38
rs757088984 Chr.1: 16015660-16015660 on Build GRCh38
rs143697552 Chr.1: 16015943-16015943 on Build GRCh38
rs752889799 Chr.1: 16015599-16015599 on Build GRCh38
rs925116101 Chr.1: 16015811-16015811 on Build GRCh38
rs754786908 Chr.1: 16015794-16015794 on Build GRCh38
rs201996831 Chr.1: 16015625-16015625 on Build GRCh38
rs779785456 Chr.1: 16015729-16015729 on Build GRCh38
rs866502789 Chr.1: 16015736-16015736 on Build GRCh38
rs138193282 Chr.1: 16015731-16015731 on Build GRCh38
rs146609918 Chr.1: 16015629-16015629 on Build GRCh38
rs527794958 Chr.1: 16015628-16015628 on Build GRCh38
rs201960746 Chr.1: 16015549-16015549 on Build GRCh38
rs750392600 Chr.1: 16015771-16015771 on Build GRCh38
rs1004882925 Chr.1: 16015880-16015880 on Build GRCh38
rs763036096 Chr.1: 16015595-16015595 on Build GRCh38
rs767416122 Chr.1: 16015718-16015718 on Build GRCh38
rs557092587 Chr.1: 16015795-16015795 on Build GRCh38
rs376565113 Chr.1: 16015667-16015667 on Build GRCh38
rs764260228 Chr.1: 16015645-16015645 on Build GRCh38
rs1013518764 Chr.1: 16015964-16015964 on Build GRCh38
rs955563781 Chr.1: 16015613-16015613 on Build GRCh38
rs748328021 Chr.1: 16015782-16015782 on Build GRCh38
rs373272623 Chr.1: 16015802-16015802 on Build GRCh38
rs754084618 Chr.1: 16015636-16015636 on Build GRCh38
rs764753899 Chr.1: 16015548-16015548 on Build GRCh38
rs1018073187 Chr.1: 16015549-16015549 on Build GRCh38
rs140061259 Chr.1: 16015928-16015928 on Build GRCh38
rs748963379 Chr.1: 16015730-16015730 on Build GRCh38
rs775935351 Chr.1: 16015754-16015754 on Build GRCh38
rs752942574 Chr.1: 16015642-16015642 on Build GRCh38
rs559032563 Chr.1: 16015857-16015857 on Build GRCh38
rs538235428 Chr.1: 16015664-16015664 on Build GRCh38
rs116499170 Chr.1: 16015694-16015694 on Build GRCh38
rs541539410 Chr.1: 16015914-16015914 on Build GRCh38
rs761047248 Chr.1: 16015634-16015634 on Build GRCh38
rs945260249 Chr.1: 16015682-16015682 on Build GRCh38
rs963725204 Chr.1: 16015606-16015606 on Build GRCh38
rs774623194 Chr.1: 16015763-16015763 on Build GRCh38
rs747801731 Chr.1: 16015797-16015797 on Build GRCh38
rs929417989 Chr.1: 16015925-16015925 on Build GRCh38
rs1017300136 Chr.1: 16015585-16015585 on Build GRCh38
rs773017695 Chr.1: 16015541-16015541 on Build GRCh38
rs755240487 Chr.1: 16015680-16015695 on Build GRCh38
rs763032522 Chr.1: 16015700-16015700 on Build GRCh38
rs879299614 Chr.1: 16015663-16015663 on Build GRCh38
rs749779823 Chr.1: 16015872-16015872 on Build GRCh38
rs781288880 Chr.1: 16015777-16015777 on Build GRCh38
rs771587197 Chr.1: 16015741-16015741 on Build GRCh38
rs987840661 Chr.1: 16015935-16015935 on Build GRCh38
rs895133463 Chr.1: 16015944-16015944 on Build GRCh38
rs748908426 Chr.1: 16015669-16015669 on Build GRCh38
rs200012555 Chr.1: 16015800-16015801 on Build GRCh38
rs142826385 Chr.1: 16015608-16015608 on Build GRCh38
rs755945385 Chr.1: 16015727-16015727 on Build GRCh38
rs963524976 Chr.1: 16015895-16015895 on Build GRCh38
rs530970423 Chr.1: 16015651-16015651 on Build GRCh38
rs764292072 Chr.1: 16015755-16015755 on Build GRCh38
rs368018183 Chr.1: 16015748-16015748 on Build GRCh38
rs756400552 Chr.1: 16015838-16015838 on Build GRCh38
rs765611158 Chr.1: 16015639-16015639 on Build GRCh38
rs770361355 Chr.1: 16015806-16015806 on Build GRCh38
rs777365756 Chr.1: 16015801-16015801 on Build GRCh38
rs201512595 Chr.1: 16015592-16015592 on Build GRCh38
rs144366876 Chr.1: 16015615-16015615 on Build GRCh38
rs777263308 Chr.1: 16015691-16015691 on Build GRCh38
rs761759 Chr.1: 16015855-16015855 on Build GRCh38
rs757875336 Chr.1: 16015799-16015799 on Build GRCh38
rs545441833 Chr.1: 16015612-16015612 on Build GRCh38
rs1739840 Chr.1: 16015742-16015742 on Build GRCh38
rs187230349 Chr.1: 16015542-16015542 on Build GRCh38
rs761760 Chr.1: 16015924-16015924 on Build GRCh38
rs561208093 Chr.1: 16015923-16015923 on Build GRCh38
rs577211747 Chr.1: 16015837-16015837 on Build GRCh38
rs150618710 Chr.1: 16015661-16015661 on Build GRCh38
rs532622086 Chr.1: 16015697-16015697 on Build GRCh38
rs778623045 Chr.1: 16015733-16015733 on Build GRCh38
rs114208608 Chr.1: 16015627-16015627 on Build GRCh38
rs568280829 Chr.1: 16015770-16015770 on Build GRCh38
rs766622342 Chr.1: 16015574-16015574 on Build GRCh38
rs191680206 Chr.1: 16015866-16015866 on Build GRCh38
rs1037165714 Chr.1: 16015887-16015887 on Build GRCh38
rs754235385 Chr.1: 16015544-16015550 on Build GRCh38
rs1038686137 Chr.1: 16015692-16015692 on Build GRCh38
rs371599498 Chr.1: 16015769-16015769 on Build GRCh38
rs759972261 Chr.1: 16015638-16015638 on Build GRCh38
rs561575565 Chr.1: 16015644-16015644 on Build GRCh38
rs755193962 Chr.1: 16015594-16015594 on Build GRCh38
rs373135452 Chr.1: 16015635-16015635 on Build GRCh38
rs761248997 Chr.1: 16015686-16015686 on Build GRCh38
rs753698681 Chr.1: 16015780-16015780 on Build GRCh38
rs200248116 Chr.1: 16015616-16015616 on Build GRCh38
rs750883423 Chr.1: 16015560-16015560 on Build GRCh38
rs771406607 Chr.1: 16015688-16015688 on Build GRCh38
rs779217024 Chr.1: 16015752-16015754 on Build GRCh38
rs374227503 Chr.1: 16015685-16015685 on Build GRCh38
rs199601455 Chr.1: 16015725-16015725 on Build GRCh38
rs973616969 Chr.1: 16015922-16015922 on Build GRCh38
rs762958256 Chr.1: 16015699-16015699 on Build GRCh38
rs1024651229 Chr.1: 16015859-16015859 on Build GRCh38

View Full Product Details