Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16016922-16017427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPB7
Entrez Gene ID
27129
Gene Name
heat shock protein family B (small) member 7
Gene Aliases
cvHSP
UniGene
Hs.502612
Gene Chromosome Location
Chr.1: 16014028-16018796 on Build GRCh38
Transcripts
NM_014424.4 XM_011541249.1 XM_011541250.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5797286 c.200-9C>T p.? Chr.1: 16017231-16017231 on Build GRCh38
5682947 c.268G>T p.A90S Chr.1: 16017154-16017154 on Build GRCh38
4399824 c.254C>T p.A85V Chr.1: 16017153-16017153 on Build GRCh38
4418327 c.207C>T p.P69P Chr.1: 16017200-16017200 on Build GRCh38
6682683 c.200-9A>G p.? Chr.1: 16017216-16017216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542498582 Chr.1: 16016982-16016982 on Build GRCh38
rs776461388 Chr.1: 16017173-16017173 on Build GRCh38
rs180869096 Chr.1: 16017027-16017027 on Build GRCh38
rs765038472 Chr.1: 16017085-16017085 on Build GRCh38
rs1050437334 Chr.1: 16017011-16017011 on Build GRCh38
rs990635855 Chr.1: 16017241-16017241 on Build GRCh38
rs1054755646 Chr.1: 16017315-16017315 on Build GRCh38
rs774178537 Chr.1: 16017271-16017271 on Build GRCh38
rs374305831 Chr.1: 16017005-16017005 on Build GRCh38
rs762840710 Chr.1: 16017202-16017202 on Build GRCh38
rs573469611 Chr.1: 16016959-16016959 on Build GRCh38
rs553330144 Chr.1: 16017331-16017331 on Build GRCh38
rs188639876 Chr.1: 16017295-16017295 on Build GRCh38
rs1739845 Chr.1: 16017320-16017320 on Build GRCh38
rs759289614 Chr.1: 16017071-16017071 on Build GRCh38
rs759465082 Chr.1: 16017180-16017180 on Build GRCh38
rs576920124 Chr.1: 16017236-16017236 on Build GRCh38
rs561574223 Chr.1: 16017008-16017008 on Build GRCh38
rs780759481 Chr.1: 16017119-16017119 on Build GRCh38
rs568400360 Chr.1: 16017261-16017261 on Build GRCh38
rs773219339 Chr.1: 16017057-16017057 on Build GRCh38
rs1763596 Chr.1: 16017340-16017340 on Build GRCh38
rs775278737 Chr.1: 16017262-16017262 on Build GRCh38
rs1023938484 Chr.1: 16017127-16017127 on Build GRCh38
rs766361550 Chr.1: 16017060-16017060 on Build GRCh38
rs748568896 Chr.1: 16017231-16017231 on Build GRCh38
rs764951901 Chr.1: 16017188-16017188 on Build GRCh38
rs199825788 Chr.1: 16017051-16017051 on Build GRCh38
rs756816628 Chr.1: 16017113-16017113 on Build GRCh38
rs774122126 Chr.1: 16017255-16017255 on Build GRCh38
rs148490812 Chr.1: 16017145-16017145 on Build GRCh38
rs575810260 Chr.1: 16017397-16017397 on Build GRCh38
rs369454490 Chr.1: 16017234-16017234 on Build GRCh38
rs752303086 Chr.1: 16017091-16017091 on Build GRCh38
rs906904469 Chr.1: 16017319-16017319 on Build GRCh38
rs769045519 Chr.1: 16017032-16017032 on Build GRCh38
rs747190986 Chr.1: 16017245-16017245 on Build GRCh38
rs529221395 Chr.1: 16017229-16017229 on Build GRCh38
rs931997199 Chr.1: 16016985-16016985 on Build GRCh38
rs1012627936 Chr.1: 16017291-16017291 on Build GRCh38
rs773407818 Chr.1: 16017161-16017161 on Build GRCh38
rs890352029 Chr.1: 16017039-16017039 on Build GRCh38
rs780713650 Chr.1: 16017218-16017218 on Build GRCh38
rs777423229 Chr.1: 16017096-16017096 on Build GRCh38
rs143274675 Chr.1: 16017204-16017204 on Build GRCh38
rs774478311 Chr.1: 16017048-16017048 on Build GRCh38
rs374526628 Chr.1: 16017049-16017049 on Build GRCh38
rs893287513 Chr.1: 16017316-16017316 on Build GRCh38
rs907529439 Chr.1: 16016999-16017001 on Build GRCh38
rs920330494 Chr.1: 16016971-16016971 on Build GRCh38
rs867387283 Chr.1: 16017066-16017066 on Build GRCh38
rs1048899244 Chr.1: 16017274-16017274 on Build GRCh38
rs751184800 Chr.1: 16017097-16017097 on Build GRCh38
rs142837502 Chr.1: 16017112-16017112 on Build GRCh38
rs747003067 Chr.1: 16017200-16017200 on Build GRCh38
rs533029438 Chr.1: 16017092-16017092 on Build GRCh38
rs530504261 Chr.1: 16017023-16017023 on Build GRCh38
rs556009404 Chr.1: 16017386-16017386 on Build GRCh38
rs777805543 Chr.1: 16017143-16017144 on Build GRCh38
rs189137394 Chr.1: 16016953-16016953 on Build GRCh38
rs752467360 Chr.1: 16017200-16017200 on Build GRCh38
rs760792798 Chr.1: 16017166-16017166 on Build GRCh38
rs753791155 Chr.1: 16017063-16017063 on Build GRCh38
rs150513998 Chr.1: 16017199-16017199 on Build GRCh38
rs975737932 Chr.1: 16017289-16017289 on Build GRCh38
rs867325449 Chr.1: 16017402-16017402 on Build GRCh38
rs779290288 Chr.1: 16017152-16017152 on Build GRCh38
rs763953969 Chr.1: 16017265-16017265 on Build GRCh38
rs751239804 Chr.1: 16017207-16017207 on Build GRCh38
rs771017698 Chr.1: 16017246-16017246 on Build GRCh38
rs375685531 Chr.1: 16017167-16017167 on Build GRCh38
rs570615500 Chr.1: 16017318-16017318 on Build GRCh38
rs760739380 Chr.1: 16017059-16017059 on Build GRCh38
rs546817616 Chr.1: 16017146-16017146 on Build GRCh38
rs184952320 Chr.1: 16017260-16017260 on Build GRCh38
rs145128881 Chr.1: 16017282-16017282 on Build GRCh38
rs1010736625 Chr.1: 16017198-16017198 on Build GRCh38
rs201407694 Chr.1: 16017212-16017212 on Build GRCh38
rs371936755 Chr.1: 16017252-16017252 on Build GRCh38
rs776361406 Chr.1: 16017262-16017262 on Build GRCh38
rs756871586 Chr.1: 16017215-16017215 on Build GRCh38
rs532763034 Chr.1: 16017382-16017382 on Build GRCh38
rs770894025 Chr.1: 16017260-16017264 on Build GRCh38
rs989620798 Chr.1: 16017395-16017395 on Build GRCh38
rs897697327 Chr.1: 16017050-16017050 on Build GRCh38
rs983098656 Chr.1: 16016986-16016986 on Build GRCh38
rs754323999 Chr.1: 16017225-16017225 on Build GRCh38
rs372898200 Chr.1: 16017158-16017158 on Build GRCh38
rs375905806 Chr.1: 16017230-16017230 on Build GRCh38

View Full Product Details