Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19217229-19217772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC1
Entrez Gene ID
23065
Gene Name
ER membrane protein complex subunit 1
Gene Aliases
CAVIPMR, KIAA0090
UniGene
Hs.439200
Gene Chromosome Location
Chr.1: 19215664-19251559 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 NR_135114.1 NR_135115.1

Gene Symbol
LOC101927895
Entrez Gene ID
101927895
Gene Name
uncharacterized LOC101927895
Gene Aliases
-
UniGene
Hs.660534
Gene Chromosome Location
Chr.1: 19210386-19240704 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 NR_135114.1 NR_135115.1

SNP Detail

SNP ID Chromosome Location
rs995767782 Chr.1: 19217426-19217426 on Build GRCh38
rs1021079258 Chr.1: 19217368-19217368 on Build GRCh38
rs183872781 Chr.1: 19217557-19217557 on Build GRCh38
rs551009087 Chr.1: 19217714-19217714 on Build GRCh38
rs571521086 Chr.1: 19217679-19217679 on Build GRCh38
rs929579070 Chr.1: 19217544-19217544 on Build GRCh38
rs893397106 Chr.1: 19217357-19217357 on Build GRCh38
rs983716580 Chr.1: 19217552-19217552 on Build GRCh38
rs1039023548 Chr.1: 19217628-19217628 on Build GRCh38
rs74928885 Chr.1: 19217518-19217518 on Build GRCh38
rs749751666 Chr.1: 19217275-19217275 on Build GRCh38
rs565125403 Chr.1: 19217384-19217384 on Build GRCh38
rs530536140 Chr.1: 19217459-19217459 on Build GRCh38
rs576876637 Chr.1: 19217472-19217472 on Build GRCh38
rs911917591 Chr.1: 19217588-19217589 on Build GRCh38
rs982751821 Chr.1: 19217469-19217469 on Build GRCh38
rs1032151722 Chr.1: 19217321-19217321 on Build GRCh38
rs1041985843 Chr.1: 19217381-19217381 on Build GRCh38
rs973984210 Chr.1: 19217489-19217489 on Build GRCh38
rs1009479440 Chr.1: 19217339-19217339 on Build GRCh38
rs944674456 Chr.1: 19217606-19217606 on Build GRCh38
rs199647646 Chr.1: 19217422-19217422 on Build GRCh38
rs796121546 Chr.1: 19217332-19217332 on Build GRCh38
rs78131788 Chr.1: 19217529-19217529 on Build GRCh38
rs140638343 Chr.1: 19217394-19217394 on Build GRCh38
rs946205498 Chr.1: 19217702-19217702 on Build GRCh38
rs532265090 Chr.1: 19217705-19217705 on Build GRCh38
rs377373719 Chr.1: 19217543-19217543 on Build GRCh38
rs549484309 Chr.1: 19217516-19217517 on Build GRCh38
rs767756935 Chr.1: 19217561-19217561 on Build GRCh38
rs946286182 Chr.1: 19217377-19217377 on Build GRCh38
rs997096906 Chr.1: 19217263-19217263 on Build GRCh38
rs1009922456 Chr.1: 19217713-19217713 on Build GRCh38
rs187946652 Chr.1: 19217635-19217635 on Build GRCh38
rs914780873 Chr.1: 19217684-19217684 on Build GRCh38
rs762041270 Chr.1: 19217410-19217410 on Build GRCh38
rs921024781 Chr.1: 19217594-19217594 on Build GRCh38
rs754499796 Chr.1: 19217589-19217589 on Build GRCh38
rs769287472 Chr.1: 19217345-19217345 on Build GRCh38
rs936060979 Chr.1: 19217664-19217664 on Build GRCh38
rs1042017068 Chr.1: 19217723-19217723 on Build GRCh38
rs1054600691 Chr.1: 19217340-19217340 on Build GRCh38
rs546624325 Chr.1: 19217675-19217675 on Build GRCh38
rs951380947 Chr.1: 19217458-19217458 on Build GRCh38
rs562904939 Chr.1: 19217438-19217438 on Build GRCh38
rs981856744 Chr.1: 19217409-19217409 on Build GRCh38
rs907243300 Chr.1: 19217389-19217389 on Build GRCh38
rs549423968 Chr.1: 19217663-19217663 on Build GRCh38
rs1034464134 Chr.1: 19217395-19217395 on Build GRCh38

View Full Product Details