Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19666150-19666575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR6
Entrez Gene ID
3362
Gene Name
5-hydroxytryptamine receptor 6
Gene Aliases
5-HT6, 5-HT6R
UniGene
Hs.22180
Gene Chromosome Location
Chr.1: 19665287-19680966 on Build GRCh38
Transcripts
NM_000871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6683046 c.514G>A p.A172T Chr.1: 19666267-19666267 on Build GRCh38
5968163 c.646C>A p.R216S Chr.1: 19666399-19666399 on Build GRCh38
4496778 c.482C>T p.P161L Chr.1: 19666235-19666235 on Build GRCh38
6683044 c.576G>A p.A192A Chr.1: 19666329-19666329 on Build GRCh38
5966315 c.477delC p.L160fs*54 Chr.1: 19666230-19666230 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759632975 Chr.1: 19666174-19666174 on Build GRCh38
rs752331612 Chr.1: 19666332-19666332 on Build GRCh38
rs754391027 Chr.1: 19666301-19666301 on Build GRCh38
rs745945587 Chr.1: 19666389-19666389 on Build GRCh38
rs539602607 Chr.1: 19666458-19666458 on Build GRCh38
rs1047123816 Chr.1: 19666288-19666288 on Build GRCh38
rs781006812 Chr.1: 19666331-19666331 on Build GRCh38
rs200720547 Chr.1: 19666242-19666242 on Build GRCh38
rs199974894 Chr.1: 19666275-19666275 on Build GRCh38
rs920123886 Chr.1: 19666523-19666523 on Build GRCh38
rs139531394 Chr.1: 19666200-19666200 on Build GRCh38
rs756004302 Chr.1: 19666356-19666356 on Build GRCh38
rs756669049 Chr.1: 19666186-19666186 on Build GRCh38
rs6660215 Chr.1: 19666296-19666296 on Build GRCh38
rs779381409 Chr.1: 19666212-19666212 on Build GRCh38
rs143672494 Chr.1: 19666321-19666321 on Build GRCh38
rs147809788 Chr.1: 19666492-19666492 on Build GRCh38
rs763506070 Chr.1: 19666518-19666518 on Build GRCh38
rs899529365 Chr.1: 19666521-19666521 on Build GRCh38
rs750886919 Chr.1: 19666432-19666432 on Build GRCh38
rs141808974 Chr.1: 19666442-19666442 on Build GRCh38
rs772070423 Chr.1: 19666399-19666399 on Build GRCh38
rs773623394 Chr.1: 19666515-19666515 on Build GRCh38
rs201695962 Chr.1: 19666180-19666180 on Build GRCh38
rs529213055 Chr.1: 19666443-19666443 on Build GRCh38
rs373493742 Chr.1: 19666410-19666410 on Build GRCh38
rs150139857 Chr.1: 19666358-19666358 on Build GRCh38
rs565763365 Chr.1: 19666455-19666455 on Build GRCh38
rs547666149 Chr.1: 19666445-19666445 on Build GRCh38
rs778718497 Chr.1: 19666456-19666456 on Build GRCh38
rs200952794 Chr.1: 19666481-19666481 on Build GRCh38
rs1008180634 Chr.1: 19666320-19666320 on Build GRCh38
rs201084777 Chr.1: 19666219-19666219 on Build GRCh38
rs762203940 Chr.1: 19666317-19666317 on Build GRCh38
rs201637638 Chr.1: 19666400-19666400 on Build GRCh38
rs370200488 Chr.1: 19666255-19666255 on Build GRCh38
rs61285556 Chr.1: 19666461-19666461 on Build GRCh38
rs768753763 Chr.1: 19666493-19666493 on Build GRCh38
rs958964017 Chr.1: 19666495-19666495 on Build GRCh38
rs200171714 Chr.1: 19666411-19666411 on Build GRCh38
rs781201635 Chr.1: 19666224-19666224 on Build GRCh38
rs8192530 Chr.1: 19666201-19666201 on Build GRCh38
rs201076514 Chr.1: 19666414-19666414 on Build GRCh38
rs767096484 Chr.1: 19666434-19666434 on Build GRCh38
rs920140973 Chr.1: 19666171-19666171 on Build GRCh38
rs777857997 Chr.1: 19666266-19666266 on Build GRCh38
rs199498808 Chr.1: 19666221-19666221 on Build GRCh38
rs1038510199 Chr.1: 19666239-19666239 on Build GRCh38
rs746722809 Chr.1: 19666213-19666213 on Build GRCh38
rs772266550 Chr.1: 19666273-19666273 on Build GRCh38
rs757957479 Chr.1: 19666209-19666209 on Build GRCh38
rs749144847 Chr.1: 19666267-19666267 on Build GRCh38
rs779119468 Chr.1: 19666379-19666379 on Build GRCh38
rs775629025 Chr.1: 19666278-19666278 on Build GRCh38
rs56027618 Chr.1: 19666215-19666215 on Build GRCh38
rs201559988 Chr.1: 19666270-19666270 on Build GRCh38
rs767922017 Chr.1: 19666176-19666176 on Build GRCh38
rs545347633 Chr.1: 19666329-19666329 on Build GRCh38
rs748475745 Chr.1: 19666508-19666508 on Build GRCh38
rs991627636 Chr.1: 19666460-19666460 on Build GRCh38
rs770746430 Chr.1: 19666363-19666363 on Build GRCh38
rs759122261 Chr.1: 19666433-19666433 on Build GRCh38
rs199909804 Chr.1: 19666435-19666435 on Build GRCh38
rs774275533 Chr.1: 19666271-19666271 on Build GRCh38
rs982966602 Chr.1: 19666489-19666490 on Build GRCh38
rs931510845 Chr.1: 19666474-19666474 on Build GRCh38
rs769655924 Chr.1: 19666262-19666262 on Build GRCh38
rs761027992 Chr.1: 19666294-19666294 on Build GRCh38
rs776828771 Chr.1: 19666413-19666413 on Build GRCh38
rs959036260 Chr.1: 19666438-19666438 on Build GRCh38
rs750682152 Chr.1: 19666480-19666480 on Build GRCh38
rs199795418 Chr.1: 19666206-19666206 on Build GRCh38
rs777110417 Chr.1: 19666496-19666496 on Build GRCh38
rs886762585 Chr.1: 19666193-19666193 on Build GRCh38
rs770131583 Chr.1: 19666514-19666514 on Build GRCh38
rs202014642 Chr.1: 19666429-19666429 on Build GRCh38
rs201579153 Chr.1: 19666181-19666181 on Build GRCh38

View Full Product Details