Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19678310-19678812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR6
Entrez Gene ID
3362
Gene Name
5-hydroxytryptamine receptor 6
Gene Aliases
5-HT6, 5-HT6R
UniGene
Hs.22180
Gene Chromosome Location
Chr.1: 19665287-19680966 on Build GRCh38
Transcripts
NM_000871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3789298 c.730C>A p.R244S Chr.1: 19678582-19678582 on Build GRCh38
4557654 c.737G>A p.G246E Chr.1: 19678589-19678589 on Build GRCh38
5379104 c.738G>A p.G246G Chr.1: 19678590-19678590 on Build GRCh38
2126081 c.746C>T p.S249F Chr.1: 19678598-19678598 on Build GRCh38
6395656 c.841T>C p.W281R Chr.1: 19678693-19678693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755046439 Chr.1: 19678613-19678613 on Build GRCh38
rs781277632 Chr.1: 19678549-19678549 on Build GRCh38
rs766123991 Chr.1: 19678703-19678703 on Build GRCh38
rs749659592 Chr.1: 19678627-19678627 on Build GRCh38
rs564362951 Chr.1: 19678656-19678656 on Build GRCh38
rs1010410900 Chr.1: 19678436-19678436 on Build GRCh38
rs201614623 Chr.1: 19678772-19678772 on Build GRCh38
rs200160454 Chr.1: 19678652-19678652 on Build GRCh38
rs995960608 Chr.1: 19678773-19678773 on Build GRCh38
rs200027010 Chr.1: 19678599-19678599 on Build GRCh38
rs995538237 Chr.1: 19678567-19678567 on Build GRCh38
rs903326651 Chr.1: 19678790-19678790 on Build GRCh38
rs760780334 Chr.1: 19678552-19678555 on Build GRCh38
rs200301309 Chr.1: 19678735-19678735 on Build GRCh38
rs566093197 Chr.1: 19678750-19678750 on Build GRCh38
rs991219572 Chr.1: 19678342-19678342 on Build GRCh38
rs543949088 Chr.1: 19678453-19678453 on Build GRCh38
rs200538675 Chr.1: 19678540-19678540 on Build GRCh38
rs200837456 Chr.1: 19678620-19678620 on Build GRCh38
rs576582739 Chr.1: 19678344-19678344 on Build GRCh38
rs200889844 Chr.1: 19678741-19678741 on Build GRCh38
rs531418003 Chr.1: 19678662-19678662 on Build GRCh38
rs200451070 Chr.1: 19678522-19678522 on Build GRCh38
rs772884793 Chr.1: 19678763-19678763 on Build GRCh38
rs759495828 Chr.1: 19678714-19678714 on Build GRCh38
rs368096897 Chr.1: 19678564-19678564 on Build GRCh38
rs199506394 Chr.1: 19678609-19678609 on Build GRCh38
rs750214863 Chr.1: 19678590-19678590 on Build GRCh38
rs200228144 Chr.1: 19678698-19678698 on Build GRCh38
rs1056343558 Chr.1: 19678746-19678746 on Build GRCh38
rs201929877 Chr.1: 19678585-19678585 on Build GRCh38
rs576634780 Chr.1: 19678781-19678781 on Build GRCh38
rs778157576 Chr.1: 19678732-19678732 on Build GRCh38
rs753460824 Chr.1: 19678528-19678528 on Build GRCh38
rs201106730 Chr.1: 19678736-19678736 on Build GRCh38
rs781284440 Chr.1: 19678616-19678616 on Build GRCh38
rs141227356 Chr.1: 19678659-19678659 on Build GRCh38
rs140928691 Chr.1: 19678726-19678726 on Build GRCh38
rs756332636 Chr.1: 19678622-19678622 on Build GRCh38
rs201083688 Chr.1: 19678658-19678658 on Build GRCh38
rs762742250 Chr.1: 19678701-19678701 on Build GRCh38
rs769420968 Chr.1: 19678692-19678692 on Build GRCh38
rs1003443679 Chr.1: 19678791-19678791 on Build GRCh38
rs758385987 Chr.1: 19678594-19678594 on Build GRCh38
rs775200937 Chr.1: 19678580-19678580 on Build GRCh38
rs564225882 Chr.1: 19678371-19678371 on Build GRCh38
rs201632507 Chr.1: 19678640-19678640 on Build GRCh38
rs976664255 Chr.1: 19678731-19678731 on Build GRCh38
rs761937245 Chr.1: 19678412-19678412 on Build GRCh38
rs541504469 Chr.1: 19678559-19678559 on Build GRCh38
rs201832081 Chr.1: 19678721-19678721 on Build GRCh38
rs1050484006 Chr.1: 19678515-19678515 on Build GRCh38
rs369367126 Chr.1: 19678463-19678463 on Build GRCh38
rs767537019 Chr.1: 19678720-19678720 on Build GRCh38
rs10917511 Chr.1: 19678755-19678755 on Build GRCh38
rs201607639 Chr.1: 19678708-19678708 on Build GRCh38
rs756351555 Chr.1: 19678730-19678730 on Build GRCh38
rs764257637 Chr.1: 19678658-19678659 on Build GRCh38
rs201508617 Chr.1: 19678523-19678523 on Build GRCh38
rs201017496 Chr.1: 19678705-19678705 on Build GRCh38
rs199795719 Chr.1: 19678578-19678578 on Build GRCh38
rs893404727 Chr.1: 19678407-19678407 on Build GRCh38
rs576058709 Chr.1: 19678377-19678377 on Build GRCh38
rs199943624 Chr.1: 19678629-19678629 on Build GRCh38
rs772134041 Chr.1: 19678475-19678475 on Build GRCh38
rs10917510 Chr.1: 19678478-19678478 on Build GRCh38
rs908407285 Chr.1: 19678446-19678446 on Build GRCh38
rs183347741 Chr.1: 19678552-19678552 on Build GRCh38
rs764973069 Chr.1: 19678608-19678608 on Build GRCh38
rs756871612 Chr.1: 19678767-19678767 on Build GRCh38
rs1041380296 Chr.1: 19678469-19678469 on Build GRCh38
rs780625049 Chr.1: 19678749-19678749 on Build GRCh38
rs201938771 Chr.1: 19678539-19678539 on Build GRCh38
rs931984991 Chr.1: 19678488-19678488 on Build GRCh38
rs763643170 Chr.1: 19678526-19678526 on Build GRCh38
rs958617751 Chr.1: 19678337-19678337 on Build GRCh38
rs200222462 Chr.1: 19678547-19678547 on Build GRCh38
rs774427000 Chr.1: 19678396-19678396 on Build GRCh38
rs775522902 Chr.1: 19678528-19678528 on Build GRCh38
rs763617024 Chr.1: 19678583-19678583 on Build GRCh38
rs201526723 Chr.1: 19678747-19678747 on Build GRCh38
rs143893286 Chr.1: 19678623-19678623 on Build GRCh38
rs368052638 Chr.1: 19678603-19678603 on Build GRCh38
rs745947620 Chr.1: 19678605-19678605 on Build GRCh38
rs200971544 Chr.1: 19678635-19678635 on Build GRCh38
rs372249233 Chr.1: 19678764-19678764 on Build GRCh38
rs757031687 Chr.1: 19678529-19678529 on Build GRCh38
rs191459000 Chr.1: 19678745-19678745 on Build GRCh38
rs558171153 Chr.1: 19678756-19678756 on Build GRCh38
rs201216539 Chr.1: 19678541-19678541 on Build GRCh38
rs764980768 Chr.1: 19678586-19678586 on Build GRCh38
rs139128448 Chr.1: 19678787-19678787 on Build GRCh38
rs199691155 Chr.1: 19678542-19678542 on Build GRCh38
rs61751019 Chr.1: 19678582-19678582 on Build GRCh38
rs200527196 Chr.1: 19678766-19678766 on Build GRCh38
rs765028517 Chr.1: 19678533-19678533 on Build GRCh38
rs767188508 Chr.1: 19678516-19678516 on Build GRCh38
rs923259357 Chr.1: 19678487-19678487 on Build GRCh38
rs779394192 Chr.1: 19678739-19678739 on Build GRCh38
rs111809215 Chr.1: 19678562-19678562 on Build GRCh38
rs749856051 Chr.1: 19678563-19678563 on Build GRCh38

View Full Product Details