Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20749268-20749812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HP1BP3
Entrez Gene ID
50809
Gene Name
heterochromatin protein 1 binding protein 3
Gene Aliases
HP1-BP74, HP1BP74
UniGene
Hs.142442
Gene Chromosome Location
Chr.1: 20742677-20787809 on Build GRCh38
Transcripts
NM_016287.3 XM_005245875.4 XM_005245876.3 XM_005245877.4 XM_005245878.4 XM_005245879.4 XM_011541532.1 XM_011541533.1 XM_011541534.2 XM_017001393.1 XM_017001394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
903422 c.1130C>A p.S377Y Chr.1: 20749734-20749734 on Build GRCh38
6262893 c.1118C>A p.P373Q Chr.1: 20749746-20749746 on Build GRCh38
4027833 c.1099T>C p.Y367H Chr.1: 20749765-20749765 on Build GRCh38
6503633 c.1102G>A p.V368I Chr.1: 20749762-20749762 on Build GRCh38
4027834 c.1067C>T p.P356L Chr.1: 20749797-20749797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138845998 Chr.1: 20749693-20749693 on Build GRCh38
rs990226087 Chr.1: 20749374-20749374 on Build GRCh38
rs774794482 Chr.1: 20749748-20749748 on Build GRCh38
rs548174177 Chr.1: 20749674-20749674 on Build GRCh38
rs1049683889 Chr.1: 20749574-20749574 on Build GRCh38
rs991062082 Chr.1: 20749354-20749354 on Build GRCh38
rs182456159 Chr.1: 20749589-20749589 on Build GRCh38
rs529121478 Chr.1: 20749421-20749421 on Build GRCh38
rs147651581 Chr.1: 20749332-20749332 on Build GRCh38
rs372620705 Chr.1: 20749694-20749694 on Build GRCh38
rs368120650 Chr.1: 20749366-20749366 on Build GRCh38
rs933796989 Chr.1: 20749551-20749551 on Build GRCh38
rs990926383 Chr.1: 20749647-20749647 on Build GRCh38
rs368351627 Chr.1: 20749389-20749389 on Build GRCh38
rs186775002 Chr.1: 20749618-20749618 on Build GRCh38
rs372080446 Chr.1: 20749709-20749709 on Build GRCh38
rs145914118 Chr.1: 20749606-20749606 on Build GRCh38
rs551442913 Chr.1: 20749594-20749594 on Build GRCh38
rs996148550 Chr.1: 20749364-20749366 on Build GRCh38
rs182718346 Chr.1: 20749382-20749382 on Build GRCh38
rs557369989 Chr.1: 20749713-20749713 on Build GRCh38
rs604728 Chr.1: 20749665-20749665 on Build GRCh38
rs886527372 Chr.1: 20749340-20749340 on Build GRCh38
rs901670094 Chr.1: 20749358-20749361 on Build GRCh38
rs567327981 Chr.1: 20749620-20749620 on Build GRCh38
rs375430390 Chr.1: 20749484-20749484 on Build GRCh38
rs764934051 Chr.1: 20749790-20749790 on Build GRCh38
rs79500933 Chr.1: 20749375-20749375 on Build GRCh38
rs940807168 Chr.1: 20749496-20749496 on Build GRCh38
rs545295791 Chr.1: 20749711-20749711 on Build GRCh38
rs368869506 Chr.1: 20749770-20749770 on Build GRCh38
rs12729575 Chr.1: 20749781-20749781 on Build GRCh38
rs17837940 Chr.1: 20749293-20749293 on Build GRCh38
rs12129245 Chr.1: 20749500-20749500 on Build GRCh38
rs781011878 Chr.1: 20749705-20749705 on Build GRCh38
rs776883359 Chr.1: 20749349-20749351 on Build GRCh38
rs996420626 Chr.1: 20749526-20749526 on Build GRCh38
rs56356353 Chr.1: 20749416-20749416 on Build GRCh38
rs566012314 Chr.1: 20749562-20749562 on Build GRCh38
rs201021126 Chr.1: 20749525-20749526 on Build GRCh38
rs545657951 Chr.1: 20749456-20749456 on Build GRCh38
rs958446862 Chr.1: 20749636-20749636 on Build GRCh38
rs12140145 Chr.1: 20749366-20749366 on Build GRCh38
rs772493212 Chr.1: 20749757-20749757 on Build GRCh38
rs77289352 Chr.1: 20749376-20749376 on Build GRCh38
rs778532890 Chr.1: 20749362-20749363 on Build GRCh38
rs561383757 Chr.1: 20749419-20749419 on Build GRCh38
rs1018745891 Chr.1: 20749354-20749356 on Build GRCh38
rs764633349 Chr.1: 20749633-20749633 on Build GRCh38
rs67026332 Chr.1: 20749375-20749376 on Build GRCh38
rs191672093 Chr.1: 20749325-20749325 on Build GRCh38
rs534905851 Chr.1: 20749664-20749664 on Build GRCh38
rs544086638 Chr.1: 20749762-20749762 on Build GRCh38
rs913141138 Chr.1: 20749602-20749602 on Build GRCh38
rs1040417349 Chr.1: 20749503-20749503 on Build GRCh38
rs760069300 Chr.1: 20749756-20749756 on Build GRCh38
rs780070092 Chr.1: 20749685-20749685 on Build GRCh38
rs375085843 Chr.1: 20749714-20749714 on Build GRCh38
rs763674949 Chr.1: 20749709-20749717 on Build GRCh38
rs981177506 Chr.1: 20749402-20749402 on Build GRCh38
rs190295875 Chr.1: 20749483-20749483 on Build GRCh38
rs969689550 Chr.1: 20749351-20749351 on Build GRCh38
rs923360593 Chr.1: 20749703-20749703 on Build GRCh38
rs537010724 Chr.1: 20749617-20749617 on Build GRCh38
rs763970895 Chr.1: 20749362-20749362 on Build GRCh38
rs925680677 Chr.1: 20749361-20749361 on Build GRCh38
rs991119854 Chr.1: 20749414-20749414 on Build GRCh38
rs11377726 Chr.1: 20749361-20749362 on Build GRCh38
rs559642645 Chr.1: 20749326-20749326 on Build GRCh38
rs901029490 Chr.1: 20749505-20749505 on Build GRCh38
rs369176150 Chr.1: 20749707-20749707 on Build GRCh38
rs907852643 Chr.1: 20749491-20749491 on Build GRCh38
rs369095738 Chr.1: 20749445-20749445 on Build GRCh38
rs991395734 Chr.1: 20749659-20749659 on Build GRCh38
rs1022006263 Chr.1: 20749605-20749605 on Build GRCh38
rs187342476 Chr.1: 20749407-20749407 on Build GRCh38
rs960861353 Chr.1: 20749408-20749408 on Build GRCh38
rs1050231062 Chr.1: 20749553-20749553 on Build GRCh38
rs368876149 Chr.1: 20749678-20749678 on Build GRCh38
rs960918545 Chr.1: 20749629-20749629 on Build GRCh38
rs979852634 Chr.1: 20749356-20749356 on Build GRCh38
rs893061769 Chr.1: 20749577-20749577 on Build GRCh38
rs916803739 Chr.1: 20749397-20749397 on Build GRCh38

View Full Product Details