Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72661274-72661771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA6B
Entrez Gene ID
55889
Gene Name
golgin A6 family member B
Gene Aliases
GOLGA, GOLGA6D
UniGene
Hs.546408
Gene Chromosome Location
Chr.15: 72638291-72669604 on Build GRCh38
Transcripts
NM_018652.4 XM_017022406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1243286 c.626C>T p.A209V Chr.15: 72661325-72661325 on Build GRCh38
6142814 c.645G>C p.V215V Chr.15: 72661344-72661344 on Build GRCh38
6142817 c.622C>T p.R208W Chr.15: 72661321-72661321 on Build GRCh38
6204106 c.651+3G>A p.? Chr.15: 72661353-72661353 on Build GRCh38
6142815 c.631C>A p.L211M Chr.15: 72661330-72661330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755698726 Chr.15: 72661298-72661298 on Build GRCh38
rs773053523 Chr.15: 72661342-72661342 on Build GRCh38
rs931353869 Chr.15: 72661683-72661683 on Build GRCh38
rs751973779 Chr.15: 72661427-72661427 on Build GRCh38
rs766308619 Chr.15: 72661343-72661343 on Build GRCh38
rs879149494 Chr.15: 72661707-72661707 on Build GRCh38
rs879016315 Chr.15: 72661692-72661692 on Build GRCh38
rs778172950 Chr.15: 72661294-72661294 on Build GRCh38
rs2081561 Chr.15: 72661297-72661297 on Build GRCh38
rs199952103 Chr.15: 72661335-72661335 on Build GRCh38
rs2081560 Chr.15: 72661293-72661293 on Build GRCh38
rs201343526 Chr.15: 72661298-72661298 on Build GRCh38
rs543869566 Chr.15: 72661296-72661296 on Build GRCh38
rs769406181 Chr.15: 72661295-72661295 on Build GRCh38
rs202082690 Chr.15: 72661322-72661322 on Build GRCh38
rs758658172 Chr.15: 72661319-72661319 on Build GRCh38
rs750300398 Chr.15: 72661303-72661303 on Build GRCh38
rs753986986 Chr.15: 72661312-72661312 on Build GRCh38
rs765396353 Chr.15: 72661353-72661353 on Build GRCh38
rs778248868 Chr.15: 72661321-72661321 on Build GRCh38
rs765393719 Chr.15: 72661314-72661314 on Build GRCh38
rs879139914 Chr.15: 72661685-72661685 on Build GRCh38
rs565278904 Chr.15: 72661412-72661412 on Build GRCh38
rs758704717 Chr.15: 72661396-72661396 on Build GRCh38
rs761537901 Chr.15: 72661300-72661300 on Build GRCh38
rs760659638 Chr.15: 72661306-72661306 on Build GRCh38
rs201629755 Chr.15: 72661560-72661560 on Build GRCh38
rs773909417 Chr.15: 72661336-72661336 on Build GRCh38
rs532504909 Chr.15: 72661414-72661414 on Build GRCh38
rs200737156 Chr.15: 72661351-72661351 on Build GRCh38
rs755127832 Chr.15: 72661313-72661313 on Build GRCh38
rs201808410 Chr.15: 72661326-72661326 on Build GRCh38
rs878880894 Chr.15: 72661688-72661688 on Build GRCh38
rs771745869 Chr.15: 72661341-72661341 on Build GRCh38
rs776636113 Chr.15: 72661350-72661350 on Build GRCh38
rs779636196 Chr.15: 72661325-72661325 on Build GRCh38
rs774135851 Chr.15: 72661299-72661299 on Build GRCh38
rs766433296 Chr.15: 72661310-72661310 on Build GRCh38
rs753064173 Chr.15: 72661318-72661318 on Build GRCh38
rs757778626 Chr.15: 72661428-72661428 on Build GRCh38
rs781769605 Chr.15: 72661498-72661498 on Build GRCh38
rs752839031 Chr.15: 72661361-72661361 on Build GRCh38

View Full Product Details