Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81225207-81225706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL16
Entrez Gene ID
3603
Gene Name
interleukin 16
Gene Aliases
LCF, NIL16, PRIL16, prIL-16
UniGene
Hs.459095
Gene Chromosome Location
Chr.15: 81181844-81312763 on Build GRCh38
Transcripts
NM_001172128.1 NM_172217.3 XM_005254342.3 XM_011521519.2 XM_011521520.2 XM_017022140.1 XM_017022141.1 XM_017022142.1 XM_017022143.1 XM_017022144.1 XM_017022145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3944281 c.273T>G p.N91K Chr.15: 81225672-81225672 on Build GRCh38
5173717 c.18C>T p.R6R Chr.15: 81225417-81225417 on Build GRCh38
4578543 c.270G>T p.G90G Chr.15: 81225669-81225669 on Build GRCh38
5350297 c.253C>G p.Q85E Chr.15: 81225652-81225652 on Build GRCh38
4878095 c.14G>T p.S5I Chr.15: 81225413-81225413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773559808 Chr.15: 81225367-81225367 on Build GRCh38
rs760626201 Chr.15: 81225530-81225530 on Build GRCh38
rs750669821 Chr.15: 81225444-81225444 on Build GRCh38
rs752252940 Chr.15: 81225641-81225641 on Build GRCh38
rs775697249 Chr.15: 81225597-81225597 on Build GRCh38
rs916779813 Chr.15: 81225236-81225236 on Build GRCh38
rs750324638 Chr.15: 81225418-81225418 on Build GRCh38
rs759686946 Chr.15: 81225407-81225407 on Build GRCh38
rs748423156 Chr.15: 81225679-81225679 on Build GRCh38
rs183847813 Chr.15: 81225431-81225431 on Build GRCh38
rs576209749 Chr.15: 81225417-81225417 on Build GRCh38
rs924078060 Chr.15: 81225531-81225531 on Build GRCh38
rs370711144 Chr.15: 81225649-81225649 on Build GRCh38
rs200842787 Chr.15: 81225671-81225671 on Build GRCh38
rs747432359 Chr.15: 81225556-81225556 on Build GRCh38
rs766454847 Chr.15: 81225642-81225642 on Build GRCh38
rs759598447 Chr.15: 81225393-81225393 on Build GRCh38
rs756134523 Chr.15: 81225422-81225422 on Build GRCh38
rs758294731 Chr.15: 81225676-81225676 on Build GRCh38
rs767203646 Chr.15: 81225503-81225503 on Build GRCh38
rs770980958 Chr.15: 81225369-81225369 on Build GRCh38
rs753351248 Chr.15: 81225545-81225545 on Build GRCh38
rs530151021 Chr.15: 81225625-81225625 on Build GRCh38
rs767690788 Chr.15: 81225292-81225292 on Build GRCh38
rs752276629 Chr.15: 81225551-81225551 on Build GRCh38
rs573604337 Chr.15: 81225234-81225234 on Build GRCh38
rs772584232 Chr.15: 81225347-81225347 on Build GRCh38
rs765933501 Chr.15: 81225425-81225425 on Build GRCh38
rs781011669 Chr.15: 81225252-81225252 on Build GRCh38
rs923711014 Chr.15: 81225313-81225313 on Build GRCh38
rs757710856 Chr.15: 81225558-81225558 on Build GRCh38
rs374725926 Chr.15: 81225681-81225681 on Build GRCh38
rs979572700 Chr.15: 81225497-81225497 on Build GRCh38
rs776702646 Chr.15: 81225637-81225637 on Build GRCh38
rs755549812 Chr.15: 81225268-81225268 on Build GRCh38
rs199973594 Chr.15: 81225448-81225448 on Build GRCh38
rs922989632 Chr.15: 81225598-81225598 on Build GRCh38
rs781604818 Chr.15: 81225650-81225650 on Build GRCh38
rs778733686 Chr.15: 81225429-81225429 on Build GRCh38
rs191307785 Chr.15: 81225416-81225416 on Build GRCh38
rs776705538 Chr.15: 81225402-81225402 on Build GRCh38
rs376483266 Chr.15: 81225341-81225341 on Build GRCh38
rs768931011 Chr.15: 81225619-81225619 on Build GRCh38
rs964409785 Chr.15: 81225294-81225294 on Build GRCh38
rs769862783 Chr.15: 81225591-81225591 on Build GRCh38
rs781648023 Chr.15: 81225572-81225572 on Build GRCh38
rs925368936 Chr.15: 81225360-81225360 on Build GRCh38
rs964294705 Chr.15: 81225576-81225576 on Build GRCh38
rs775546068 Chr.15: 81225468-81225468 on Build GRCh38
rs562794729 Chr.15: 81225646-81225646 on Build GRCh38
rs777933017 Chr.15: 81225553-81225553 on Build GRCh38
rs773073585 Chr.15: 81225506-81225506 on Build GRCh38
rs748935965 Chr.15: 81225301-81225301 on Build GRCh38
rs562915648 Chr.15: 81225283-81225283 on Build GRCh38
rs764866226 Chr.15: 81225550-81225550 on Build GRCh38
rs200683061 Chr.15: 81225340-81225340 on Build GRCh38
rs758682758 Chr.15: 81225432-81225432 on Build GRCh38
rs745968376 Chr.15: 81225450-81225450 on Build GRCh38
rs751259131 Chr.15: 81225648-81225648 on Build GRCh38
rs371174340 Chr.15: 81225677-81225677 on Build GRCh38
rs747412827 Chr.15: 81225440-81225440 on Build GRCh38
rs955647189 Chr.15: 81225228-81225228 on Build GRCh38
rs779357406 Chr.15: 81225338-81225338 on Build GRCh38
rs145164115 Chr.15: 81225465-81225465 on Build GRCh38
rs776652388 Chr.15: 81225449-81225449 on Build GRCh38
rs750918945 Chr.15: 81225654-81225654 on Build GRCh38
rs779777950 Chr.15: 81225280-81225280 on Build GRCh38
rs753406289 Chr.15: 81225426-81225426 on Build GRCh38
rs750346610 Chr.15: 81225301-81225301 on Build GRCh38
rs200742971 Chr.15: 81225626-81225626 on Build GRCh38
rs764590315 Chr.15: 81225640-81225640 on Build GRCh38
rs565192328 Chr.15: 81225435-81225435 on Build GRCh38
rs767618370 Chr.15: 81225615-81225616 on Build GRCh38
rs202125479 Chr.15: 81225662-81225662 on Build GRCh38
rs373015962 Chr.15: 81225494-81225494 on Build GRCh38
rs754641043 Chr.15: 81225427-81225427 on Build GRCh38
rs763828717 Chr.15: 81225647-81225647 on Build GRCh38
rs955553429 Chr.15: 81225486-81225486 on Build GRCh38
rs750426438 Chr.15: 81225257-81225257 on Build GRCh38
rs202011202 Chr.15: 81225415-81225415 on Build GRCh38
rs762170767 Chr.15: 81225235-81225235 on Build GRCh38
rs199723975 Chr.15: 81225631-81225631 on Build GRCh38
rs758078437 Chr.15: 81225552-81225552 on Build GRCh38
rs766454583 Chr.15: 81225541-81225541 on Build GRCh38
rs968249097 Chr.15: 81225493-81225493 on Build GRCh38
rs768441598 Chr.15: 81225310-81225310 on Build GRCh38
rs979543028 Chr.15: 81225295-81225295 on Build GRCh38
rs199906235 Chr.15: 81225599-81225599 on Build GRCh38
rs1044869104 Chr.15: 81225669-81225669 on Build GRCh38
rs868740491 Chr.15: 81225589-81225589 on Build GRCh38
rs761308358 Chr.15: 81225368-81225368 on Build GRCh38
rs201225720 Chr.15: 81225408-81225408 on Build GRCh38
rs759603341 Chr.15: 81225638-81225638 on Build GRCh38

View Full Product Details