Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81279475-81279985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL16
Entrez Gene ID
3603
Gene Name
interleukin 16
Gene Aliases
LCF, NIL16, PRIL16, prIL-16
UniGene
Hs.459095
Gene Chromosome Location
Chr.15: 81181844-81312763 on Build GRCh38
Transcripts
NM_001172128.1 NM_172217.3 XM_005254342.3 XM_011521519.2 XM_011521520.2 XM_017022140.1 XM_017022141.1 XM_017022142.1 XM_017022143.1 XM_017022144.1 XM_017022145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
296917 c.880C>T p.L294F Chr.15: 81279573-81279573 on Build GRCh38
5172909 c.1010A>G p.E337G Chr.15: 81279703-81279703 on Build GRCh38
6267262 c.971G>T p.S324I Chr.15: 81279664-81279664 on Build GRCh38
7117335 c.1029C>G p.I343M Chr.15: 81279722-81279722 on Build GRCh38
5136931 c.991A>T p.S331C Chr.15: 81279684-81279684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202091691 Chr.15: 81279712-81279712 on Build GRCh38
rs751857020 Chr.15: 81279751-81279751 on Build GRCh38
rs773079032 Chr.15: 81279619-81279619 on Build GRCh38
rs923560574 Chr.15: 81279871-81279871 on Build GRCh38
rs779889911 Chr.15: 81279756-81279756 on Build GRCh38
rs377290170 Chr.15: 81279608-81279608 on Build GRCh38
rs763196394 Chr.15: 81279739-81279739 on Build GRCh38
rs771540143 Chr.15: 81279516-81279516 on Build GRCh38
rs757057964 Chr.15: 81279597-81279597 on Build GRCh38
rs59298568 Chr.15: 81279508-81279508 on Build GRCh38
rs760127526 Chr.15: 81279728-81279728 on Build GRCh38
rs908282920 Chr.15: 81279845-81279845 on Build GRCh38
rs1036050917 Chr.15: 81279752-81279752 on Build GRCh38
rs201210766 Chr.15: 81279753-81279753 on Build GRCh38
rs931066075 Chr.15: 81279530-81279530 on Build GRCh38
rs184301414 Chr.15: 81279799-81279799 on Build GRCh38
rs761208055 Chr.15: 81279639-81279639 on Build GRCh38
rs933592419 Chr.15: 81279894-81279894 on Build GRCh38
rs754005697 Chr.15: 81279580-81279580 on Build GRCh38
rs373482725 Chr.15: 81279695-81279695 on Build GRCh38
rs756594873 Chr.15: 81279565-81279565 on Build GRCh38
rs1056603151 Chr.15: 81279575-81279575 on Build GRCh38
rs750206291 Chr.15: 81279672-81279672 on Build GRCh38
rs775015723 Chr.15: 81279605-81279605 on Build GRCh38
rs761191691 Chr.15: 81279587-81279587 on Build GRCh38
rs534940384 Chr.15: 81279956-81279956 on Build GRCh38
rs559778163 Chr.15: 81279725-81279725 on Build GRCh38
rs767837781 Chr.15: 81279548-81279548 on Build GRCh38
rs200383193 Chr.15: 81279821-81279821 on Build GRCh38
rs777277475 Chr.15: 81279727-81279727 on Build GRCh38
rs759957683 Chr.15: 81279941-81279941 on Build GRCh38
rs765353678 Chr.15: 81279729-81279729 on Build GRCh38
rs771003655 Chr.15: 81279656-81279656 on Build GRCh38
rs771648519 Chr.15: 81279786-81279786 on Build GRCh38
rs4076201 Chr.15: 81279962-81279962 on Build GRCh38
rs748445175 Chr.15: 81279588-81279588 on Build GRCh38
rs375892392 Chr.15: 81279640-81279640 on Build GRCh38
rs200719244 Chr.15: 81279649-81279649 on Build GRCh38
rs199981575 Chr.15: 81279555-81279555 on Build GRCh38
rs765753793 Chr.15: 81279960-81279960 on Build GRCh38
rs755984887 Chr.15: 81279692-81279692 on Build GRCh38
rs759681017 Chr.15: 81279642-81279642 on Build GRCh38
rs770267179 Chr.15: 81279598-81279598 on Build GRCh38
rs773455913 Chr.15: 81279636-81279636 on Build GRCh38
rs368469782 Chr.15: 81279611-81279611 on Build GRCh38
rs771388895 Chr.15: 81279726-81279726 on Build GRCh38
rs769288387 Chr.15: 81279507-81279507 on Build GRCh38
rs189634718 Chr.15: 81279875-81279875 on Build GRCh38
rs765461860 Chr.15: 81279648-81279648 on Build GRCh38
rs201714634 Chr.15: 81279696-81279696 on Build GRCh38
rs777152647 Chr.15: 81279816-81279816 on Build GRCh38
rs903638545 Chr.15: 81279851-81279851 on Build GRCh38
rs1007838497 Chr.15: 81279945-81279945 on Build GRCh38
rs1021693022 Chr.15: 81279665-81279665 on Build GRCh38
rs147320559 Chr.15: 81279886-81279886 on Build GRCh38
rs758550662 Chr.15: 81279650-81279650 on Build GRCh38
rs752848664 Chr.15: 81279731-81279731 on Build GRCh38
rs774039413 Chr.15: 81279541-81279541 on Build GRCh38
rs745749777 Chr.15: 81279603-81279603 on Build GRCh38
rs773580839 Chr.15: 81279719-81279719 on Build GRCh38
rs545808485 Chr.15: 81279713-81279713 on Build GRCh38
rs769711053 Chr.15: 81279607-81279621 on Build GRCh38
rs762134212 Chr.15: 81279546-81279546 on Build GRCh38
rs373067172 Chr.15: 81279747-81279747 on Build GRCh38
rs767867834 Chr.15: 81279626-81279626 on Build GRCh38
rs777234991 Chr.15: 81279732-81279732 on Build GRCh38
rs537866391 Chr.15: 81279604-81279604 on Build GRCh38
rs547007573 Chr.15: 81279904-81279904 on Build GRCh38
rs998683560 Chr.15: 81279723-81279723 on Build GRCh38
rs755206497 Chr.15: 81279584-81279584 on Build GRCh38
rs372350374 Chr.15: 81279569-81279569 on Build GRCh38
rs889137763 Chr.15: 81279911-81279911 on Build GRCh38
rs748773612 Chr.15: 81279607-81279607 on Build GRCh38
rs1051084511 Chr.15: 81279901-81279901 on Build GRCh38
rs997160222 Chr.15: 81279896-81279896 on Build GRCh38
rs201929052 Chr.15: 81279759-81279759 on Build GRCh38
rs780999465 Chr.15: 81279659-81279659 on Build GRCh38
rs753706846 Chr.15: 81279757-81279757 on Build GRCh38
rs983854873 Chr.15: 81279834-81279834 on Build GRCh38
rs565947582 Chr.15: 81279921-81279921 on Build GRCh38
rs761512344 Chr.15: 81279612-81279612 on Build GRCh38
rs747639609 Chr.15: 81279764-81279764 on Build GRCh38
rs889021848 Chr.15: 81279936-81279936 on Build GRCh38
rs149649915 Chr.15: 81279867-81279867 on Build GRCh38
rs763758350 Chr.15: 81279540-81279540 on Build GRCh38
rs940892152 Chr.15: 81279853-81279853 on Build GRCh38
rs762551203 Chr.15: 81279512-81279512 on Build GRCh38
rs777264798 Chr.15: 81279793-81279793 on Build GRCh38
rs561947527 Chr.15: 81279815-81279815 on Build GRCh38

View Full Product Details