Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83127302-83127807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDGFRP3
Entrez Gene ID
50810
Gene Name
hepatoma-derived growth factor, related protein 3
Gene Aliases
CGI-142, HDGF-2, HDGF2, HRP-3
UniGene
Hs.513954
Gene Chromosome Location
Chr.15: 83112738-83208018 on Build GRCh38
Transcripts
XM_006720554.3 NM_001144903.1 NM_023003.3 XM_011521679.2 XM_011521680.2 XM_017022320.1 XM_017022321.1 XM_017022322.1 XM_017022323.1 XM_017022324.1 XM_017022325.1 XM_017022326.1 XM_017022327.1 XM_017022328.1 XM_017022329.1 XM_017022330.1 XM_017022331.1

Gene Symbol
TM6SF1
Entrez Gene ID
53346
Gene Name
transmembrane 6 superfamily member 1
Gene Aliases
-
UniGene
Hs.513094
Gene Chromosome Location
Chr.15: 83107572-83145403 on Build GRCh38
Transcripts
XM_006720554.3 NM_001144903.1 NM_023003.3 XM_011521679.2 XM_011521680.2 XM_017022320.1 XM_017022321.1 XM_017022322.1 XM_017022323.1 XM_017022324.1 XM_017022325.1 XM_017022326.1 XM_017022327.1 XM_017022328.1 XM_017022329.1 XM_017022330.1 XM_017022331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6561821 c.809C>G p.A270G Chr.15: 83127365-83127365 on Build GRCh38
5805194 c.908G>A p.G303E Chr.15: 83127464-83127464 on Build GRCh38
4734047 c.854G>A p.G285D Chr.15: 83127410-83127410 on Build GRCh38
6432459 c.831T>A p.P277P Chr.15: 83127387-83127387 on Build GRCh38
6802635 c.863G>A p.G288D Chr.15: 83127410-83127410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752529131 Chr.15: 83127485-83127485 on Build GRCh38
rs763382900 Chr.15: 83127429-83127429 on Build GRCh38
rs769317988 Chr.15: 83127476-83127476 on Build GRCh38
rs1027655969 Chr.15: 83127602-83127602 on Build GRCh38
rs758148077 Chr.15: 83127516-83127516 on Build GRCh38
rs761505263 Chr.15: 83127418-83127427 on Build GRCh38
rs573527715 Chr.15: 83127355-83127355 on Build GRCh38
rs531127818 Chr.15: 83127647-83127647 on Build GRCh38
rs1019036802 Chr.15: 83127393-83127393 on Build GRCh38
rs770417835 Chr.15: 83127401-83127401 on Build GRCh38
rs761809468 Chr.15: 83127447-83127447 on Build GRCh38
rs771798887 Chr.15: 83127576-83127576 on Build GRCh38
rs879510096 Chr.15: 83127769-83127769 on Build GRCh38
rs994020681 Chr.15: 83127510-83127510 on Build GRCh38
rs920922821 Chr.15: 83127343-83127343 on Build GRCh38
rs766564557 Chr.15: 83127481-83127481 on Build GRCh38
rs1023721473 Chr.15: 83127743-83127743 on Build GRCh38
rs535305399 Chr.15: 83127748-83127748 on Build GRCh38
rs760510163 Chr.15: 83127711-83127711 on Build GRCh38
rs750599062 Chr.15: 83127350-83127350 on Build GRCh38
rs148691649 Chr.15: 83127416-83127416 on Build GRCh38
rs747924656 Chr.15: 83127364-83127364 on Build GRCh38
rs1053510731 Chr.15: 83127376-83127376 on Build GRCh38
rs965700427 Chr.15: 83127662-83127662 on Build GRCh38
rs756382718 Chr.15: 83127461-83127461 on Build GRCh38
rs767653249 Chr.15: 83127334-83127334 on Build GRCh38
rs1003794587 Chr.15: 83127456-83127456 on Build GRCh38
rs200315449 Chr.15: 83127444-83127444 on Build GRCh38
rs769557782 Chr.15: 83127408-83127408 on Build GRCh38
rs952555448 Chr.15: 83127628-83127628 on Build GRCh38
rs976685529 Chr.15: 83127663-83127663 on Build GRCh38
rs920940494 Chr.15: 83127685-83127685 on Build GRCh38
rs746464492 Chr.15: 83127774-83127774 on Build GRCh38
rs984074814 Chr.15: 83127725-83127725 on Build GRCh38
rs776527009 Chr.15: 83127365-83127366 on Build GRCh38
rs953712796 Chr.15: 83127733-83127733 on Build GRCh38
rs750494301 Chr.15: 83127458-83127458 on Build GRCh38
rs1015380695 Chr.15: 83127653-83127653 on Build GRCh38
rs758379606 Chr.15: 83127368-83127368 on Build GRCh38
rs769824986 Chr.15: 83127498-83127500 on Build GRCh38
rs776153603 Chr.15: 83127405-83127405 on Build GRCh38
rs551225510 Chr.15: 83127762-83127762 on Build GRCh38
rs768361112 Chr.15: 83127342-83127343 on Build GRCh38
rs746531547 Chr.15: 83127398-83127398 on Build GRCh38
rs562270944 Chr.15: 83127451-83127451 on Build GRCh38
rs747875705 Chr.15: 83127468-83127468 on Build GRCh38
rs893995778 Chr.15: 83127407-83127407 on Build GRCh38
rs778742801 Chr.15: 83127357-83127357 on Build GRCh38
rs764543192 Chr.15: 83127435-83127435 on Build GRCh38
rs534989796 Chr.15: 83127680-83127680 on Build GRCh38
rs550724033 Chr.15: 83127581-83127581 on Build GRCh38
rs112539541 Chr.15: 83127344-83127344 on Build GRCh38
rs777732938 Chr.15: 83127369-83127369 on Build GRCh38

View Full Product Details