Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11869225-11869737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSPT1
Entrez Gene ID
2935
Gene Name
G1 to S phase transition 1
Gene Aliases
551G9.2, ETF3A, GST1, eRF3a
UniGene
Hs.528780
Gene Chromosome Location
Chr.16: 11868128-11916662 on Build GRCh38
Transcripts
NM_001130006.1 NM_001130007.1 NM_002094.3

SNP Detail

SNP ID Chromosome Location
rs951346282 Chr.16: 11869313-11869313 on Build GRCh38
rs117726386 Chr.16: 11869510-11869510 on Build GRCh38
rs1034962662 Chr.16: 11869281-11869281 on Build GRCh38
rs935476845 Chr.16: 11869704-11869704 on Build GRCh38
rs941312538 Chr.16: 11869446-11869446 on Build GRCh38
rs780177409 Chr.16: 11869555-11869555 on Build GRCh38
rs1013556795 Chr.16: 11869328-11869328 on Build GRCh38
rs1031806767 Chr.16: 11869680-11869680 on Build GRCh38
rs142198868 Chr.16: 11869533-11869533 on Build GRCh38
rs187823788 Chr.16: 11869569-11869569 on Build GRCh38
rs554546647 Chr.16: 11869448-11869448 on Build GRCh38
rs956203075 Chr.16: 11869700-11869700 on Build GRCh38
rs780307875 Chr.16: 11869625-11869625 on Build GRCh38
rs772120992 Chr.16: 11869272-11869273 on Build GRCh38
rs33659 Chr.16: 11869504-11869504 on Build GRCh38
rs1050085257 Chr.16: 11869420-11869420 on Build GRCh38
rs761517393 Chr.16: 11869289-11869290 on Build GRCh38
rs35759493 Chr.16: 11869434-11869435 on Build GRCh38
rs1056230482 Chr.16: 11869548-11869548 on Build GRCh38
rs973340164 Chr.16: 11869613-11869613 on Build GRCh38
rs894777829 Chr.16: 11869560-11869560 on Build GRCh38
rs755892826 Chr.16: 11869331-11869331 on Build GRCh38
rs963357829 Chr.16: 11869601-11869601 on Build GRCh38
rs40311 Chr.16: 11869333-11869333 on Build GRCh38
rs773810638 Chr.16: 11869317-11869317 on Build GRCh38
rs773054783 Chr.16: 11869627-11869627 on Build GRCh38
rs559472436 Chr.16: 11869329-11869332 on Build GRCh38
rs909783279 Chr.16: 11869426-11869426 on Build GRCh38
rs932685149 Chr.16: 11869416-11869416 on Build GRCh38
rs988014072 Chr.16: 11869386-11869386 on Build GRCh38
rs182396836 Chr.16: 11869449-11869449 on Build GRCh38
rs763788083 Chr.16: 11869485-11869485 on Build GRCh38
rs145953219 Chr.16: 11869634-11869634 on Build GRCh38
rs548273026 Chr.16: 11869412-11869412 on Build GRCh38
rs752328499 Chr.16: 11869329-11869329 on Build GRCh38
rs765878921 Chr.16: 11869295-11869295 on Build GRCh38
rs921979225 Chr.16: 11869357-11869357 on Build GRCh38
rs1000489891 Chr.16: 11869633-11869633 on Build GRCh38
rs912084036 Chr.16: 11869659-11869659 on Build GRCh38
rs904917334 Chr.16: 11869514-11869514 on Build GRCh38
rs191016399 Chr.16: 11869363-11869363 on Build GRCh38
rs569600985 Chr.16: 11869370-11869370 on Build GRCh38
rs535634514 Chr.16: 11869596-11869596 on Build GRCh38
rs571198024 Chr.16: 11869518-11869518 on Build GRCh38
rs987552796 Chr.16: 11869642-11869642 on Build GRCh38
rs1003390906 Chr.16: 11869543-11869543 on Build GRCh38
rs935591208 Chr.16: 11869427-11869427 on Build GRCh38
rs752264496 Chr.16: 11869379-11869379 on Build GRCh38
rs956106864 Chr.16: 11869384-11869384 on Build GRCh38
rs1022405298 Chr.16: 11869626-11869626 on Build GRCh38
rs868093062 Chr.16: 11869695-11869695 on Build GRCh38
rs112592586 Chr.16: 11869654-11869654 on Build GRCh38
rs556844934 Chr.16: 11869562-11869562 on Build GRCh38
rs968949709 Chr.16: 11869629-11869629 on Build GRCh38
rs976806911 Chr.16: 11869706-11869706 on Build GRCh38
rs1003747879 Chr.16: 11869253-11869253 on Build GRCh38
rs976837941 Chr.16: 11869392-11869392 on Build GRCh38
rs907906686 Chr.16: 11869327-11869327 on Build GRCh38

View Full Product Details