Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21647042-21647552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF6
Entrez Gene ID
10261
Gene Name
immunoglobulin superfamily member 6
Gene Aliases
DORA
UniGene
Hs.530902
Gene Chromosome Location
Chr.16: 21641284-21652666 on Build GRCh38
Transcripts
NM_005849.3 NM_001077180.2 NM_001288659.1 NM_001288660.1 NM_016025.4 XM_017023269.1

Gene Symbol
METTL9
Entrez Gene ID
51108
Gene Name
methyltransferase like 9
Gene Aliases
CGI-81, DREV, DREV1, PAP1
UniGene
Hs.279583
Gene Chromosome Location
Chr.16: 21597221-21657473 on Build GRCh38
Transcripts
NM_005849.3 NM_001077180.2 NM_001288659.1 NM_001288660.1 NM_016025.4 XM_017023269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578797 c.112C>T p.L38L Chr.16: 21647448-21647448 on Build GRCh38
6684614 c.68-1G>T p.? Chr.16: 21647493-21647493 on Build GRCh38
1493574 c.411C>T p.T137T Chr.16: 21647149-21647149 on Build GRCh38
6555504 c.298C>A p.Q100K Chr.16: 21647262-21647262 on Build GRCh38
1376617 c.75G>A p.V25V Chr.16: 21647485-21647485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748194189 Chr.16: 21647350-21647350 on Build GRCh38
rs772985937 Chr.16: 21647075-21647075 on Build GRCh38
rs563502288 Chr.16: 21647297-21647297 on Build GRCh38
rs967018221 Chr.16: 21647316-21647316 on Build GRCh38
rs777054630 Chr.16: 21647117-21647117 on Build GRCh38
rs781448340 Chr.16: 21647349-21647349 on Build GRCh38
rs762179953 Chr.16: 21647198-21647198 on Build GRCh38
rs199824045 Chr.16: 21647512-21647512 on Build GRCh38
rs1019913195 Chr.16: 21647314-21647314 on Build GRCh38
rs780238868 Chr.16: 21647424-21647426 on Build GRCh38
rs1041774696 Chr.16: 21647122-21647122 on Build GRCh38
rs761553423 Chr.16: 21647466-21647466 on Build GRCh38
rs772492797 Chr.16: 21647518-21647518 on Build GRCh38
rs756317662 Chr.16: 21647425-21647425 on Build GRCh38
rs541756545 Chr.16: 21647253-21647253 on Build GRCh38
rs751033130 Chr.16: 21647137-21647137 on Build GRCh38
rs202168035 Chr.16: 21647247-21647247 on Build GRCh38
rs777956569 Chr.16: 21647490-21647490 on Build GRCh38
rs777993032 Chr.16: 21647437-21647437 on Build GRCh38
rs748247374 Chr.16: 21647424-21647424 on Build GRCh38
rs564145745 Chr.16: 21647392-21647392 on Build GRCh38
rs746090681 Chr.16: 21647513-21647513 on Build GRCh38
rs778571289 Chr.16: 21647312-21647312 on Build GRCh38
rs749276966 Chr.16: 21647505-21647505 on Build GRCh38
rs1454373 Chr.16: 21647068-21647068 on Build GRCh38
rs754528260 Chr.16: 21647467-21647470 on Build GRCh38
rs763582744 Chr.16: 21647402-21647402 on Build GRCh38
rs770877981 Chr.16: 21647094-21647094 on Build GRCh38
rs754450106 Chr.16: 21647145-21647145 on Build GRCh38
rs757530396 Chr.16: 21647509-21647509 on Build GRCh38
rs773508193 Chr.16: 21647379-21647379 on Build GRCh38
rs548553916 Chr.16: 21647489-21647489 on Build GRCh38
rs200635568 Chr.16: 21647417-21647417 on Build GRCh38
rs763636136 Chr.16: 21647463-21647463 on Build GRCh38
rs17851574 Chr.16: 21647339-21647339 on Build GRCh38
rs760122055 Chr.16: 21647398-21647398 on Build GRCh38
rs749263205 Chr.16: 21647081-21647081 on Build GRCh38
rs370573910 Chr.16: 21647391-21647391 on Build GRCh38
rs755785405 Chr.16: 21647274-21647274 on Build GRCh38
rs746178604 Chr.16: 21647453-21647453 on Build GRCh38
rs116192352 Chr.16: 21647421-21647421 on Build GRCh38
rs771034810 Chr.16: 21647445-21647445 on Build GRCh38
rs756961815 Chr.16: 21647362-21647362 on Build GRCh38
rs766807948 Chr.16: 21647237-21647237 on Build GRCh38
rs771885163 Chr.16: 21647346-21647346 on Build GRCh38
rs750798170 Chr.16: 21647127-21647127 on Build GRCh38
rs749455636 Chr.16: 21647386-21647386 on Build GRCh38
rs531915033 Chr.16: 21647139-21647139 on Build GRCh38
rs770779016 Chr.16: 21647169-21647169 on Build GRCh38
rs768774248 Chr.16: 21647104-21647104 on Build GRCh38
rs756831346 Chr.16: 21647406-21647406 on Build GRCh38
rs370477124 Chr.16: 21647311-21647311 on Build GRCh38
rs776035836 Chr.16: 21647457-21647457 on Build GRCh38
rs753647394 Chr.16: 21647299-21647299 on Build GRCh38
rs752410978 Chr.16: 21647159-21647159 on Build GRCh38
rs749563320 Chr.16: 21647443-21647443 on Build GRCh38
rs766865720 Chr.16: 21647135-21647135 on Build GRCh38
rs144603059 Chr.16: 21647287-21647287 on Build GRCh38
rs752179827 Chr.16: 21647258-21647258 on Build GRCh38
rs764761471 Chr.16: 21647474-21647474 on Build GRCh38
rs975254774 Chr.16: 21647465-21647465 on Build GRCh38
rs766260469 Chr.16: 21647482-21647482 on Build GRCh38
rs753594905 Chr.16: 21647405-21647405 on Build GRCh38
rs151150797 Chr.16: 21647481-21647481 on Build GRCh38
rs763573457 Chr.16: 21647129-21647129 on Build GRCh38
rs140742307 Chr.16: 21647123-21647123 on Build GRCh38
rs150845052 Chr.16: 21647389-21647389 on Build GRCh38
rs774800987 Chr.16: 21647446-21647446 on Build GRCh38
rs368474150 Chr.16: 21647464-21647464 on Build GRCh38
rs773739508 Chr.16: 21647228-21647228 on Build GRCh38
rs144045651 Chr.16: 21647168-21647168 on Build GRCh38
rs1454374 Chr.16: 21647317-21647317 on Build GRCh38
rs374014200 Chr.16: 21647397-21647397 on Build GRCh38
rs770095575 Chr.16: 21647209-21647209 on Build GRCh38
rs576841517 Chr.16: 21647182-21647182 on Build GRCh38
rs755732200 Chr.16: 21647161-21647161 on Build GRCh38
rs760743163 Chr.16: 21647102-21647102 on Build GRCh38
rs767963050 Chr.16: 21647393-21647393 on Build GRCh38
rs1027551065 Chr.16: 21647166-21647166 on Build GRCh38
rs199841207 Chr.16: 21647112-21647112 on Build GRCh38
rs189739425 Chr.16: 21647413-21647413 on Build GRCh38
rs376880564 Chr.16: 21647151-21647151 on Build GRCh38
rs767052856 Chr.16: 21647256-21647256 on Build GRCh38
rs747653210 Chr.16: 21647522-21647540 on Build GRCh38
rs1001697670 Chr.16: 21647239-21647239 on Build GRCh38
rs750232505 Chr.16: 21647477-21647477 on Build GRCh38
rs747420250 Chr.16: 21647467-21647468 on Build GRCh38
rs780824615 Chr.16: 21647517-21647519 on Build GRCh38
rs933924758 Chr.16: 21647146-21647146 on Build GRCh38
rs758754048 Chr.16: 21647153-21647155 on Build GRCh38
rs147257312 Chr.16: 21647177-21647177 on Build GRCh38
rs201381483 Chr.16: 21647455-21647455 on Build GRCh38
rs542957838 Chr.16: 21647176-21647176 on Build GRCh38
rs1034961459 Chr.16: 21647266-21647266 on Build GRCh38
rs373342802 Chr.16: 21647355-21647355 on Build GRCh38
rs552933355 Chr.16: 21647236-21647236 on Build GRCh38
rs186067099 Chr.16: 21647183-21647183 on Build GRCh38
rs775464475 Chr.16: 21647100-21647100 on Build GRCh38
rs970342564 Chr.16: 21647210-21647210 on Build GRCh38
rs761166964 Chr.16: 21647461-21647461 on Build GRCh38

View Full Product Details