Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74342547-74343054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF19
Entrez Gene ID
124602
Gene Name
kinesin family member 19
Gene Aliases
KIF19A
UniGene
Hs.372773
Gene Chromosome Location
Chr.17: 74326212-74355820 on Build GRCh38
Transcripts
NM_153209.3 XM_005257029.1 XM_005257030.1 XM_017024150.1 XM_017024151.1 XM_017024152.1 XM_017024153.1 XM_017024154.1 XM_017024155.1 XM_017024156.1 XM_017024157.1 XM_017024158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4835220 c.258G>C p.K86N Chr.17: 74342656-74342656 on Build GRCh38
3890326 c.310G>A p.G104S Chr.17: 74342708-74342708 on Build GRCh38
7191989 c.287A>G p.Y96C Chr.17: 74342685-74342685 on Build GRCh38
3989301 c.267C>G p.I89M Chr.17: 74342665-74342665 on Build GRCh38
148337 c.232-4G>A p.? Chr.17: 74342626-74342626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760997570 Chr.17: 74342744-74342744 on Build GRCh38
rs1035268222 Chr.17: 74342571-74342571 on Build GRCh38
rs768751891 Chr.17: 74342992-74342992 on Build GRCh38
rs754784199 Chr.17: 74343007-74343007 on Build GRCh38
rs774563945 Chr.17: 74342603-74342603 on Build GRCh38
rs911805266 Chr.17: 74342743-74342743 on Build GRCh38
rs867987095 Chr.17: 74342586-74342586 on Build GRCh38
rs1024208590 Chr.17: 74342633-74342633 on Build GRCh38
rs957128461 Chr.17: 74342573-74342573 on Build GRCh38
rs746932945 Chr.17: 74342591-74342591 on Build GRCh38
rs551276245 Chr.17: 74342808-74342808 on Build GRCh38
rs970987111 Chr.17: 74342653-74342653 on Build GRCh38
rs939060868 Chr.17: 74343003-74343003 on Build GRCh38
rs547257814 Chr.17: 74342749-74342749 on Build GRCh38
rs773399909 Chr.17: 74342594-74342594 on Build GRCh38
rs753068117 Chr.17: 74342625-74342625 on Build GRCh38
rs749971599 Chr.17: 74342995-74342995 on Build GRCh38
rs746096971 Chr.17: 74342750-74342750 on Build GRCh38
rs758047071 Chr.17: 74342670-74342670 on Build GRCh38
rs765231753 Chr.17: 74342857-74342857 on Build GRCh38
rs920388108 Chr.17: 74342753-74342753 on Build GRCh38
rs756048450 Chr.17: 74343024-74343024 on Build GRCh38
rs143245689 Chr.17: 74342996-74342996 on Build GRCh38
rs775809118 Chr.17: 74342754-74342754 on Build GRCh38
rs147978023 Chr.17: 74342681-74342681 on Build GRCh38
rs763074585 Chr.17: 74342601-74342601 on Build GRCh38
rs754365782 Chr.17: 74342648-74342648 on Build GRCh38
rs1010928793 Chr.17: 74342930-74342930 on Build GRCh38
rs1045275967 Chr.17: 74342878-74342878 on Build GRCh38
rs748098332 Chr.17: 74342710-74342710 on Build GRCh38
rs746844897 Chr.17: 74342672-74342672 on Build GRCh38
rs1051642482 Chr.17: 74342809-74342809 on Build GRCh38
rs751266389 Chr.17: 74343006-74343006 on Build GRCh38
rs762323940 Chr.17: 74342980-74342980 on Build GRCh38
rs577100138 Chr.17: 74342943-74342943 on Build GRCh38
rs117809084 Chr.17: 74342770-74342770 on Build GRCh38
rs759688484 Chr.17: 74342611-74342611 on Build GRCh38
rs781232133 Chr.17: 74342700-74342700 on Build GRCh38
rs367977040 Chr.17: 74342968-74342968 on Build GRCh38
rs568051542 Chr.17: 74342907-74342907 on Build GRCh38
rs975902607 Chr.17: 74342958-74342958 on Build GRCh38
rs964197040 Chr.17: 74342956-74342956 on Build GRCh38
rs113193465 Chr.17: 74342825-74342825 on Build GRCh38
rs748079343 Chr.17: 74343023-74343023 on Build GRCh38
rs568603209 Chr.17: 74342838-74342838 on Build GRCh38
rs372391374 Chr.17: 74342757-74342757 on Build GRCh38
rs750036187 Chr.17: 74342666-74342666 on Build GRCh38
rs767908881 Chr.17: 74342623-74342623 on Build GRCh38
rs371038827 Chr.17: 74342695-74342695 on Build GRCh38
rs547242562 Chr.17: 74342874-74342874 on Build GRCh38
rs376348767 Chr.17: 74342602-74342602 on Build GRCh38
rs756717798 Chr.17: 74342650-74342650 on Build GRCh38
rs892723349 Chr.17: 74342793-74342793 on Build GRCh38
rs1019821086 Chr.17: 74342965-74342981 on Build GRCh38
rs939014587 Chr.17: 74342774-74342774 on Build GRCh38
rs141818925 Chr.17: 74342946-74342946 on Build GRCh38
rs1017572436 Chr.17: 74342902-74342902 on Build GRCh38
rs764633982 Chr.17: 74342645-74342645 on Build GRCh38
rs779047979 Chr.17: 74343030-74343030 on Build GRCh38
rs531061555 Chr.17: 74342771-74342771 on Build GRCh38
rs975467405 Chr.17: 74342661-74342661 on Build GRCh38
rs368432623 Chr.17: 74342665-74342665 on Build GRCh38
rs370742356 Chr.17: 74342879-74342879 on Build GRCh38
rs921756900 Chr.17: 74342780-74342780 on Build GRCh38
rs371480015 Chr.17: 74342904-74342904 on Build GRCh38
rs572959222 Chr.17: 74342987-74342987 on Build GRCh38
rs557240061 Chr.17: 74342924-74342924 on Build GRCh38
rs764474466 Chr.17: 74342762-74342762 on Build GRCh38
rs12601559 Chr.17: 74342626-74342626 on Build GRCh38
rs1022476119 Chr.17: 74342828-74342828 on Build GRCh38
rs747059707 Chr.17: 74342631-74342632 on Build GRCh38
rs376191167 Chr.17: 74342659-74342659 on Build GRCh38
rs536921167 Chr.17: 74342908-74342908 on Build GRCh38
rs537444829 Chr.17: 74343020-74343020 on Build GRCh38
rs779898771 Chr.17: 74342671-74342671 on Build GRCh38
rs373796045 Chr.17: 74342791-74342791 on Build GRCh38
rs369540361 Chr.17: 74342976-74342976 on Build GRCh38
rs186566355 Chr.17: 74342751-74342751 on Build GRCh38
rs534750830 Chr.17: 74342901-74342901 on Build GRCh38
rs372766186 Chr.17: 74342590-74342590 on Build GRCh38
rs947787476 Chr.17: 74342870-74342870 on Build GRCh38
rs533553190 Chr.17: 74342866-74342866 on Build GRCh38

View Full Product Details