Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74344669-74345162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF19
Entrez Gene ID
124602
Gene Name
kinesin family member 19
Gene Aliases
KIF19A
UniGene
Hs.372773
Gene Chromosome Location
Chr.17: 74326212-74355820 on Build GRCh38
Transcripts
NM_153209.3 XM_005257029.1 XM_005257030.1 XM_017024150.1 XM_017024151.1 XM_017024152.1 XM_017024153.1 XM_017024154.1 XM_017024155.1 XM_017024156.1 XM_017024157.1 XM_017024158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3421874 c.748G>A p.D250N Chr.17: 74344926-74344926 on Build GRCh38
6881225 c.751C>T p.L251L Chr.17: 74344929-74344929 on Build GRCh38
1479955 c.768C>T p.R256R Chr.17: 74344946-74344946 on Build GRCh38
5495468 c.648G>A p.T216T Chr.17: 74344826-74344826 on Build GRCh38
6881224 c.717G>A p.Q239Q Chr.17: 74344895-74344895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34914485 Chr.17: 74344826-74344826 on Build GRCh38
rs567683553 Chr.17: 74344698-74344698 on Build GRCh38
rs1024497514 Chr.17: 74345081-74345081 on Build GRCh38
rs759414680 Chr.17: 74344870-74344870 on Build GRCh38
rs767467259 Chr.17: 74344724-74344724 on Build GRCh38
rs539441171 Chr.17: 74344843-74344843 on Build GRCh38
rs754064985 Chr.17: 74344833-74344833 on Build GRCh38
rs762120737 Chr.17: 74344877-74344877 on Build GRCh38
rs976621698 Chr.17: 74344914-74344914 on Build GRCh38
rs558677969 Chr.17: 74345120-74345120 on Build GRCh38
rs760079664 Chr.17: 74345001-74345001 on Build GRCh38
rs748925466 Chr.17: 74344925-74344925 on Build GRCh38
rs776277390 Chr.17: 74344807-74344807 on Build GRCh38
rs377449569 Chr.17: 74344750-74344750 on Build GRCh38
rs957397999 Chr.17: 74345108-74345108 on Build GRCh38
rs547925839 Chr.17: 74344971-74344971 on Build GRCh38
rs34016821 Chr.17: 74344842-74344842 on Build GRCh38
rs763308420 Chr.17: 74344896-74344896 on Build GRCh38
rs368550936 Chr.17: 74344722-74344722 on Build GRCh38
rs1014235158 Chr.17: 74345059-74345059 on Build GRCh38
rs757849063 Chr.17: 74345010-74345010 on Build GRCh38
rs868303686 Chr.17: 74344933-74344933 on Build GRCh38
rs115500074 Chr.17: 74344757-74344757 on Build GRCh38
rs187861420 Chr.17: 74344721-74344721 on Build GRCh38
rs1017990318 Chr.17: 74345137-74345137 on Build GRCh38
rs746354359 Chr.17: 74344758-74344758 on Build GRCh38
rs570244557 Chr.17: 74345089-74345089 on Build GRCh38
rs765612756 Chr.17: 74344879-74344879 on Build GRCh38
rs201262345 Chr.17: 74344789-74344789 on Build GRCh38
rs1053686893 Chr.17: 74344748-74344748 on Build GRCh38
rs200024337 Chr.17: 74344834-74344834 on Build GRCh38
rs777341825 Chr.17: 74344924-74344924 on Build GRCh38
rs113296978 Chr.17: 74344691-74344691 on Build GRCh38
rs370212175 Chr.17: 74344972-74344972 on Build GRCh38
rs564644989 Chr.17: 74344817-74344817 on Build GRCh38
rs565085903 Chr.17: 74344945-74344945 on Build GRCh38
rs144185563 Chr.17: 74344815-74344815 on Build GRCh38
rs752890699 Chr.17: 74344728-74344728 on Build GRCh38
rs942573780 Chr.17: 74344904-74344904 on Build GRCh38
rs764320324 Chr.17: 74344737-74344737 on Build GRCh38
rs200721967 Chr.17: 74344844-74344844 on Build GRCh38
rs759329655 Chr.17: 74344863-74344863 on Build GRCh38
rs888141740 Chr.17: 74344988-74344988 on Build GRCh38
rs201328999 Chr.17: 74344884-74344884 on Build GRCh38
rs144888082 Chr.17: 74344835-74344835 on Build GRCh38
rs748179750 Chr.17: 74344951-74344951 on Build GRCh38
rs780681154 Chr.17: 74344911-74344911 on Build GRCh38
rs756278210 Chr.17: 74344729-74344729 on Build GRCh38
rs754118046 Chr.17: 74344749-74344749 on Build GRCh38
rs769558861 Chr.17: 74344811-74344811 on Build GRCh38
rs530018088 Chr.17: 74345035-74345035 on Build GRCh38
rs144743924 Chr.17: 74344850-74344850 on Build GRCh38
rs1032182653 Chr.17: 74345104-74345104 on Build GRCh38
rs779422970 Chr.17: 74344782-74344782 on Build GRCh38
rs139160580 Chr.17: 74344810-74344810 on Build GRCh38
rs148507128 Chr.17: 74344819-74344819 on Build GRCh38
rs752025633 Chr.17: 74345090-74345090 on Build GRCh38
rs957347162 Chr.17: 74344876-74344876 on Build GRCh38
rs774648618 Chr.17: 74344992-74344992 on Build GRCh38
rs770710316 Chr.17: 74344937-74344937 on Build GRCh38
rs762571318 Chr.17: 74344743-74344745 on Build GRCh38
rs542642249 Chr.17: 74344860-74344860 on Build GRCh38
rs77811595 Chr.17: 74345096-74345096 on Build GRCh38
rs941020412 Chr.17: 74345006-74345006 on Build GRCh38
rs376096990 Chr.17: 74344761-74344761 on Build GRCh38
rs372182489 Chr.17: 74344862-74344862 on Build GRCh38
rs374106339 Chr.17: 74344711-74344711 on Build GRCh38
rs925250818 Chr.17: 74344712-74344712 on Build GRCh38
rs957828415 Chr.17: 74345114-74345114 on Build GRCh38
rs766327825 Chr.17: 74344821-74344821 on Build GRCh38
rs145463528 Chr.17: 74344875-74344875 on Build GRCh38
rs760743166 Chr.17: 74344825-74344825 on Build GRCh38
rs193276897 Chr.17: 74344903-74344903 on Build GRCh38
rs1001750786 Chr.17: 74345023-74345023 on Build GRCh38
rs896051717 Chr.17: 74345058-74345058 on Build GRCh38
rs112674750 Chr.17: 74345107-74345107 on Build GRCh38
rs755638085 Chr.17: 74344912-74344912 on Build GRCh38
rs770638270 Chr.17: 74344867-74344867 on Build GRCh38
rs774170996 Chr.17: 74344820-74344820 on Build GRCh38
rs373527071 Chr.17: 74344902-74344902 on Build GRCh38
rs750882046 Chr.17: 74344841-74344841 on Build GRCh38
rs763102021 Chr.17: 74344965-74344965 on Build GRCh38
rs146234533 Chr.17: 74344788-74344788 on Build GRCh38
rs568463083 Chr.17: 74345009-74345009 on Build GRCh38
rs761536492 Chr.17: 74344715-74344715 on Build GRCh38
rs769996166 Chr.17: 74344952-74344952 on Build GRCh38
rs550195220 Chr.17: 74345084-74345084 on Build GRCh38
rs527755806 Chr.17: 74344946-74344946 on Build GRCh38
rs200390291 Chr.17: 74344869-74344869 on Build GRCh38
rs149952681 Chr.17: 74344814-74344814 on Build GRCh38
rs766681366 Chr.17: 74344899-74344899 on Build GRCh38
rs768758945 Chr.17: 74344871-74344871 on Build GRCh38
rs562183394 Chr.17: 74344859-74344859 on Build GRCh38
rs773524953 Chr.17: 74344958-74344958 on Build GRCh38
rs776836395 Chr.17: 74344947-74344947 on Build GRCh38
rs967820155 Chr.17: 74344690-74344690 on Build GRCh38
rs749026804 Chr.17: 74344864-74344864 on Build GRCh38
rs752336468 Chr.17: 74344944-74344944 on Build GRCh38
rs567779129 Chr.17: 74345028-74345028 on Build GRCh38
rs565955155 Chr.17: 74345138-74345138 on Build GRCh38

View Full Product Details