Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75727113-75727615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB4
Entrez Gene ID
3691
Gene Name
integrin subunit beta 4
Gene Aliases
CD104, GP150
UniGene
Hs.632226
Gene Chromosome Location
Chr.17: 75721328-75757819 on Build GRCh38
Transcripts
NM_000213.4 NM_001005619.1 NM_001005731.2 NM_001321123.1 XM_005257309.2 XM_005257311.4 XM_006721866.3 XM_006721867.3 XM_006721868.3 XM_006721870.3 XM_011524751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147909 c.194C>T p.A65V Chr.17: 75727435-75727435 on Build GRCh38
4069645 c.199C>T p.L67L Chr.17: 75727440-75727440 on Build GRCh38
4922735 c.145G>T p.A49S Chr.17: 75727260-75727260 on Build GRCh38
4069643 c.145G>A p.A49T Chr.17: 75727260-75727260 on Build GRCh38
6998875 c.179G>A p.R60H Chr.17: 75727420-75727420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759870083 Chr.17: 75727275-75727275 on Build GRCh38
rs765142082 Chr.17: 75727298-75727298 on Build GRCh38
rs1015321334 Chr.17: 75727439-75727439 on Build GRCh38
rs398031629 Chr.17: 75727378-75727378 on Build GRCh38
rs753782989 Chr.17: 75727235-75727235 on Build GRCh38
rs772979254 Chr.17: 75727380-75727380 on Build GRCh38
rs774764615 Chr.17: 75727274-75727274 on Build GRCh38
rs758821844 Chr.17: 75727534-75727534 on Build GRCh38
rs372947104 Chr.17: 75727231-75727231 on Build GRCh38
rs767333175 Chr.17: 75727523-75727523 on Build GRCh38
rs1009490043 Chr.17: 75727384-75727384 on Build GRCh38
rs763732398 Chr.17: 75727234-75727234 on Build GRCh38
rs926803832 Chr.17: 75727584-75727584 on Build GRCh38
rs765187788 Chr.17: 75727374-75727374 on Build GRCh38
rs939494246 Chr.17: 75727591-75727591 on Build GRCh38
rs774591158 Chr.17: 75727216-75727216 on Build GRCh38
rs771728733 Chr.17: 75727176-75727176 on Build GRCh38
rs762395308 Chr.17: 75727373-75727373 on Build GRCh38
rs201532846 Chr.17: 75727417-75727417 on Build GRCh38
rs778748791 Chr.17: 75727243-75727243 on Build GRCh38
rs973694265 Chr.17: 75727478-75727478 on Build GRCh38
rs772790459 Chr.17: 75727461-75727461 on Build GRCh38
rs774110524 Chr.17: 75727484-75727484 on Build GRCh38
rs566677029 Chr.17: 75727318-75727318 on Build GRCh38
rs751359332 Chr.17: 75727390-75727390 on Build GRCh38
rs745322866 Chr.17: 75727212-75727212 on Build GRCh38
rs747199450 Chr.17: 75727171-75727171 on Build GRCh38
rs200853535 Chr.17: 75727286-75727286 on Build GRCh38
rs776264602 Chr.17: 75727361-75727361 on Build GRCh38
rs146966502 Chr.17: 75727260-75727260 on Build GRCh38
rs925126060 Chr.17: 75727556-75727556 on Build GRCh38
rs750938314 Chr.17: 75727378-75727379 on Build GRCh38
rs371650219 Chr.17: 75727158-75727158 on Build GRCh38
rs775938419 Chr.17: 75727326-75727326 on Build GRCh38
rs398079175 Chr.17: 75727377-75727377 on Build GRCh38
rs376523967 Chr.17: 75727262-75727262 on Build GRCh38
rs767136048 Chr.17: 75727226-75727226 on Build GRCh38
rs569235037 Chr.17: 75727449-75727449 on Build GRCh38
rs757739484 Chr.17: 75727322-75727322 on Build GRCh38
rs980991265 Chr.17: 75727527-75727527 on Build GRCh38
rs761834593 Chr.17: 75727296-75727296 on Build GRCh38
rs764349772 Chr.17: 75727409-75727409 on Build GRCh38
rs373500013 Chr.17: 75727303-75727303 on Build GRCh38
rs80338755 Chr.17: 75727423-75727423 on Build GRCh38
rs766405875 Chr.17: 75727480-75727480 on Build GRCh38
rs775284751 Chr.17: 75727395-75727395 on Build GRCh38
rs757276151 Chr.17: 75727242-75727242 on Build GRCh38
rs150023264 Chr.17: 75727259-75727259 on Build GRCh38
rs772279298 Chr.17: 75727325-75727325 on Build GRCh38
rs747831589 Chr.17: 75727450-75727450 on Build GRCh38
rs762732702 Chr.17: 75727472-75727472 on Build GRCh38
rs945470804 Chr.17: 75727512-75727512 on Build GRCh38
rs748230223 Chr.17: 75727180-75727180 on Build GRCh38
rs746420073 Chr.17: 75727323-75727323 on Build GRCh38
rs756323532 Chr.17: 75727400-75727400 on Build GRCh38
rs121912465 Chr.17: 75727227-75727227 on Build GRCh38
rs372797886 Chr.17: 75727436-75727436 on Build GRCh38
rs199911115 Chr.17: 75727315-75727315 on Build GRCh38
rs2584100 Chr.17: 75727159-75727159 on Build GRCh38
rs369324606 Chr.17: 75727299-75727299 on Build GRCh38
rs757517774 Chr.17: 75727528-75727528 on Build GRCh38
rs769790730 Chr.17: 75727513-75727513 on Build GRCh38
rs200966154 Chr.17: 75727435-75727435 on Build GRCh38
rs773871702 Chr.17: 75727194-75727194 on Build GRCh38
rs373707270 Chr.17: 75727547-75727547 on Build GRCh38
rs771726104 Chr.17: 75727284-75727284 on Build GRCh38
rs1056451931 Chr.17: 75727349-75727349 on Build GRCh38
rs749736808 Chr.17: 75727266-75727266 on Build GRCh38
rs1043654582 Chr.17: 75727387-75727387 on Build GRCh38
rs750249253 Chr.17: 75727248-75727248 on Build GRCh38
rs368937384 Chr.17: 75727285-75727285 on Build GRCh38
rs781314847 Chr.17: 75727253-75727253 on Build GRCh38
rs781325992 Chr.17: 75727175-75727175 on Build GRCh38
rs746597537 Chr.17: 75727356-75727356 on Build GRCh38
rs755212864 Chr.17: 75727251-75727251 on Build GRCh38
rs754012248 Chr.17: 75727525-75727525 on Build GRCh38
rs753406366 Chr.17: 75727293-75727293 on Build GRCh38
rs775074217 Chr.17: 75727186-75727187 on Build GRCh38
rs750587121 Chr.17: 75727533-75727533 on Build GRCh38
rs544136973 Chr.17: 75727416-75727416 on Build GRCh38
rs936449869 Chr.17: 75727573-75727573 on Build GRCh38
rs765166765 Chr.17: 75727529-75727529 on Build GRCh38
rs550392338 Chr.17: 75727419-75727419 on Build GRCh38
rs766028777 Chr.17: 75727388-75727388 on Build GRCh38
rs200500313 Chr.17: 75727451-75727451 on Build GRCh38
rs920810686 Chr.17: 75727236-75727236 on Build GRCh38
rs752230205 Chr.17: 75727228-75727228 on Build GRCh38
rs759223935 Chr.17: 75727218-75727218 on Build GRCh38
rs557836108 Chr.17: 75727473-75727473 on Build GRCh38
rs760303034 Chr.17: 75727232-75727232 on Build GRCh38
rs972322019 Chr.17: 75727225-75727225 on Build GRCh38
rs571682079 Chr.17: 75727570-75727570 on Build GRCh38
rs772995224 Chr.17: 75727290-75727290 on Build GRCh38
rs558845154 Chr.17: 75727524-75727524 on Build GRCh38
rs1042466757 Chr.17: 75727551-75727551 on Build GRCh38
rs892480182 Chr.17: 75727373-75727373 on Build GRCh38
rs762968243 Chr.17: 75727386-75727386 on Build GRCh38
rs768493013 Chr.17: 75727360-75727360 on Build GRCh38
rs746305033 Chr.17: 75727420-75727420 on Build GRCh38
rs138695324 Chr.17: 75727200-75727200 on Build GRCh38
rs776953208 Chr.17: 75727448-75727448 on Build GRCh38
rs752866356 Chr.17: 75727309-75727309 on Build GRCh38
rs530665868 Chr.17: 75727201-75727201 on Build GRCh38

View Full Product Details