Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34300366-34300862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0355
Entrez Gene ID
9710
Gene Name
KIAA0355
Gene Aliases
-
UniGene
Hs.330073
Gene Chromosome Location
Chr.19: 34254551-34355566 on Build GRCh38
Transcripts
NM_014686.3 XM_005259444.1 XM_017027525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5150115 c.190C>T p.H64Y Chr.19: 34300663-34300663 on Build GRCh38
4076831 c.223G>A p.D75N Chr.19: 34300696-34300696 on Build GRCh38
5895050 c.173C>T p.T58I Chr.19: 34300646-34300646 on Build GRCh38
1189997 c.59G>T p.G20V Chr.19: 34300532-34300532 on Build GRCh38
1719252 c.155C>T p.A52V Chr.19: 34300628-34300628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764020643 Chr.19: 34300696-34300696 on Build GRCh38
rs754003690 Chr.19: 34300729-34300729 on Build GRCh38
rs760940741 Chr.19: 34300799-34300799 on Build GRCh38
rs770572065 Chr.19: 34300755-34300755 on Build GRCh38
rs778258992 Chr.19: 34300748-34300748 on Build GRCh38
rs760079039 Chr.19: 34300611-34300611 on Build GRCh38
rs779758980 Chr.19: 34300439-34300439 on Build GRCh38
rs751405374 Chr.19: 34300798-34300798 on Build GRCh38
rs757429444 Chr.19: 34300572-34300572 on Build GRCh38
rs747401571 Chr.19: 34300842-34300842 on Build GRCh38
rs915180197 Chr.19: 34300551-34300551 on Build GRCh38
rs775951539 Chr.19: 34300493-34300493 on Build GRCh38
rs558834376 Chr.19: 34300533-34300533 on Build GRCh38
rs903192757 Chr.19: 34300708-34300708 on Build GRCh38
rs756262638 Chr.19: 34300636-34300636 on Build GRCh38
rs759067421 Chr.19: 34300783-34300783 on Build GRCh38
rs111551327 Chr.19: 34300790-34300790 on Build GRCh38
rs777929074 Chr.19: 34300833-34300833 on Build GRCh38
rs752436465 Chr.19: 34300628-34300628 on Build GRCh38
rs367800219 Chr.19: 34300562-34300562 on Build GRCh38
rs760028635 Chr.19: 34300476-34300476 on Build GRCh38
rs754736372 Chr.19: 34300552-34300552 on Build GRCh38
rs766994852 Chr.19: 34300514-34300514 on Build GRCh38
rs73580724 Chr.19: 34300581-34300581 on Build GRCh38
rs199681316 Chr.19: 34300471-34300471 on Build GRCh38
rs772707616 Chr.19: 34300663-34300663 on Build GRCh38
rs980676510 Chr.19: 34300801-34300801 on Build GRCh38
rs759725775 Chr.19: 34300549-34300549 on Build GRCh38
rs776076607 Chr.19: 34300780-34300780 on Build GRCh38
rs530657729 Chr.19: 34300390-34300390 on Build GRCh38
rs149799888 Chr.19: 34300738-34300738 on Build GRCh38
rs772727313 Chr.19: 34300609-34300609 on Build GRCh38
rs761431454 Chr.19: 34300513-34300513 on Build GRCh38
rs375199695 Chr.19: 34300437-34300437 on Build GRCh38
rs368217436 Chr.19: 34300447-34300447 on Build GRCh38
rs779987848 Chr.19: 34300536-34300536 on Build GRCh38
rs752731347 Chr.19: 34300694-34300694 on Build GRCh38
rs777949055 Chr.19: 34300446-34300446 on Build GRCh38
rs953283950 Chr.19: 34300785-34300785 on Build GRCh38
rs140257965 Chr.19: 34300617-34300617 on Build GRCh38
rs200244481 Chr.19: 34300651-34300651 on Build GRCh38
rs770204594 Chr.19: 34300484-34300484 on Build GRCh38
rs778928865 Chr.19: 34300824-34300825 on Build GRCh38
rs201404557 Chr.19: 34300616-34300616 on Build GRCh38
rs763743064 Chr.19: 34300796-34300796 on Build GRCh38
rs201207182 Chr.19: 34300692-34300692 on Build GRCh38
rs754962398 Chr.19: 34300618-34300618 on Build GRCh38
rs151200468 Chr.19: 34300591-34300591 on Build GRCh38
rs763390949 Chr.19: 34300497-34300497 on Build GRCh38
rs201416108 Chr.19: 34300498-34300498 on Build GRCh38
rs776127731 Chr.19: 34300687-34300687 on Build GRCh38
rs537194127 Chr.19: 34300598-34300598 on Build GRCh38
rs373446167 Chr.19: 34300747-34300747 on Build GRCh38
rs772231710 Chr.19: 34300443-34300443 on Build GRCh38
rs146815070 Chr.19: 34300830-34300830 on Build GRCh38
rs372162457 Chr.19: 34300669-34300669 on Build GRCh38
rs749880333 Chr.19: 34300593-34300593 on Build GRCh38
rs753131048 Chr.19: 34300826-34300826 on Build GRCh38
rs781251259 Chr.19: 34300632-34300632 on Build GRCh38
rs757458282 Chr.19: 34300629-34300629 on Build GRCh38
rs375023608 Chr.19: 34300434-34300434 on Build GRCh38
rs376785044 Chr.19: 34300806-34300806 on Build GRCh38
rs192513488 Chr.19: 34300605-34300605 on Build GRCh38
rs750602156 Chr.19: 34300699-34300699 on Build GRCh38
rs769388748 Chr.19: 34300784-34300784 on Build GRCh38
rs996985670 Chr.19: 34300395-34300395 on Build GRCh38
rs369572498 Chr.19: 34300740-34300740 on Build GRCh38
rs751256442 Chr.19: 34300499-34300499 on Build GRCh38
rs920744745 Chr.19: 34300604-34300604 on Build GRCh38
rs550377095 Chr.19: 34300442-34300442 on Build GRCh38
rs779115366 Chr.19: 34300811-34300811 on Build GRCh38
rs780217364 Chr.19: 34300718-34300718 on Build GRCh38
rs1024832279 Chr.19: 34300418-34300418 on Build GRCh38
rs140588030 Chr.19: 34300571-34300571 on Build GRCh38
rs765115581 Chr.19: 34300693-34300693 on Build GRCh38
rs373975583 Chr.19: 34300519-34300519 on Build GRCh38
rs756418226 Chr.19: 34300713-34300713 on Build GRCh38
rs750361197 Chr.19: 34300433-34300433 on Build GRCh38
rs746144405 Chr.19: 34300633-34300633 on Build GRCh38
rs184853377 Chr.19: 34300682-34300682 on Build GRCh38
rs780635187 Chr.19: 34300427-34300427 on Build GRCh38
rs989218758 Chr.19: 34300491-34300491 on Build GRCh38
rs368671239 Chr.19: 34300648-34300648 on Build GRCh38
rs774593069 Chr.19: 34300603-34300603 on Build GRCh38
rs753820886 Chr.19: 34300548-34300548 on Build GRCh38
rs764520420 Chr.19: 34300658-34300658 on Build GRCh38
rs765861221 Chr.19: 34300675-34300675 on Build GRCh38
rs755075889 Chr.19: 34300685-34300687 on Build GRCh38
rs200592725 Chr.19: 34300757-34300757 on Build GRCh38
rs572714541 Chr.19: 34300794-34300794 on Build GRCh38
rs764950563 Chr.19: 34300623-34300623 on Build GRCh38
rs780342636 Chr.19: 34300647-34300647 on Build GRCh38
rs769130065 Chr.19: 34300599-34300599 on Build GRCh38
rs754164774 Chr.19: 34300800-34300800 on Build GRCh38
rs772534773 Chr.19: 34300656-34300656 on Build GRCh38
rs535765849 Chr.19: 34300472-34300472 on Build GRCh38
rs999310612 Chr.19: 34300444-34300445 on Build GRCh38
rs867083071 Chr.19: 34300490-34300490 on Build GRCh38
rs956384018 Chr.19: 34300518-34300518 on Build GRCh38
rs927780887 Chr.19: 34300585-34300585 on Build GRCh38
rs202077029 Chr.19: 34300695-34300695 on Build GRCh38
rs371396471 Chr.19: 34300589-34300589 on Build GRCh38
rs188387566 Chr.19: 34300824-34300824 on Build GRCh38

View Full Product Details