Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34339762-34340277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0355
Entrez Gene ID
9710
Gene Name
KIAA0355
Gene Aliases
-
UniGene
Hs.330073
Gene Chromosome Location
Chr.19: 34254551-34355566 on Build GRCh38
Transcripts
NM_014686.3 XM_005259444.1 XM_017027525.1 XM_017027526.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2751742 c.1425C>T p.T475T Chr.19: 34339930-34339930 on Build GRCh38
4641124 c.1458G>A p.V486V Chr.19: 34339963-34339963 on Build GRCh38
994747 c.1372G>T p.E458* Chr.19: 34339877-34339877 on Build GRCh38
128312 c.1368G>A p.L456L Chr.19: 34339873-34339873 on Build GRCh38
4752608 c.1471C>T p.R491C Chr.19: 34339976-34339976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919779267 Chr.19: 34340075-34340075 on Build GRCh38
rs895643890 Chr.19: 34340181-34340181 on Build GRCh38
rs749645515 Chr.19: 34340019-34340019 on Build GRCh38
rs773124931 Chr.19: 34339954-34339954 on Build GRCh38
rs751954787 Chr.19: 34340201-34340201 on Build GRCh38
rs372164070 Chr.19: 34339854-34339854 on Build GRCh38
rs760672387 Chr.19: 34339962-34339962 on Build GRCh38
rs760675961 Chr.19: 34339945-34339946 on Build GRCh38
rs766097339 Chr.19: 34339948-34339949 on Build GRCh38
rs904469507 Chr.19: 34340143-34340143 on Build GRCh38
rs544437477 Chr.19: 34339976-34339976 on Build GRCh38
rs752750303 Chr.19: 34339977-34339977 on Build GRCh38
rs1042089277 Chr.19: 34340194-34340194 on Build GRCh38
rs371451448 Chr.19: 34340001-34340001 on Build GRCh38
rs1014994714 Chr.19: 34340186-34340186 on Build GRCh38
rs1027617327 Chr.19: 34340215-34340215 on Build GRCh38
rs962049630 Chr.19: 34339957-34339957 on Build GRCh38
rs780234633 Chr.19: 34340008-34340008 on Build GRCh38
rs986373426 Chr.19: 34340230-34340230 on Build GRCh38
rs16969204 Chr.19: 34339802-34339802 on Build GRCh38
rs1001338793 Chr.19: 34339871-34339871 on Build GRCh38
rs372625002 Chr.19: 34339817-34339817 on Build GRCh38
rs547948744 Chr.19: 34340117-34340117 on Build GRCh38
rs192998161 Chr.19: 34339827-34339827 on Build GRCh38
rs533767319 Chr.19: 34340202-34340202 on Build GRCh38
rs200784438 Chr.19: 34339988-34339988 on Build GRCh38
rs759754898 Chr.19: 34340021-34340022 on Build GRCh38
rs891453879 Chr.19: 34339786-34339786 on Build GRCh38
rs976526753 Chr.19: 34340081-34340081 on Build GRCh38
rs945720746 Chr.19: 34340087-34340087 on Build GRCh38
rs545818491 Chr.19: 34339911-34339911 on Build GRCh38
rs113639122 Chr.19: 34340092-34340092 on Build GRCh38
rs374868846 Chr.19: 34340225-34340225 on Build GRCh38
rs776516887 Chr.19: 34339964-34339964 on Build GRCh38
rs1014776392 Chr.19: 34340185-34340185 on Build GRCh38
rs778536181 Chr.19: 34340026-34340026 on Build GRCh38
rs201716497 Chr.19: 34339930-34339930 on Build GRCh38
rs761283498 Chr.19: 34339945-34339945 on Build GRCh38
rs530387778 Chr.19: 34339996-34339996 on Build GRCh38
rs776426135 Chr.19: 34339949-34339950 on Build GRCh38
rs1054893850 Chr.19: 34340105-34340105 on Build GRCh38
rs758649320 Chr.19: 34339862-34339862 on Build GRCh38
rs753011361 Chr.19: 34340049-34340049 on Build GRCh38
rs570771189 Chr.19: 34340248-34340248 on Build GRCh38
rs771641269 Chr.19: 34339946-34339946 on Build GRCh38
rs764177747 Chr.19: 34340168-34340168 on Build GRCh38
rs765340032 Chr.19: 34339939-34339939 on Build GRCh38
rs184564406 Chr.19: 34340129-34340129 on Build GRCh38
rs576090527 Chr.19: 34339942-34339942 on Build GRCh38
rs781755195 Chr.19: 34339912-34339912 on Build GRCh38
rs562440671 Chr.19: 34339935-34339935 on Build GRCh38
rs532390363 Chr.19: 34340033-34340033 on Build GRCh38
rs912912738 Chr.19: 34340082-34340082 on Build GRCh38
rs375867102 Chr.19: 34339820-34339820 on Build GRCh38
rs35990349 Chr.19: 34339882-34339882 on Build GRCh38
rs986300271 Chr.19: 34340021-34340021 on Build GRCh38
rs369367294 Chr.19: 34339963-34339963 on Build GRCh38
rs755341055 Chr.19: 34340025-34340025 on Build GRCh38
rs552590197 Chr.19: 34340074-34340074 on Build GRCh38
rs201498230 Chr.19: 34339836-34339836 on Build GRCh38
rs1055563918 Chr.19: 34340161-34340161 on Build GRCh38
rs765153618 Chr.19: 34339850-34339850 on Build GRCh38
rs765409620 Chr.19: 34340041-34340043 on Build GRCh38
rs752800135 Chr.19: 34339859-34339859 on Build GRCh38
rs939986229 Chr.19: 34340046-34340046 on Build GRCh38
rs199945585 Chr.19: 34340017-34340017 on Build GRCh38
rs943818983 Chr.19: 34340060-34340060 on Build GRCh38
rs937222115 Chr.19: 34340149-34340149 on Build GRCh38
rs540904383 Chr.19: 34340004-34340004 on Build GRCh38
rs746444067 Chr.19: 34339927-34339927 on Build GRCh38
rs143541447 Chr.19: 34339981-34339981 on Build GRCh38
rs531844305 Chr.19: 34340103-34340103 on Build GRCh38
rs139810249 Chr.19: 34340100-34340100 on Build GRCh38
rs576408605 Chr.19: 34339841-34339841 on Build GRCh38
rs949073676 Chr.19: 34340163-34340163 on Build GRCh38

View Full Product Details