Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46675067-46675569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB14
Entrez Gene ID
9813
Gene Name
EF-hand calcium binding domain 14
Gene Aliases
KIAA0494
UniGene
Hs.707688
Gene Chromosome Location
Chr.1: 46675159-46719064 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Gene Symbol
EFCAB14-AS1
Entrez Gene ID
100130197
Gene Name
EFCAB14 antisense RNA 1
Gene Aliases
KIAA0494-AS1
UniGene
Hs.568643
Gene Chromosome Location
Chr.1: 46674036-46692098 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

SNP Detail

SNP ID Chromosome Location
rs781747331 Chr.1: 46675206-46675206 on Build GRCh38
rs748148309 Chr.1: 46675451-46675451 on Build GRCh38
rs909303078 Chr.1: 46675449-46675449 on Build GRCh38
rs78841814 Chr.1: 46675104-46675104 on Build GRCh38
rs1801800 Chr.1: 46675250-46675250 on Build GRCh38
rs185283737 Chr.1: 46675143-46675143 on Build GRCh38
rs1042860392 Chr.1: 46675237-46675237 on Build GRCh38
rs1051945053 Chr.1: 46675176-46675176 on Build GRCh38
rs548301184 Chr.1: 46675112-46675112 on Build GRCh38
rs909238214 Chr.1: 46675144-46675144 on Build GRCh38
rs773300300 Chr.1: 46675275-46675275 on Build GRCh38
rs991617576 Chr.1: 46675390-46675390 on Build GRCh38
rs933597007 Chr.1: 46675521-46675521 on Build GRCh38
rs148225926 Chr.1: 46675159-46675159 on Build GRCh38
rs1020134311 Chr.1: 46675352-46675352 on Build GRCh38
rs190322994 Chr.1: 46675436-46675436 on Build GRCh38
rs1051933780 Chr.1: 46675546-46675546 on Build GRCh38
rs748377325 Chr.1: 46675478-46675478 on Build GRCh38
rs1002484579 Chr.1: 46675353-46675353 on Build GRCh38
rs762834384 Chr.1: 46675385-46675385 on Build GRCh38
rs925814230 Chr.1: 46675249-46675249 on Build GRCh38
rs967269086 Chr.1: 46675370-46675370 on Build GRCh38
rs1044492509 Chr.1: 46675342-46675342 on Build GRCh38
rs530165610 Chr.1: 46675102-46675102 on Build GRCh38
rs997646515 Chr.1: 46675175-46675175 on Build GRCh38
rs1006678521 Chr.1: 46675215-46675215 on Build GRCh38
rs942263996 Chr.1: 46675151-46675151 on Build GRCh38
rs1801801 Chr.1: 46675404-46675404 on Build GRCh38
rs952828439 Chr.1: 46675388-46675388 on Build GRCh38
rs928520846 Chr.1: 46675384-46675384 on Build GRCh38
rs554727884 Chr.1: 46675136-46675136 on Build GRCh38
rs1801799 Chr.1: 46675314-46675314 on Build GRCh38
rs772602017 Chr.1: 46675227-46675227 on Build GRCh38
rs571121774 Chr.1: 46675301-46675301 on Build GRCh38
rs937449003 Chr.1: 46675142-46675142 on Build GRCh38
rs585257 Chr.1: 46675113-46675113 on Build GRCh38
rs1036368135 Chr.1: 46675484-46675484 on Build GRCh38
rs905434961 Chr.1: 46675347-46675347 on Build GRCh38
rs898099139 Chr.1: 46675172-46675172 on Build GRCh38
rs200087519 Chr.1: 46675218-46675218 on Build GRCh38
rs1052772461 Chr.1: 46675277-46675277 on Build GRCh38
rs895560897 Chr.1: 46675281-46675281 on Build GRCh38
rs540393099 Chr.1: 46675214-46675214 on Build GRCh38
rs764040385 Chr.1: 46675502-46675502 on Build GRCh38
rs770856947 Chr.1: 46675324-46675324 on Build GRCh38
rs538161537 Chr.1: 46675382-46675382 on Build GRCh38
rs776776691 Chr.1: 46675327-46675327 on Build GRCh38
rs980932238 Chr.1: 46675103-46675104 on Build GRCh38

View Full Product Details