Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46676288-46676786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB14
Entrez Gene ID
9813
Gene Name
EF-hand calcium binding domain 14
Gene Aliases
KIAA0494
UniGene
Hs.707688
Gene Chromosome Location
Chr.1: 46675159-46719064 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Gene Symbol
EFCAB14-AS1
Entrez Gene ID
100130197
Gene Name
EFCAB14 antisense RNA 1
Gene Aliases
KIAA0494-AS1
UniGene
Hs.568643
Gene Chromosome Location
Chr.1: 46674036-46692098 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

SNP Detail

SNP ID Chromosome Location
rs1057210967 Chr.1: 46676412-46676412 on Build GRCh38
rs964986627 Chr.1: 46676629-46676629 on Build GRCh38
rs915506916 Chr.1: 46676656-46676656 on Build GRCh38
rs993342279 Chr.1: 46676575-46676575 on Build GRCh38
rs924383727 Chr.1: 46676373-46676373 on Build GRCh38
rs989157488 Chr.1: 46676403-46676403 on Build GRCh38
rs927026379 Chr.1: 46676556-46676556 on Build GRCh38
rs1054368379 Chr.1: 46676752-46676752 on Build GRCh38
rs955942967 Chr.1: 46676390-46676390 on Build GRCh38
rs889861446 Chr.1: 46676668-46676668 on Build GRCh38
rs938322942 Chr.1: 46676377-46676377 on Build GRCh38
rs984441354 Chr.1: 46676580-46676580 on Build GRCh38
rs978233591 Chr.1: 46676754-46676754 on Build GRCh38
rs558484689 Chr.1: 46676382-46676382 on Build GRCh38
rs990162572 Chr.1: 46676646-46676646 on Build GRCh38
rs906301485 Chr.1: 46676519-46676519 on Build GRCh38
rs867962716 Chr.1: 46676538-46676538 on Build GRCh38
rs531625182 Chr.1: 46676365-46676365 on Build GRCh38
rs566566696 Chr.1: 46676364-46676364 on Build GRCh38
rs1024244060 Chr.1: 46676454-46676454 on Build GRCh38
rs1000267119 Chr.1: 46676706-46676706 on Build GRCh38
rs537878330 Chr.1: 46676614-46676614 on Build GRCh38
rs979963489 Chr.1: 46676502-46676502 on Build GRCh38
rs971603824 Chr.1: 46676495-46676495 on Build GRCh38
rs947119914 Chr.1: 46676358-46676358 on Build GRCh38
rs1037525049 Chr.1: 46676570-46676570 on Build GRCh38
rs894386203 Chr.1: 46676431-46676431 on Build GRCh38
rs771214816 Chr.1: 46676660-46676660 on Build GRCh38
rs533875688 Chr.1: 46676372-46676372 on Build GRCh38
rs940384477 Chr.1: 46676614-46676614 on Build GRCh38
rs1008865054 Chr.1: 46676616-46676616 on Build GRCh38
rs574434768 Chr.1: 46676722-46676722 on Build GRCh38
rs998374561 Chr.1: 46676346-46676346 on Build GRCh38
rs899024272 Chr.1: 46676572-46676572 on Build GRCh38
rs778210160 Chr.1: 46676336-46676336 on Build GRCh38
rs1028790698 Chr.1: 46676361-46676361 on Build GRCh38
rs373324884 Chr.1: 46676608-46676608 on Build GRCh38
rs556172228 Chr.1: 46676623-46676623 on Build GRCh38
rs746845066 Chr.1: 46676618-46676618 on Build GRCh38
rs201303978 Chr.1: 46676609-46676610 on Build GRCh38

View Full Product Details