Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 46684399-46684896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB14
Entrez Gene ID
9813
Gene Name
EF-hand calcium binding domain 14
Gene Aliases
KIAA0494
UniGene
Hs.707688
Gene Chromosome Location
Chr.1: 46675159-46719064 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Gene Symbol
EFCAB14-AS1
Entrez Gene ID
100130197
Gene Name
EFCAB14 antisense RNA 1
Gene Aliases
KIAA0494-AS1
UniGene
Hs.568643
Gene Chromosome Location
Chr.1: 46674036-46692098 on Build GRCh38
Transcripts
NM_014774.2 NR_038827.1 NR_038828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445854 c.1186+3G>C p.? Chr.1: 46684488-46684488 on Build GRCh38
6394661 c.1180G>A p.D394N Chr.1: 46684497-46684497 on Build GRCh38
5599769 c.1078G>A p.E360K Chr.1: 46684599-46684599 on Build GRCh38
6398229 c.1172A>T p.E391V Chr.1: 46684505-46684505 on Build GRCh38
6290112 c.1135G>T p.A379S Chr.1: 46684542-46684542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773329606 Chr.1: 46684522-46684522 on Build GRCh38
rs758547678 Chr.1: 46684631-46684631 on Build GRCh38
rs753006251 Chr.1: 46684559-46684559 on Build GRCh38
rs915822099 Chr.1: 46684542-46684542 on Build GRCh38
rs951706231 Chr.1: 46684767-46684767 on Build GRCh38
rs763648233 Chr.1: 46684507-46684507 on Build GRCh38
rs749021694 Chr.1: 46684500-46684500 on Build GRCh38
rs754200074 Chr.1: 46684445-46684445 on Build GRCh38
rs188449602 Chr.1: 46684693-46684693 on Build GRCh38
rs1039048926 Chr.1: 46684718-46684718 on Build GRCh38
rs779637293 Chr.1: 46684498-46684498 on Build GRCh38
rs746062714 Chr.1: 46684647-46684647 on Build GRCh38
rs764563884 Chr.1: 46684570-46684570 on Build GRCh38
rs770054838 Chr.1: 46684652-46684652 on Build GRCh38
rs141207881 Chr.1: 46684801-46684801 on Build GRCh38
rs889765534 Chr.1: 46684531-46684531 on Build GRCh38
rs749255054 Chr.1: 46684789-46684790 on Build GRCh38
rs151155208 Chr.1: 46684687-46684687 on Build GRCh38
rs755582345 Chr.1: 46684575-46684575 on Build GRCh38
rs779696602 Chr.1: 46684576-46684576 on Build GRCh38
rs777578798 Chr.1: 46684612-46684612 on Build GRCh38
rs200934704 Chr.1: 46684511-46684511 on Build GRCh38
rs759848365 Chr.1: 46684545-46684545 on Build GRCh38
rs201689563 Chr.1: 46684497-46684497 on Build GRCh38
rs765677886 Chr.1: 46684458-46684458 on Build GRCh38
rs1047784373 Chr.1: 46684657-46684657 on Build GRCh38
rs764327591 Chr.1: 46684778-46684778 on Build GRCh38
rs376178646 Chr.1: 46684534-46684534 on Build GRCh38
rs367996064 Chr.1: 46684471-46684471 on Build GRCh38
rs748014271 Chr.1: 46684601-46684601 on Build GRCh38
rs199735021 Chr.1: 46684476-46684476 on Build GRCh38
rs753565759 Chr.1: 46684587-46684587 on Build GRCh38
rs763498441 Chr.1: 46684569-46684569 on Build GRCh38
rs371127428 Chr.1: 46684539-46684539 on Build GRCh38
rs776853477 Chr.1: 46684543-46684543 on Build GRCh38
rs907052051 Chr.1: 46684692-46684692 on Build GRCh38
rs140908900 Chr.1: 46684517-46684517 on Build GRCh38
rs375819291 Chr.1: 46684622-46684622 on Build GRCh38
rs80015621 Chr.1: 46684459-46684459 on Build GRCh38
rs756720119 Chr.1: 46684473-46684473 on Build GRCh38
rs189748464 Chr.1: 46684448-46684448 on Build GRCh38
rs200699927 Chr.1: 46684495-46684495 on Build GRCh38
rs1044383078 Chr.1: 46684850-46684850 on Build GRCh38
rs76664198 Chr.1: 46684432-46684432 on Build GRCh38
rs772042712 Chr.1: 46684604-46684604 on Build GRCh38
rs961634131 Chr.1: 46684842-46684842 on Build GRCh38
rs778819707 Chr.1: 46684588-46684588 on Build GRCh38
rs749947077 Chr.1: 46684477-46684477 on Build GRCh38
rs867325139 Chr.1: 46684527-46684530 on Build GRCh38
rs182780830 Chr.1: 46684510-46684510 on Build GRCh38
rs142055266 Chr.1: 46684501-46684501 on Build GRCh38
rs374519800 Chr.1: 46684573-46684573 on Build GRCh38
rs929147330 Chr.1: 46684640-46684640 on Build GRCh38
rs568685093 Chr.1: 46684685-46684685 on Build GRCh38
rs776795868 Chr.1: 46684635-46684635 on Build GRCh38
rs778944195 Chr.1: 46684505-46684505 on Build GRCh38
rs944677034 Chr.1: 46684717-46684717 on Build GRCh38
rs200025396 Chr.1: 46684513-46684513 on Build GRCh38
rs566596539 Chr.1: 46684796-46684796 on Build GRCh38
rs753105504 Chr.1: 46684564-46684564 on Build GRCh38
rs770940754 Chr.1: 46684626-46684626 on Build GRCh38

View Full Product Details