Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55032075-55032569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GP6
Entrez Gene ID
51206
Gene Name
glycoprotein VI platelet
Gene Aliases
BDPLT11, GPIV, GPVI
UniGene
Hs.661752
Gene Chromosome Location
Chr.19: 55013705-55038264 on Build GRCh38
Transcripts
NM_001083899.2 NM_001256017.2 NM_016363.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3414100 c.222C>T p.F74F Chr.19: 55032242-55032242 on Build GRCh38
3539323 c.148G>A p.G50R Chr.19: 55032316-55032316 on Build GRCh38
4512885 c.91C>T p.Q31* Chr.19: 55032373-55032373 on Build GRCh38
320628 c.325G>T p.G109* Chr.19: 55032139-55032139 on Build GRCh38
5765286 c.307C>A p.L103M Chr.19: 55032157-55032157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575231122 Chr.19: 55032148-55032148 on Build GRCh38
rs201025246 Chr.19: 55032513-55032513 on Build GRCh38
rs554527184 Chr.19: 55032203-55032203 on Build GRCh38
rs559176407 Chr.19: 55032215-55032215 on Build GRCh38
rs779634946 Chr.19: 55032490-55032490 on Build GRCh38
rs932070312 Chr.19: 55032499-55032499 on Build GRCh38
rs756955728 Chr.19: 55032484-55032484 on Build GRCh38
rs957967730 Chr.19: 55032322-55032322 on Build GRCh38
rs761400559 Chr.19: 55032181-55032181 on Build GRCh38
rs760145522 Chr.19: 55032471-55032471 on Build GRCh38
rs1014609232 Chr.19: 55032199-55032199 on Build GRCh38
rs776751773 Chr.19: 55032290-55032290 on Build GRCh38
rs771430932 Chr.19: 55032160-55032160 on Build GRCh38
rs759426349 Chr.19: 55032204-55032204 on Build GRCh38
rs752925351 Chr.19: 55032211-55032211 on Build GRCh38
rs146751606 Chr.19: 55032266-55032266 on Build GRCh38
rs750477581 Chr.19: 55032395-55032395 on Build GRCh38
rs888030341 Chr.19: 55032186-55032186 on Build GRCh38
rs745426989 Chr.19: 55032437-55032437 on Build GRCh38
rs1032125399 Chr.19: 55032501-55032501 on Build GRCh38
rs772847163 Chr.19: 55032273-55032273 on Build GRCh38
rs545004274 Chr.19: 55032188-55032188 on Build GRCh38
rs764293365 Chr.19: 55032446-55032446 on Build GRCh38
rs774416479 Chr.19: 55032484-55032485 on Build GRCh38
rs774089152 Chr.19: 55032179-55032179 on Build GRCh38
rs28385643 Chr.19: 55032367-55032367 on Build GRCh38
rs775648841 Chr.19: 55032426-55032445 on Build GRCh38
rs1671204 Chr.19: 55032455-55032455 on Build GRCh38
rs773349443 Chr.19: 55032523-55032523 on Build GRCh38
rs368001192 Chr.19: 55032473-55032473 on Build GRCh38
rs745762025 Chr.19: 55032362-55032362 on Build GRCh38
rs751209179 Chr.19: 55032424-55032424 on Build GRCh38
rs28385644 Chr.19: 55032245-55032245 on Build GRCh38
rs113219102 Chr.19: 55032327-55032327 on Build GRCh38
rs528105487 Chr.19: 55032096-55032096 on Build GRCh38
rs528065959 Chr.19: 55032460-55032460 on Build GRCh38
rs749915633 Chr.19: 55032488-55032488 on Build GRCh38
rs746184218 Chr.19: 55032242-55032242 on Build GRCh38
rs28969501 Chr.19: 55032157-55032157 on Build GRCh38
rs775341524 Chr.19: 55032379-55032379 on Build GRCh38
rs769686181 Chr.19: 55032378-55032378 on Build GRCh38
rs780748321 Chr.19: 55032430-55032430 on Build GRCh38
rs199588110 Chr.19: 55032292-55032292 on Build GRCh38
rs370941526 Chr.19: 55032173-55032173 on Build GRCh38
rs375999257 Chr.19: 55032459-55032459 on Build GRCh38
rs1025596226 Chr.19: 55032297-55032297 on Build GRCh38
rs769747276 Chr.19: 55032438-55032438 on Build GRCh38
rs746487471 Chr.19: 55032353-55032353 on Build GRCh38
rs764355964 Chr.19: 55032309-55032309 on Build GRCh38
rs770869085 Chr.19: 55032525-55032525 on Build GRCh38
rs767613010 Chr.19: 55032316-55032316 on Build GRCh38
rs567724105 Chr.19: 55032146-55032146 on Build GRCh38
rs773333506 Chr.19: 55032461-55032461 on Build GRCh38
rs987990282 Chr.19: 55032337-55032337 on Build GRCh38
rs775151408 Chr.19: 55032305-55032305 on Build GRCh38
rs758112169 Chr.19: 55032343-55032343 on Build GRCh38
rs767523302 Chr.19: 55032393-55032393 on Build GRCh38
rs995736633 Chr.19: 55032293-55032293 on Build GRCh38
rs773719373 Chr.19: 55032107-55032107 on Build GRCh38
rs201283598 Chr.19: 55032382-55032382 on Build GRCh38
rs758571097 Chr.19: 55032213-55032213 on Build GRCh38
rs951109424 Chr.19: 55032124-55032124 on Build GRCh38
rs374068596 Chr.19: 55032449-55032449 on Build GRCh38
rs572860909 Chr.19: 55032163-55032163 on Build GRCh38
rs370323646 Chr.19: 55032429-55032429 on Build GRCh38
rs774514319 Chr.19: 55032387-55032387 on Build GRCh38
rs749163054 Chr.19: 55032497-55032497 on Build GRCh38
rs1050599191 Chr.19: 55032406-55032406 on Build GRCh38
rs763231845 Chr.19: 55032308-55032308 on Build GRCh38
rs28385645 Chr.19: 55032227-55032227 on Build GRCh38
rs760718784 Chr.19: 55032465-55032465 on Build GRCh38
rs374599974 Chr.19: 55032149-55032149 on Build GRCh38
rs561370230 Chr.19: 55032291-55032291 on Build GRCh38
rs770526790 Chr.19: 55032358-55032358 on Build GRCh38
rs762938065 Chr.19: 55032432-55032433 on Build GRCh38
rs774141069 Chr.19: 55032458-55032458 on Build GRCh38
rs369642453 Chr.19: 55032514-55032514 on Build GRCh38
rs756588702 Chr.19: 55032325-55032325 on Build GRCh38
rs2288905 Chr.19: 55032466-55032466 on Build GRCh38
rs753693386 Chr.19: 55032128-55032128 on Build GRCh38
rs1002260406 Chr.19: 55032284-55032284 on Build GRCh38
rs370005974 Chr.19: 55032122-55032122 on Build GRCh38
rs200403102 Chr.19: 55032281-55032281 on Build GRCh38
rs757364878 Chr.19: 55032315-55032315 on Build GRCh38
rs757745719 Chr.19: 55032236-55032236 on Build GRCh38
rs936938328 Chr.19: 55032183-55032183 on Build GRCh38
rs778394293 Chr.19: 55032097-55032097 on Build GRCh38
rs771558250 Chr.19: 55032187-55032187 on Build GRCh38
rs752698122 Chr.19: 55032140-55032140 on Build GRCh38
rs937978607 Chr.19: 55032450-55032450 on Build GRCh38
rs765902536 Chr.19: 55032544-55032544 on Build GRCh38
rs771631715 Chr.19: 55032269-55032269 on Build GRCh38
rs369128928 Chr.19: 55032110-55032110 on Build GRCh38
rs1019203799 Chr.19: 55032376-55032376 on Build GRCh38
rs776994233 Chr.19: 55032105-55032105 on Build GRCh38
rs200424845 Chr.19: 55032222-55032222 on Build GRCh38
rs866721211 Chr.19: 55032237-55032237 on Build GRCh38
rs777620820 Chr.19: 55032347-55032347 on Build GRCh38
rs766494410 Chr.19: 55032192-55032192 on Build GRCh38
rs780301940 Chr.19: 55032258-55032258 on Build GRCh38
rs779255434 Chr.19: 55032139-55032139 on Build GRCh38
rs778679076 Chr.19: 55032511-55032511 on Build GRCh38
rs760053904 Chr.19: 55032542-55032542 on Build GRCh38
rs925006141 Chr.19: 55032389-55032389 on Build GRCh38
rs751615170 Chr.19: 55032310-55032310 on Build GRCh38
rs200428516 Chr.19: 55032106-55032106 on Build GRCh38
rs76221126 Chr.19: 55032120-55032120 on Build GRCh38
rs764268085 Chr.19: 55032384-55032384 on Build GRCh38
rs1025144124 Chr.19: 55032279-55032279 on Build GRCh38
rs771246475 Chr.19: 55032104-55032104 on Build GRCh38
rs764106486 Chr.19: 55032408-55032408 on Build GRCh38
rs776785166 Chr.19: 55032534-55032534 on Build GRCh38
rs189503167 Chr.19: 55032239-55032239 on Build GRCh38
rs750889036 Chr.19: 55032324-55032324 on Build GRCh38
rs374449011 Chr.19: 55032503-55032503 on Build GRCh38
rs781531121 Chr.19: 55032161-55032161 on Build GRCh38
rs186278363 Chr.19: 55032474-55032474 on Build GRCh38
rs374599246 Chr.19: 55032412-55032412 on Build GRCh38
rs761244764 Chr.19: 55032396-55032396 on Build GRCh38
rs747344218 Chr.19: 55032099-55032099 on Build GRCh38
rs115456071 Chr.19: 55032121-55032121 on Build GRCh38
rs780756092 Chr.19: 55032486-55032486 on Build GRCh38
rs768168959 Chr.19: 55032164-55032164 on Build GRCh38
rs1671205 Chr.19: 55032467-55032467 on Build GRCh38
rs757111978 Chr.19: 55032439-55032439 on Build GRCh38
rs753563209 Chr.19: 55032411-55032411 on Build GRCh38
rs199731675 Chr.19: 55032489-55032489 on Build GRCh38
rs748269983 Chr.19: 55032524-55032524 on Build GRCh38
rs372189835 Chr.19: 55032399-55032399 on Build GRCh38
rs768388261 Chr.19: 55032456-55032456 on Build GRCh38
rs773976878 Chr.19: 55032390-55032390 on Build GRCh38
rs749686530 Chr.19: 55032339-55032339 on Build GRCh38
rs780402836 Chr.19: 55032330-55032330 on Build GRCh38
rs755152911 Chr.19: 55032129-55032129 on Build GRCh38
rs368522136 Chr.19: 55032392-55032392 on Build GRCh38
rs975105725 Chr.19: 55032386-55032386 on Build GRCh38
rs200313847 Chr.19: 55032360-55032360 on Build GRCh38

View Full Product Details