Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112977951-112978456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL36G
Entrez Gene ID
56300
Gene Name
interleukin 36, gamma
Gene Aliases
IL-1F9, IL-1H1, IL-1RP2, IL1E, IL1F9, IL1H1, IL1RP2
UniGene
Hs.211238
Gene Chromosome Location
Chr.2: 112978019-112985672 on Build GRCh38
Transcripts
NM_001278568.1 NM_019618.3

SNP Detail

SNP ID Chromosome Location
rs941629887 Chr.2: 112978289-112978289 on Build GRCh38
rs755728141 Chr.2: 112978294-112978294 on Build GRCh38
rs761630976 Chr.2: 112978281-112978281 on Build GRCh38
rs977996545 Chr.2: 112978323-112978323 on Build GRCh38
rs207462177 Chr.2: 112978262-112978262 on Build GRCh38
rs764108759 Chr.2: 112978241-112978241 on Build GRCh38
rs189336460 Chr.2: 112977988-112977988 on Build GRCh38
rs925285616 Chr.2: 112978324-112978324 on Build GRCh38
rs918780559 Chr.2: 112978302-112978302 on Build GRCh38
rs1055181583 Chr.2: 112978137-112978137 on Build GRCh38
rs769805137 Chr.2: 112978063-112978063 on Build GRCh38
rs936659110 Chr.2: 112978371-112978371 on Build GRCh38
rs905947637 Chr.2: 112978415-112978415 on Build GRCh38
rs1020157970 Chr.2: 112978173-112978173 on Build GRCh38
rs976275435 Chr.2: 112978256-112978256 on Build GRCh38
rs905901323 Chr.2: 112978071-112978071 on Build GRCh38
rs553510365 Chr.2: 112978127-112978127 on Build GRCh38
rs765869468 Chr.2: 112978315-112978315 on Build GRCh38
rs957290507 Chr.2: 112978110-112978110 on Build GRCh38
rs1001999674 Chr.2: 112978425-112978425 on Build GRCh38
rs569073954 Chr.2: 112978046-112978046 on Build GRCh38
rs771991826 Chr.2: 112978052-112978052 on Build GRCh38
rs182168986 Chr.2: 112978090-112978090 on Build GRCh38
rs950245587 Chr.2: 112978349-112978349 on Build GRCh38
rs897385582 Chr.2: 112978195-112978195 on Build GRCh38
rs1026396450 Chr.2: 112978232-112978232 on Build GRCh38
rs529192985 Chr.2: 112978012-112978012 on Build GRCh38
rs1033384769 Chr.2: 112978104-112978104 on Build GRCh38
rs549217131 Chr.2: 112978026-112978026 on Build GRCh38
rs572471279 Chr.2: 112978394-112978394 on Build GRCh38
rs147627165 Chr.2: 112978287-112978287 on Build GRCh38
rs771352696 Chr.2: 112978032-112978032 on Build GRCh38
rs185907526 Chr.2: 112978194-112978194 on Build GRCh38
rs1001549044 Chr.2: 112978079-112978079 on Build GRCh38
rs28947205 Chr.2: 112978286-112978286 on Build GRCh38
rs910387634 Chr.2: 112978022-112978022 on Build GRCh38
rs538034732 Chr.2: 112978064-112978064 on Build GRCh38
rs556436053 Chr.2: 112978283-112978283 on Build GRCh38
rs925431485 Chr.2: 112978094-112978094 on Build GRCh38
rs1042774640 Chr.2: 112978066-112978066 on Build GRCh38
rs774091570 Chr.2: 112978261-112978261 on Build GRCh38
rs951706033 Chr.2: 112978247-112978247 on Build GRCh38
rs868023735 Chr.2: 112978433-112978433 on Build GRCh38
rs1017279193 Chr.2: 112978258-112978258 on Build GRCh38
rs910142619 Chr.2: 112978011-112978011 on Build GRCh38
rs12328178 Chr.2: 112978391-112978391 on Build GRCh38
rs533811367 Chr.2: 112978093-112978093 on Build GRCh38
rs993209689 Chr.2: 112978215-112978215 on Build GRCh38
rs551540413 Chr.2: 112978087-112978087 on Build GRCh38
rs572567847 Chr.2: 112978035-112978035 on Build GRCh38
rs145291752 Chr.2: 112978169-112978169 on Build GRCh38

View Full Product Details