Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 113005770-113006265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL36A
Entrez Gene ID
27179
Gene Name
interleukin 36, alpha
Gene Aliases
FIL1, FIL1(EPSILON), FIL1E, IL-1F6, IL1(EPSILON), IL1F6
UniGene
Hs.278910
Gene Chromosome Location
Chr.2: 113005461-113011071 on Build GRCh38
Transcripts
NM_014440.2 XM_005263639.2 XM_011510965.1 XM_017003806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5609143 c.10G>A p.A4T Chr.2: 113005881-113005881 on Build GRCh38
5603798 c.68G>A p.W23* Chr.2: 113006031-113006031 on Build GRCh38
5532688 c.15G>A p.L5L Chr.2: 113005978-113005978 on Build GRCh38
5576716 c.76C>T p.Q26* Chr.2: 113006039-113006039 on Build GRCh38
6509655 c.58C>A p.H20N Chr.2: 113006021-113006021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551561855 Chr.2: 113006147-113006147 on Build GRCh38
rs766864713 Chr.2: 113005904-113005904 on Build GRCh38
rs2305152 Chr.2: 113005886-113005886 on Build GRCh38
rs776581757 Chr.2: 113005893-113005893 on Build GRCh38
rs372549382 Chr.2: 113006119-113006119 on Build GRCh38
rs981555967 Chr.2: 113005792-113005792 on Build GRCh38
rs777920919 Chr.2: 113006020-113006020 on Build GRCh38
rs202001688 Chr.2: 113006076-113006076 on Build GRCh38
rs540129063 Chr.2: 113005810-113005810 on Build GRCh38
rs763397803 Chr.2: 113005841-113005841 on Build GRCh38
rs766240065 Chr.2: 113006072-113006072 on Build GRCh38
rs750529063 Chr.2: 113006091-113006092 on Build GRCh38
rs761956828 Chr.2: 113005826-113005826 on Build GRCh38
rs752169360 Chr.2: 113005843-113005843 on Build GRCh38
rs1047390586 Chr.2: 113006208-113006208 on Build GRCh38
rs955699539 Chr.2: 113006101-113006101 on Build GRCh38
rs200675990 Chr.2: 113005816-113005816 on Build GRCh38
rs765176889 Chr.2: 113005948-113005949 on Build GRCh38
rs375931124 Chr.2: 113005842-113005842 on Build GRCh38
rs764931788 Chr.2: 113006064-113006064 on Build GRCh38
rs755687109 Chr.2: 113005844-113005844 on Build GRCh38
rs979825533 Chr.2: 113006213-113006213 on Build GRCh38
rs1011231828 Chr.2: 113006009-113006009 on Build GRCh38
rs749638456 Chr.2: 113006021-113006021 on Build GRCh38
rs555679914 Chr.2: 113005911-113005911 on Build GRCh38
rs544436443 Chr.2: 113005992-113005992 on Build GRCh38
rs778721171 Chr.2: 113005858-113005858 on Build GRCh38
rs137983289 Chr.2: 113006024-113006024 on Build GRCh38
rs370538371 Chr.2: 113005850-113005850 on Build GRCh38
rs780011138 Chr.2: 113005997-113005997 on Build GRCh38
rs745391211 Chr.2: 113005991-113005991 on Build GRCh38
rs895497 Chr.2: 113005998-113005998 on Build GRCh38
rs907051463 Chr.2: 113006210-113006210 on Build GRCh38
rs201497922 Chr.2: 113005817-113005817 on Build GRCh38
rs915454213 Chr.2: 113005896-113005896 on Build GRCh38
rs756913946 Chr.2: 113005949-113005949 on Build GRCh38
rs763592530 Chr.2: 113005939-113005939 on Build GRCh38
rs539941311 Chr.2: 113006049-113006049 on Build GRCh38
rs367615872 Chr.2: 113005918-113005918 on Build GRCh38
rs748221855 Chr.2: 113006008-113006008 on Build GRCh38
rs577725399 Chr.2: 113005861-113005861 on Build GRCh38
rs377053319 Chr.2: 113006086-113006086 on Build GRCh38
rs771173093 Chr.2: 113006105-113006105 on Build GRCh38
rs926929076 Chr.2: 113005945-113005945 on Build GRCh38
rs367920192 Chr.2: 113006000-113006000 on Build GRCh38
rs773599574 Chr.2: 113005897-113005897 on Build GRCh38
rs1002636888 Chr.2: 113005966-113005966 on Build GRCh38
rs760960192 Chr.2: 113006180-113006180 on Build GRCh38
rs1034024181 Chr.2: 113005796-113005796 on Build GRCh38
rs956972756 Chr.2: 113006134-113006134 on Build GRCh38
rs149472911 Chr.2: 113006099-113006099 on Build GRCh38
rs776177247 Chr.2: 113006054-113006054 on Build GRCh38
rs753319244 Chr.2: 113005947-113005947 on Build GRCh38
rs1057407087 Chr.2: 113005970-113005970 on Build GRCh38
rs776973376 Chr.2: 113005821-113005821 on Build GRCh38
rs752661666 Chr.2: 113005800-113005800 on Build GRCh38
rs374841383 Chr.2: 113006133-113006133 on Build GRCh38
rs894220144 Chr.2: 113006240-113006240 on Build GRCh38
rs1022587462 Chr.2: 113006037-113006037 on Build GRCh38
rs200879333 Chr.2: 113005894-113005894 on Build GRCh38
rs765575924 Chr.2: 113005837-113005837 on Build GRCh38
rs774759023 Chr.2: 113006071-113006071 on Build GRCh38
rs374285811 Chr.2: 113005859-113005859 on Build GRCh38
rs868045893 Chr.2: 113006216-113006216 on Build GRCh38
rs761298474 Chr.2: 113006057-113006057 on Build GRCh38
rs756864026 Chr.2: 113005852-113005852 on Build GRCh38
rs764837494 Chr.2: 113005978-113005978 on Build GRCh38
rs374607104 Chr.2: 113005903-113005903 on Build GRCh38
rs780927160 Chr.2: 113005956-113005956 on Build GRCh38
rs371448929 Chr.2: 113005961-113005961 on Build GRCh38
rs927077577 Chr.2: 113006215-113006215 on Build GRCh38
rs1003099089 Chr.2: 113006227-113006227 on Build GRCh38
rs760048472 Chr.2: 113005936-113005936 on Build GRCh38
rs1055575158 Chr.2: 113006235-113006235 on Build GRCh38
rs751426211 Chr.2: 113006075-113006075 on Build GRCh38
rs533989566 Chr.2: 113005884-113005884 on Build GRCh38
rs750241837 Chr.2: 113005981-113005981 on Build GRCh38
rs370196932 Chr.2: 113006112-113006112 on Build GRCh38
rs781027848 Chr.2: 113006085-113006085 on Build GRCh38
rs766617384 Chr.2: 113006189-113006189 on Build GRCh38
rs929900268 Chr.2: 113006198-113006198 on Build GRCh38
rs201500964 Chr.2: 113006111-113006111 on Build GRCh38
rs770023543 Chr.2: 113006018-113006018 on Build GRCh38
rs866220179 Chr.2: 113005875-113005875 on Build GRCh38
rs372617143 Chr.2: 113006100-113006100 on Build GRCh38
rs759995735 Chr.2: 113006027-113006027 on Build GRCh38
rs750041642 Chr.2: 113006067-113006067 on Build GRCh38
rs531844877 Chr.2: 113006136-113006136 on Build GRCh38
rs1025475176 Chr.2: 113005942-113005942 on Build GRCh38
rs1057364181 Chr.2: 113005803-113005803 on Build GRCh38
rs182845963 Chr.2: 113006025-113006025 on Build GRCh38
rs775920306 Chr.2: 113006132-113006132 on Build GRCh38

View Full Product Details