Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9892459-9893001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAGN1
Entrez Gene ID
84522
Gene Name
jagunal homolog 1
Gene Aliases
GL009, SCN6
UniGene
Hs.24054
Gene Chromosome Location
Chr.3: 9890587-9894349 on Build GRCh38
Transcripts
NM_032492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1049296 c.126C>T p.Y42Y Chr.3: 9892951-9892951 on Build GRCh38
1049295 c.91G>T p.V31L Chr.3: 9892916-9892916 on Build GRCh38
3993355 c.167G>A p.S56N Chr.3: 9892992-9892992 on Build GRCh38
6386630 c.119T>A p.L40Q Chr.3: 9892944-9892944 on Build GRCh38
132711 c.124T>C p.Y42H Chr.3: 9892949-9892949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931673377 Chr.3: 9892847-9892847 on Build GRCh38
rs140024249 Chr.3: 9892948-9892948 on Build GRCh38
rs279554 Chr.3: 9892547-9892547 on Build GRCh38
rs772221562 Chr.3: 9892890-9892890 on Build GRCh38
rs941911283 Chr.3: 9892643-9892643 on Build GRCh38
rs764869404 Chr.3: 9892603-9892603 on Build GRCh38
rs1008427440 Chr.3: 9892810-9892810 on Build GRCh38
rs537497964 Chr.3: 9892898-9892898 on Build GRCh38
rs941630668 Chr.3: 9892604-9892604 on Build GRCh38
rs745932619 Chr.3: 9892960-9892960 on Build GRCh38
rs753747801 Chr.3: 9892949-9892949 on Build GRCh38
rs898850500 Chr.3: 9892619-9892619 on Build GRCh38
rs1033409404 Chr.3: 9892738-9892738 on Build GRCh38
rs917371420 Chr.3: 9892777-9892777 on Build GRCh38
rs780119317 Chr.3: 9892977-9892977 on Build GRCh38
rs180988466 Chr.3: 9892899-9892899 on Build GRCh38
rs763900045 Chr.3: 9892942-9892942 on Build GRCh38
rs139681239 Chr.3: 9892678-9892678 on Build GRCh38
rs773759588 Chr.3: 9892816-9892816 on Build GRCh38
rs568674262 Chr.3: 9892770-9892773 on Build GRCh38
rs279553 Chr.3: 9892921-9892921 on Build GRCh38
rs890288480 Chr.3: 9892644-9892644 on Build GRCh38
rs758380213 Chr.3: 9892737-9892737 on Build GRCh38
rs530437210 Chr.3: 9892635-9892635 on Build GRCh38
rs371216049 Chr.3: 9892888-9892888 on Build GRCh38
rs533811445 Chr.3: 9892799-9892799 on Build GRCh38
rs150624071 Chr.3: 9892488-9892488 on Build GRCh38
rs866615076 Chr.3: 9892564-9892564 on Build GRCh38
rs868799195 Chr.3: 9892525-9892525 on Build GRCh38
rs372730920 Chr.3: 9892702-9892702 on Build GRCh38
rs775993295 Chr.3: 9892894-9892894 on Build GRCh38
rs941586594 Chr.3: 9892807-9892807 on Build GRCh38
rs187981807 Chr.3: 9892895-9892895 on Build GRCh38
rs938580278 Chr.3: 9892633-9892633 on Build GRCh38
rs1028778947 Chr.3: 9892639-9892639 on Build GRCh38
rs528473640 Chr.3: 9892963-9892963 on Build GRCh38
rs778975542 Chr.3: 9892952-9892952 on Build GRCh38
rs1055596292 Chr.3: 9892636-9892636 on Build GRCh38
rs369545172 Chr.3: 9892912-9892912 on Build GRCh38
rs1055831114 Chr.3: 9892502-9892502 on Build GRCh38
rs2279795 Chr.3: 9892744-9892744 on Build GRCh38
rs563021924 Chr.3: 9892588-9892588 on Build GRCh38
rs765992554 Chr.3: 9892910-9892910 on Build GRCh38
rs587777728 Chr.3: 9892955-9892955 on Build GRCh38
rs759750222 Chr.3: 9892673-9892673 on Build GRCh38
rs754539279 Chr.3: 9892896-9892897 on Build GRCh38
rs982881465 Chr.3: 9892782-9892782 on Build GRCh38
rs371744656 Chr.3: 9892897-9892897 on Build GRCh38
rs914352652 Chr.3: 9892523-9892523 on Build GRCh38
rs201260563 Chr.3: 9892927-9892927 on Build GRCh38
rs898817022 Chr.3: 9892828-9892828 on Build GRCh38
rs373761568 Chr.3: 9892712-9892712 on Build GRCh38
rs894290002 Chr.3: 9892640-9892640 on Build GRCh38
rs201733831 Chr.3: 9892896-9892896 on Build GRCh38
rs1047300322 Chr.3: 9892549-9892549 on Build GRCh38
rs945682983 Chr.3: 9892538-9892538 on Build GRCh38
rs375147795 Chr.3: 9892623-9892623 on Build GRCh38
rs1037746653 Chr.3: 9892609-9892609 on Build GRCh38
rs201225696 Chr.3: 9892915-9892915 on Build GRCh38
rs762370289 Chr.3: 9892906-9892906 on Build GRCh38
rs964542016 Chr.3: 9892859-9892859 on Build GRCh38
rs140564109 Chr.3: 9892951-9892951 on Build GRCh38
rs898608436 Chr.3: 9892705-9892705 on Build GRCh38
rs746766942 Chr.3: 9892749-9892749 on Build GRCh38
rs937398459 Chr.3: 9892490-9892490 on Build GRCh38
rs778920284 Chr.3: 9892887-9892887 on Build GRCh38
rs950142889 Chr.3: 9892778-9892780 on Build GRCh38
rs990304283 Chr.3: 9892516-9892516 on Build GRCh38
rs964278584 Chr.3: 9892483-9892483 on Build GRCh38
rs758662868 Chr.3: 9892969-9892969 on Build GRCh38

View Full Product Details