Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28533876-28534371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX5
Entrez Gene ID
2880
Gene Name
glutathione peroxidase 5
Gene Aliases
HEL-S-75p
UniGene
Hs.248129
Gene Chromosome Location
Chr.6: 28526012-28534951 on Build GRCh38
Transcripts
NM_001509.2 NM_003996.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6483023 c.616T>A p.S206T Chr.6: 28534117-28534117 on Build GRCh38
400033 c.626C>A p.T209K Chr.6: 28534127-28534127 on Build GRCh38
6674729 c.491G>A p.G164D Chr.6: 28533992-28533992 on Build GRCh38
357041 c.658A>C p.T220P Chr.6: 28534159-28534159 on Build GRCh38
6674730 c.609G>A p.T203T Chr.6: 28534110-28534110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889389755 Chr.6: 28534011-28534011 on Build GRCh38
rs749579808 Chr.6: 28534199-28534199 on Build GRCh38
rs986317860 Chr.6: 28534345-28534345 on Build GRCh38
rs944681849 Chr.6: 28534012-28534012 on Build GRCh38
rs946841540 Chr.6: 28534302-28534302 on Build GRCh38
rs772777598 Chr.6: 28534041-28534041 on Build GRCh38
rs180855511 Chr.6: 28534261-28534261 on Build GRCh38
rs199538306 Chr.6: 28534001-28534001 on Build GRCh38
rs753534946 Chr.6: 28534074-28534074 on Build GRCh38
rs371361550 Chr.6: 28534109-28534109 on Build GRCh38
rs868153995 Chr.6: 28534167-28534167 on Build GRCh38
rs766042467 Chr.6: 28534063-28534063 on Build GRCh38
rs368865536 Chr.6: 28534190-28534190 on Build GRCh38
rs897868936 Chr.6: 28534082-28534082 on Build GRCh38
rs567969552 Chr.6: 28534110-28534110 on Build GRCh38
rs771083393 Chr.6: 28533968-28533968 on Build GRCh38
rs140888955 Chr.6: 28534092-28534092 on Build GRCh38
rs372426231 Chr.6: 28534106-28534106 on Build GRCh38
rs956658321 Chr.6: 28534234-28534234 on Build GRCh38
rs765613212 Chr.6: 28534090-28534090 on Build GRCh38
rs752032816 Chr.6: 28534166-28534166 on Build GRCh38
rs974385255 Chr.6: 28534281-28534281 on Build GRCh38
rs367937430 Chr.6: 28533949-28533949 on Build GRCh38
rs780148040 Chr.6: 28534197-28534197 on Build GRCh38
rs778266328 Chr.6: 28534096-28534096 on Build GRCh38
rs779211755 Chr.6: 28533917-28533917 on Build GRCh38
rs137907958 Chr.6: 28533992-28533992 on Build GRCh38
rs374429132 Chr.6: 28534127-28534127 on Build GRCh38
rs538763073 Chr.6: 28534303-28534303 on Build GRCh38
rs1002189036 Chr.6: 28534101-28534101 on Build GRCh38
rs868415334 Chr.6: 28533976-28533976 on Build GRCh38
rs28402737 Chr.6: 28534324-28534324 on Build GRCh38
rs1047506303 Chr.6: 28534304-28534304 on Build GRCh38
rs759150125 Chr.6: 28534088-28534088 on Build GRCh38
rs887288464 Chr.6: 28534087-28534087 on Build GRCh38
rs774065092 Chr.6: 28534039-28534039 on Build GRCh38
rs531172601 Chr.6: 28534139-28534139 on Build GRCh38
rs915401727 Chr.6: 28534258-28534258 on Build GRCh38
rs747189329 Chr.6: 28534113-28534113 on Build GRCh38
rs780349667 Chr.6: 28534102-28534102 on Build GRCh38
rs764802592 Chr.6: 28534146-28534146 on Build GRCh38
rs781296013 Chr.6: 28534337-28534337 on Build GRCh38
rs536914105 Chr.6: 28534114-28534114 on Build GRCh38
rs1051721410 Chr.6: 28533921-28533921 on Build GRCh38
rs865861025 Chr.6: 28534067-28534067 on Build GRCh38
rs764511160 Chr.6: 28534158-28534158 on Build GRCh38
rs911841599 Chr.6: 28534223-28534223 on Build GRCh38
rs753024954 Chr.6: 28534091-28534091 on Build GRCh38
rs1034726818 Chr.6: 28534214-28534214 on Build GRCh38
rs759178505 Chr.6: 28534144-28534144 on Build GRCh38
rs555265561 Chr.6: 28533903-28533903 on Build GRCh38
rs534236562 Chr.6: 28534178-28534178 on Build GRCh38
rs920262385 Chr.6: 28534295-28534295 on Build GRCh38
rs768471015 Chr.6: 28534032-28534032 on Build GRCh38
rs757812985 Chr.6: 28534174-28534174 on Build GRCh38
rs371805986 Chr.6: 28533960-28533960 on Build GRCh38
rs760272553 Chr.6: 28534062-28534062 on Build GRCh38
rs781379923 Chr.6: 28533970-28533970 on Build GRCh38
rs763215871 Chr.6: 28534156-28534156 on Build GRCh38
rs755356054 Chr.6: 28533912-28533912 on Build GRCh38
rs761764258 Chr.6: 28534040-28534040 on Build GRCh38
rs28382610 Chr.6: 28534134-28534134 on Build GRCh38
rs943282155 Chr.6: 28534248-28534248 on Build GRCh38
rs758080094 Chr.6: 28534103-28534104 on Build GRCh38
rs756289630 Chr.6: 28534184-28534184 on Build GRCh38
rs28382609 Chr.6: 28533937-28533937 on Build GRCh38
rs757328065 Chr.6: 28534105-28534105 on Build GRCh38
rs777791304 Chr.6: 28533944-28533944 on Build GRCh38

View Full Product Details