Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 29829738-29830241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-G
Entrez Gene ID
3135
Gene Name
major histocompatibility complex, class I, G
Gene Aliases
MHC-G
UniGene
Hs.512152
Gene Chromosome Location
Chr.6: 29826967-29831130 on Build GRCh38
Transcripts
NM_002127.5 XM_017010816.1 XM_017010817.1 XM_017010818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4992686 c.920C>T p.P307L Chr.6: 29829840-29829840 on Build GRCh38
1559748 c.991C>A p.L331M Chr.6: 29829911-29829911 on Build GRCh38
1559747 c.982G>C p.A328P Chr.6: 29829902-29829902 on Build GRCh38
1559746 c.977C>T p.A326V Chr.6: 29829897-29829897 on Build GRCh38
1559745 c.956C>G p.A319G Chr.6: 29829876-29829876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779746040 Chr.6: 29829863-29829863 on Build GRCh38
rs766671384 Chr.6: 29829819-29829819 on Build GRCh38
rs1043968312 Chr.6: 29830034-29830034 on Build GRCh38
rs774006543 Chr.6: 29829789-29829789 on Build GRCh38
rs761503504 Chr.6: 29829802-29829802 on Build GRCh38
rs913892318 Chr.6: 29829991-29829991 on Build GRCh38
rs144724454 Chr.6: 29829814-29829814 on Build GRCh38
rs553193145 Chr.6: 29829845-29829845 on Build GRCh38
rs757425434 Chr.6: 29829972-29829972 on Build GRCh38
rs772870109 Chr.6: 29829914-29829914 on Build GRCh38
rs573045964 Chr.6: 29829850-29829850 on Build GRCh38
rs752741808 Chr.6: 29829937-29829937 on Build GRCh38
rs761985705 Chr.6: 29830120-29830121 on Build GRCh38
rs758548929 Chr.6: 29829843-29829843 on Build GRCh38
rs758613787 Chr.6: 29829851-29829851 on Build GRCh38
rs1611627 Chr.6: 29830005-29830005 on Build GRCh38
rs1632933 Chr.6: 29830156-29830156 on Build GRCh38
rs536606226 Chr.6: 29829783-29829783 on Build GRCh38
rs746175910 Chr.6: 29829782-29829782 on Build GRCh38
rs904016712 Chr.6: 29830078-29830078 on Build GRCh38
rs1130363 Chr.6: 29829919-29829919 on Build GRCh38
rs865963836 Chr.6: 29829933-29829933 on Build GRCh38
rs1007705583 Chr.6: 29830080-29830080 on Build GRCh38
rs768731691 Chr.6: 29829897-29829897 on Build GRCh38
rs557607229 Chr.6: 29829765-29829765 on Build GRCh38
rs141130133 Chr.6: 29829869-29829869 on Build GRCh38
rs778011464 Chr.6: 29829853-29829853 on Build GRCh38
rs1025941295 Chr.6: 29830177-29830177 on Build GRCh38
rs1056432783 Chr.6: 29829988-29829988 on Build GRCh38
rs765798458 Chr.6: 29829932-29829932 on Build GRCh38
rs1632937 Chr.6: 29829776-29829776 on Build GRCh38
rs796748483 Chr.6: 29829888-29829888 on Build GRCh38
rs761148652 Chr.6: 29829817-29829817 on Build GRCh38
rs1736912 Chr.6: 29830174-29830174 on Build GRCh38
rs777549945 Chr.6: 29829944-29829944 on Build GRCh38
rs939338724 Chr.6: 29829983-29829983 on Build GRCh38
rs564772039 Chr.6: 29829898-29829898 on Build GRCh38
rs1026642003 Chr.6: 29829846-29829846 on Build GRCh38
rs546622677 Chr.6: 29830154-29830154 on Build GRCh38
rs773666498 Chr.6: 29829766-29829766 on Build GRCh38
rs950382076 Chr.6: 29830217-29830217 on Build GRCh38
rs556746961 Chr.6: 29829927-29829927 on Build GRCh38
rs760023923 Chr.6: 29829931-29829931 on Build GRCh38
rs745630585 Chr.6: 29829854-29829854 on Build GRCh38
rs533677430 Chr.6: 29829766-29829766 on Build GRCh38
rs771824819 Chr.6: 29829904-29829904 on Build GRCh38
rs762821306 Chr.6: 29829781-29829781 on Build GRCh38
rs544200079 Chr.6: 29829973-29829973 on Build GRCh38
rs747314068 Chr.6: 29829902-29829902 on Build GRCh38
rs1049033 Chr.6: 29829862-29829862 on Build GRCh38
rs778906176 Chr.6: 29829848-29829848 on Build GRCh38
rs745741578 Chr.6: 29829767-29829767 on Build GRCh38
rs777740260 Chr.6: 29829949-29829949 on Build GRCh38
rs769588222 Chr.6: 29829769-29829769 on Build GRCh38
rs1632934 Chr.6: 29830122-29830122 on Build GRCh38
rs766696298 Chr.6: 29829922-29829922 on Build GRCh38

View Full Product Details