Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30908074-30908586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H4
Entrez Gene ID
2968
Gene Name
general transcription factor IIH subunit 4
Gene Aliases
P52, TFB2, TFIIH
UniGene
Hs.485070
Gene Chromosome Location
Chr.6: 30908200-30914103 on Build GRCh38
Transcripts
NM_001517.4

SNP Detail

SNP ID Chromosome Location
rs1006316243 Chr.6: 30908202-30908202 on Build GRCh38
rs3218809 Chr.6: 30908414-30908414 on Build GRCh38
rs929300908 Chr.6: 30908360-30908360 on Build GRCh38
rs766871441 Chr.6: 30908169-30908169 on Build GRCh38
rs959959211 Chr.6: 30908260-30908260 on Build GRCh38
rs116013990 Chr.6: 30908447-30908447 on Build GRCh38
rs3218808 Chr.6: 30908240-30908240 on Build GRCh38
rs937819647 Chr.6: 30908548-30908548 on Build GRCh38
rs1264309 Chr.6: 30908122-30908122 on Build GRCh38
rs765728270 Chr.6: 30908102-30908102 on Build GRCh38
rs1001940463 Chr.6: 30908204-30908204 on Build GRCh38
rs867838826 Chr.6: 30908097-30908097 on Build GRCh38
rs889494812 Chr.6: 30908119-30908119 on Build GRCh38
rs913067953 Chr.6: 30908266-30908266 on Build GRCh38
rs1017795295 Chr.6: 30908268-30908268 on Build GRCh38
rs1053468223 Chr.6: 30908558-30908558 on Build GRCh38
rs1032024936 Chr.6: 30908210-30908210 on Build GRCh38
rs763350125 Chr.6: 30908123-30908123 on Build GRCh38
rs1021631353 Chr.6: 30908243-30908243 on Build GRCh38
rs546906242 Chr.6: 30908464-30908464 on Build GRCh38
rs910943255 Chr.6: 30908299-30908299 on Build GRCh38
rs950906595 Chr.6: 30908190-30908190 on Build GRCh38
rs2074509 Chr.6: 30908461-30908461 on Build GRCh38
rs544144076 Chr.6: 30908132-30908132 on Build GRCh38
rs373441040 Chr.6: 30908536-30908536 on Build GRCh38
rs1042851408 Chr.6: 30908292-30908292 on Build GRCh38
rs188491108 Chr.6: 30908211-30908211 on Build GRCh38
rs191842165 Chr.6: 30908281-30908281 on Build GRCh38
rs920639027 Chr.6: 30908348-30908348 on Build GRCh38
rs1047425640 Chr.6: 30908107-30908107 on Build GRCh38
rs942397486 Chr.6: 30908320-30908320 on Build GRCh38
rs997692681 Chr.6: 30908160-30908160 on Build GRCh38
rs147236117 Chr.6: 30908426-30908426 on Build GRCh38
rs949320576 Chr.6: 30908121-30908121 on Build GRCh38
rs2074510 Chr.6: 30908257-30908257 on Build GRCh38
rs964861573 Chr.6: 30908273-30908273 on Build GRCh38
rs886997518 Chr.6: 30908112-30908112 on Build GRCh38
rs779229286 Chr.6: 30908441-30908442 on Build GRCh38
rs1048409588 Chr.6: 30908440-30908441 on Build GRCh38
rs1026499307 Chr.6: 30908165-30908165 on Build GRCh38
rs973503080 Chr.6: 30908335-30908335 on Build GRCh38
rs369535959 Chr.6: 30908129-30908129 on Build GRCh38
rs997723739 Chr.6: 30908427-30908427 on Build GRCh38
rs143868324 Chr.6: 30908262-30908262 on Build GRCh38
rs920622640 Chr.6: 30908096-30908096 on Build GRCh38
rs1052693 Chr.6: 30908375-30908375 on Build GRCh38
rs560849645 Chr.6: 30908189-30908189 on Build GRCh38
rs970088195 Chr.6: 30908228-30908228 on Build GRCh38
rs138184407 Chr.6: 30908225-30908225 on Build GRCh38
rs905049282 Chr.6: 30908476-30908476 on Build GRCh38

View Full Product Details