Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111259201-111259710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD4B
Entrez Gene ID
91749
Gene Name
major facilitator superfamily domain containing 4B
Gene Aliases
KIAA1919, NaGLT1
UniGene
Hs.400572
Gene Chromosome Location
Chr.6: 111259279-111280005 on Build GRCh38
Transcripts
NM_153369.2 XM_017011483.1

SNP Detail

SNP ID Chromosome Location
rs979356586 Chr.6: 111259396-111259396 on Build GRCh38
rs919491796 Chr.6: 111259444-111259444 on Build GRCh38
rs527303499 Chr.6: 111259598-111259598 on Build GRCh38
rs937092102 Chr.6: 111259527-111259527 on Build GRCh38
rs928446024 Chr.6: 111259502-111259502 on Build GRCh38
rs1011573688 Chr.6: 111259258-111259258 on Build GRCh38
rs550959860 Chr.6: 111259278-111259278 on Build GRCh38
rs981508404 Chr.6: 111259501-111259501 on Build GRCh38
rs947197108 Chr.6: 111259555-111259555 on Build GRCh38
rs547719742 Chr.6: 111259538-111259538 on Build GRCh38
rs963510501 Chr.6: 111259415-111259415 on Build GRCh38
rs1001759060 Chr.6: 111259234-111259234 on Build GRCh38
rs932002793 Chr.6: 111259283-111259283 on Build GRCh38
rs979352301 Chr.6: 111259621-111259621 on Build GRCh38
rs539339317 Chr.6: 111259317-111259317 on Build GRCh38
rs942255669 Chr.6: 111259337-111259337 on Build GRCh38
rs1052195973 Chr.6: 111259604-111259604 on Build GRCh38
rs1030585202 Chr.6: 111259334-111259353 on Build GRCh38
rs773575556 Chr.6: 111259366-111259366 on Build GRCh38
rs903959576 Chr.6: 111259292-111259292 on Build GRCh38
rs772760986 Chr.6: 111259652-111259652 on Build GRCh38
rs765808119 Chr.6: 111259663-111259663 on Build GRCh38
rs17072492 Chr.6: 111259363-111259363 on Build GRCh38
rs773152947 Chr.6: 111259237-111259237 on Build GRCh38
rs182092932 Chr.6: 111259348-111259348 on Build GRCh38
rs11153277 Chr.6: 111259358-111259358 on Build GRCh38
rs966151659 Chr.6: 111259567-111259567 on Build GRCh38
rs549022366 Chr.6: 111259626-111259626 on Build GRCh38
rs953226967 Chr.6: 111259583-111259583 on Build GRCh38
rs532722453 Chr.6: 111259270-111259270 on Build GRCh38
rs868620938 Chr.6: 111259443-111259443 on Build GRCh38
rs1034260541 Chr.6: 111259235-111259235 on Build GRCh38
rs1007661281 Chr.6: 111259613-111259613 on Build GRCh38
rs921842079 Chr.6: 111259282-111259282 on Build GRCh38
rs917963717 Chr.6: 111259412-111259412 on Build GRCh38
rs1014170471 Chr.6: 111259333-111259333 on Build GRCh38
rs1055552311 Chr.6: 111259368-111259368 on Build GRCh38
rs918269381 Chr.6: 111259349-111259349 on Build GRCh38
rs780234211 Chr.6: 111259274-111259274 on Build GRCh38
rs892934474 Chr.6: 111259369-111259369 on Build GRCh38
rs545360099 Chr.6: 111259370-111259370 on Build GRCh38
rs996183200 Chr.6: 111259447-111259447 on Build GRCh38
rs567252174 Chr.6: 111259650-111259650 on Build GRCh38
rs953594952 Chr.6: 111259327-111259327 on Build GRCh38
rs567419881 Chr.6: 111259329-111259329 on Build GRCh38
rs909497375 Chr.6: 111259332-111259332 on Build GRCh38
rs997559926 Chr.6: 111259572-111259572 on Build GRCh38
rs553485 Chr.6: 111259531-111259531 on Build GRCh38
rs893048352 Chr.6: 111259243-111259243 on Build GRCh38
rs565186874 Chr.6: 111259442-111259442 on Build GRCh38
rs531895654 Chr.6: 111259399-111259399 on Build GRCh38
rs189833053 Chr.6: 111259429-111259429 on Build GRCh38
rs566386705 Chr.6: 111259285-111259285 on Build GRCh38
rs143896125 Chr.6: 111259372-111259372 on Build GRCh38
rs976204329 Chr.6: 111259301-111259301 on Build GRCh38
rs1009794606 Chr.6: 111259525-111259525 on Build GRCh38
rs929608550 Chr.6: 111259353-111259353 on Build GRCh38
rs1020242545 Chr.6: 111259275-111259275 on Build GRCh38
rs111515930 Chr.6: 111259400-111259400 on Build GRCh38
rs956710388 Chr.6: 111259643-111259643 on Build GRCh38
rs773975443 Chr.6: 111259685-111259685 on Build GRCh38
rs1048662629 Chr.6: 111259493-111259493 on Build GRCh38
rs571618543 Chr.6: 111259233-111259233 on Build GRCh38
rs561243236 Chr.6: 111259389-111259389 on Build GRCh38
rs748889824 Chr.6: 111259379-111259379 on Build GRCh38
rs951110957 Chr.6: 111259457-111259457 on Build GRCh38
rs150662869 Chr.6: 111259328-111259328 on Build GRCh38
rs925200917 Chr.6: 111259636-111259637 on Build GRCh38
rs747096051 Chr.6: 111259310-111259310 on Build GRCh38
rs953579431 Chr.6: 111259395-111259395 on Build GRCh38
rs976280246 Chr.6: 111259386-111259386 on Build GRCh38
rs935443093 Chr.6: 111259312-111259312 on Build GRCh38
rs751357220 Chr.6: 111259561-111259561 on Build GRCh38
rs78273629 Chr.6: 111259387-111259387 on Build GRCh38
rs889082711 Chr.6: 111259612-111259612 on Build GRCh38
rs905464181 Chr.6: 111259436-111259437 on Build GRCh38
rs988070395 Chr.6: 111259662-111259662 on Build GRCh38
rs1052508506 Chr.6: 111259407-111259407 on Build GRCh38
rs572374004 Chr.6: 111259377-111259377 on Build GRCh38
rs906935211 Chr.6: 111259361-111259361 on Build GRCh38
rs905411195 Chr.6: 111259341-111259341 on Build GRCh38
rs113185274 Chr.6: 111259568-111259568 on Build GRCh38
rs1045614659 Chr.6: 111259422-111259422 on Build GRCh38

View Full Product Details