Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111267395-111267938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD4B
Entrez Gene ID
91749
Gene Name
major facilitator superfamily domain containing 4B
Gene Aliases
KIAA1919, NaGLT1
UniGene
Hs.400572
Gene Chromosome Location
Chr.6: 111259279-111280005 on Build GRCh38
Transcripts
NM_153369.2 XM_017011483.1

SNP Detail

SNP ID Chromosome Location
rs763473823 Chr.6: 111267566-111267566 on Build GRCh38
rs916719530 Chr.6: 111267779-111267779 on Build GRCh38
rs1038988871 Chr.6: 111267665-111267665 on Build GRCh38
rs957908580 Chr.6: 111267456-111267456 on Build GRCh38
rs982029129 Chr.6: 111267860-111267860 on Build GRCh38
rs1052836892 Chr.6: 111267617-111267617 on Build GRCh38
rs565969987 Chr.6: 111267816-111267816 on Build GRCh38
rs948454537 Chr.6: 111267551-111267551 on Build GRCh38
rs960895702 Chr.6: 111267751-111267751 on Build GRCh38
rs994731447 Chr.6: 111267690-111267690 on Build GRCh38
rs771605731 Chr.6: 111267896-111267896 on Build GRCh38
rs1005616112 Chr.6: 111267601-111267601 on Build GRCh38
rs572024421 Chr.6: 111267437-111267437 on Build GRCh38
rs900418872 Chr.6: 111267681-111267681 on Build GRCh38
rs376894124 Chr.6: 111267791-111267791 on Build GRCh38
rs529983697 Chr.6: 111267837-111267837 on Build GRCh38
rs980684662 Chr.6: 111267875-111267875 on Build GRCh38
rs911639496 Chr.6: 111267893-111267893 on Build GRCh38
rs532354465 Chr.6: 111267687-111267687 on Build GRCh38
rs1017374427 Chr.6: 111267814-111267814 on Build GRCh38
rs1046340342 Chr.6: 111267530-111267530 on Build GRCh38
rs146356862 Chr.6: 111267658-111267658 on Build GRCh38
rs1055223461 Chr.6: 111267564-111267564 on Build GRCh38
rs560904236 Chr.6: 111267492-111267492 on Build GRCh38
rs548168261 Chr.6: 111267840-111267840 on Build GRCh38
rs376895752 Chr.6: 111267501-111267501 on Build GRCh38
rs569750677 Chr.6: 111267901-111267901 on Build GRCh38
rs960843503 Chr.6: 111267525-111267525 on Build GRCh38
rs1028085204 Chr.6: 111267694-111267694 on Build GRCh38
rs890319615 Chr.6: 111267648-111267648 on Build GRCh38
rs547336648 Chr.6: 111267815-111267815 on Build GRCh38
rs1035055066 Chr.6: 111267746-111267746 on Build GRCh38
rs938007988 Chr.6: 111267872-111267872 on Build GRCh38
rs773183450 Chr.6: 111267544-111267544 on Build GRCh38
rs916811452 Chr.6: 111267547-111267547 on Build GRCh38
rs1007149376 Chr.6: 111267812-111267812 on Build GRCh38
rs183873867 Chr.6: 111267543-111267543 on Build GRCh38
rs950697791 Chr.6: 111267736-111267736 on Build GRCh38
rs968718452 Chr.6: 111267786-111267786 on Build GRCh38
rs770108245 Chr.6: 111267634-111267635 on Build GRCh38
rs75135648 Chr.6: 111267495-111267495 on Build GRCh38
rs989233540 Chr.6: 111267836-111267836 on Build GRCh38
rs542639053 Chr.6: 111267475-111267475 on Build GRCh38
rs897312274 Chr.6: 111267682-111267684 on Build GRCh38
rs1043122571 Chr.6: 111267580-111267580 on Build GRCh38
rs998431100 Chr.6: 111267689-111267689 on Build GRCh38
rs566866607 Chr.6: 111267745-111267745 on Build GRCh38
rs1055558346 Chr.6: 111267879-111267879 on Build GRCh38
rs944430964 Chr.6: 111267894-111267894 on Build GRCh38
rs991394145 Chr.6: 111267769-111267769 on Build GRCh38
rs989697215 Chr.6: 111267485-111267485 on Build GRCh38
rs527509699 Chr.6: 111267462-111267462 on Build GRCh38
rs934415119 Chr.6: 111267593-111267593 on Build GRCh38
rs979683666 Chr.6: 111267790-111267790 on Build GRCh38
rs945305521 Chr.6: 111267511-111267513 on Build GRCh38
rs1052887706 Chr.6: 111267868-111267868 on Build GRCh38
rs927990853 Chr.6: 111267539-111267539 on Build GRCh38
rs543568595 Chr.6: 111267592-111267592 on Build GRCh38

View Full Product Details