Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111268528-111269059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD4B
Entrez Gene ID
91749
Gene Name
major facilitator superfamily domain containing 4B
Gene Aliases
KIAA1919, NaGLT1
UniGene
Hs.400572
Gene Chromosome Location
Chr.6: 111259279-111280005 on Build GRCh38
Transcripts
NM_153369.2 XM_017011483.1

SNP Detail

SNP ID Chromosome Location
rs200022313 Chr.6: 111268625-111268626 on Build GRCh38
rs986851530 Chr.6: 111268781-111268781 on Build GRCh38
rs946394477 Chr.6: 111268619-111268619 on Build GRCh38
rs966135119 Chr.6: 111268732-111268732 on Build GRCh38
rs892668598 Chr.6: 111269031-111269031 on Build GRCh38
rs557703995 Chr.6: 111268655-111268655 on Build GRCh38
rs936758555 Chr.6: 111268974-111268974 on Build GRCh38
rs942845997 Chr.6: 111268584-111268584 on Build GRCh38
rs1052502473 Chr.6: 111269008-111269008 on Build GRCh38
rs903937828 Chr.6: 111268659-111268659 on Build GRCh38
rs948071567 Chr.6: 111268638-111268638 on Build GRCh38
rs866383371 Chr.6: 111268592-111268592 on Build GRCh38
rs190978557 Chr.6: 111268808-111268808 on Build GRCh38
rs240987 Chr.6: 111268552-111268552 on Build GRCh38
rs240988 Chr.6: 111268891-111268891 on Build GRCh38
rs79074797 Chr.6: 111268561-111268561 on Build GRCh38
rs892616718 Chr.6: 111268695-111268695 on Build GRCh38
rs938965025 Chr.6: 111268939-111268939 on Build GRCh38
rs995856792 Chr.6: 111268734-111268734 on Build GRCh38
rs536125560 Chr.6: 111268608-111268608 on Build GRCh38
rs1012050221 Chr.6: 111268646-111268646 on Build GRCh38
rs1000916529 Chr.6: 111268661-111268661 on Build GRCh38
rs748350012 Chr.6: 111268587-111268587 on Build GRCh38
rs574524861 Chr.6: 111268673-111268673 on Build GRCh38
rs947925467 Chr.6: 111268906-111268906 on Build GRCh38
rs946352748 Chr.6: 111268869-111268869 on Build GRCh38
rs893360171 Chr.6: 111268617-111268617 on Build GRCh38
rs962679483 Chr.6: 111268714-111268714 on Build GRCh38
rs1017281323 Chr.6: 111268675-111268675 on Build GRCh38
rs781564015 Chr.6: 111268912-111268912 on Build GRCh38
rs568697257 Chr.6: 111268590-111268590 on Build GRCh38
rs142489535 Chr.6: 111268786-111268786 on Build GRCh38
rs1008508279 Chr.6: 111268724-111268724 on Build GRCh38
rs1056291559 Chr.6: 111268625-111268625 on Build GRCh38
rs955374270 Chr.6: 111268748-111268748 on Build GRCh38
rs925244874 Chr.6: 111268961-111268961 on Build GRCh38
rs758258808 Chr.6: 111268555-111268555 on Build GRCh38
rs556128840 Chr.6: 111268644-111268645 on Build GRCh38
rs760724717 Chr.6: 111269026-111269027 on Build GRCh38
rs374150627 Chr.6: 111268626-111268626 on Build GRCh38
rs1045080057 Chr.6: 111268956-111268956 on Build GRCh38
rs1041987844 Chr.6: 111268883-111268883 on Build GRCh38
rs749376046 Chr.6: 111269022-111269022 on Build GRCh38
rs990311702 Chr.6: 111268844-111268844 on Build GRCh38
rs542041677 Chr.6: 111268846-111268846 on Build GRCh38
rs992069113 Chr.6: 111268867-111268867 on Build GRCh38
rs964573367 Chr.6: 111268840-111268840 on Build GRCh38
rs1019852465 Chr.6: 111268727-111268727 on Build GRCh38
rs79850296 Chr.6: 111268702-111268702 on Build GRCh38
rs984481252 Chr.6: 111268792-111268792 on Build GRCh38
rs1029039101 Chr.6: 111268736-111268736 on Build GRCh38
rs994480212 Chr.6: 111268717-111268717 on Build GRCh38

View Full Product Details