Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16087208-16087720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISPD
Entrez Gene ID
729920
Gene Name
isoprenoid synthase domain containing
Gene Aliases
MDDGA7, MDDGC7, Nip, hCG_1745121
UniGene
Hs.636502
Gene Chromosome Location
Chr.7: 16087527-16422562 on Build GRCh38
Transcripts
NM_001101417.3 NM_001101426.3

SNP Detail

SNP ID Chromosome Location
rs1000282529 Chr.7: 16087472-16087472 on Build GRCh38
rs201991429 Chr.7: 16087330-16087331 on Build GRCh38
rs527811569 Chr.7: 16087473-16087473 on Build GRCh38
rs565491860 Chr.7: 16087588-16087592 on Build GRCh38
rs1010337240 Chr.7: 16087502-16087502 on Build GRCh38
rs1033162702 Chr.7: 16087278-16087279 on Build GRCh38
rs200292569 Chr.7: 16087354-16087355 on Build GRCh38
rs12539174 Chr.7: 16087672-16087672 on Build GRCh38
rs886062138 Chr.7: 16087579-16087579 on Build GRCh38
rs188823385 Chr.7: 16087603-16087603 on Build GRCh38
rs923090552 Chr.7: 16087644-16087644 on Build GRCh38
rs923215799 Chr.7: 16087397-16087397 on Build GRCh38
rs974097804 Chr.7: 16087688-16087688 on Build GRCh38
rs34543368 Chr.7: 16087260-16087261 on Build GRCh38
rs951209905 Chr.7: 16087623-16087623 on Build GRCh38
rs967812072 Chr.7: 16087359-16087359 on Build GRCh38
rs553353474 Chr.7: 16087590-16087590 on Build GRCh38
rs551547475 Chr.7: 16087382-16087382 on Build GRCh38
rs909099473 Chr.7: 16087673-16087673 on Build GRCh38
rs994312948 Chr.7: 16087277-16087277 on Build GRCh38
rs931887418 Chr.7: 16087418-16087418 on Build GRCh38
rs994782027 Chr.7: 16087506-16087506 on Build GRCh38
rs775071114 Chr.7: 16087585-16087585 on Build GRCh38
rs979765101 Chr.7: 16087334-16087334 on Build GRCh38
rs1056626019 Chr.7: 16087507-16087507 on Build GRCh38
rs116899576 Chr.7: 16087363-16087363 on Build GRCh38
rs911240540 Chr.7: 16087433-16087433 on Build GRCh38
rs762312522 Chr.7: 16087636-16087636 on Build GRCh38
rs563239790 Chr.7: 16087342-16087342 on Build GRCh38
rs912510993 Chr.7: 16087650-16087650 on Build GRCh38
rs891967966 Chr.7: 16087482-16087482 on Build GRCh38
rs971609592 Chr.7: 16087506-16087506 on Build GRCh38
rs886062139 Chr.7: 16087685-16087685 on Build GRCh38
rs1038431191 Chr.7: 16087252-16087252 on Build GRCh38
rs573114796 Chr.7: 16087626-16087626 on Build GRCh38
rs71007741 Chr.7: 16087274-16087275 on Build GRCh38
rs1000799069 Chr.7: 16087277-16087278 on Build GRCh38
rs956750521 Chr.7: 16087381-16087381 on Build GRCh38
rs192924503 Chr.7: 16087695-16087695 on Build GRCh38
rs17169260 Chr.7: 16087465-16087465 on Build GRCh38
rs539238298 Chr.7: 16087678-16087678 on Build GRCh38
rs1035682980 Chr.7: 16087321-16087321 on Build GRCh38
rs557404340 Chr.7: 16087687-16087687 on Build GRCh38
rs369313549 Chr.7: 16087306-16087307 on Build GRCh38
rs1039021463 Chr.7: 16087441-16087441 on Build GRCh38
rs897328664 Chr.7: 16087258-16087258 on Build GRCh38
rs550979092 Chr.7: 16087305-16087306 on Build GRCh38
rs1049741222 Chr.7: 16087428-16087428 on Build GRCh38
rs369915144 Chr.7: 16087563-16087563 on Build GRCh38
rs184324561 Chr.7: 16087536-16087536 on Build GRCh38
rs1023290434 Chr.7: 16087344-16087344 on Build GRCh38
rs57652322 Chr.7: 16087377-16087378 on Build GRCh38
rs145625852 Chr.7: 16087665-16087665 on Build GRCh38
rs960944325 Chr.7: 16087332-16087332 on Build GRCh38
rs929759294 Chr.7: 16087379-16087379 on Build GRCh38
rs976350662 Chr.7: 16087634-16087634 on Build GRCh38
rs386409589 Chr.7: 16087264-16087265 on Build GRCh38
rs10242711 Chr.7: 16087365-16087365 on Build GRCh38
rs1023867663 Chr.7: 16087571-16087571 on Build GRCh38
rs67347055 Chr.7: 16087261-16087262 on Build GRCh38
rs941337091 Chr.7: 16087438-16087438 on Build GRCh38
rs912458633 Chr.7: 16087389-16087389 on Build GRCh38
rs548902504 Chr.7: 16087505-16087506 on Build GRCh38
rs186948323 Chr.7: 16087298-16087298 on Build GRCh38

View Full Product Details